Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9628644-9629144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP52
Entrez Gene ID
146845
Gene Name
cilia and flagella associated protein 52
Gene Aliases
WDR16, WDRPUH
UniGene
Hs.232270
Gene Chromosome Location
Chr.17: 9576627-9643459 on Build GRCh38
Transcripts
NM_001080556.1 NM_145054.4 XM_017024227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4649936 c.1063G>A p.A355T Chr.17: 9628679-9628679 on Build GRCh38
7222854 c.1144G>T p.V382L Chr.17: 9628760-9628760 on Build GRCh38
7222853 c.1114G>T p.V372L Chr.17: 9628760-9628760 on Build GRCh38
4599078 c.1062G>T p.K354N Chr.17: 9628708-9628708 on Build GRCh38
4592540 c.1143C>A p.D381E Chr.17: 9628789-9628789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374787564 Chr.17: 9629052-9629052 on Build GRCh38
rs746932861 Chr.17: 9628738-9628738 on Build GRCh38
rs904539123 Chr.17: 9629017-9629017 on Build GRCh38
rs139863455 Chr.17: 9628974-9628974 on Build GRCh38
rs367791419 Chr.17: 9628717-9628717 on Build GRCh38
rs776460331 Chr.17: 9628751-9628751 on Build GRCh38
rs374339508 Chr.17: 9628787-9628787 on Build GRCh38
rs768387591 Chr.17: 9628745-9628745 on Build GRCh38
rs765507258 Chr.17: 9628864-9628864 on Build GRCh38
rs139936399 Chr.17: 9628918-9628918 on Build GRCh38
rs767576439 Chr.17: 9628759-9628759 on Build GRCh38
rs372924224 Chr.17: 9628825-9628825 on Build GRCh38
rs774619043 Chr.17: 9628826-9628826 on Build GRCh38
rs199988393 Chr.17: 9628761-9628761 on Build GRCh38
rs369770347 Chr.17: 9628786-9628786 on Build GRCh38
rs369133428 Chr.17: 9629091-9629091 on Build GRCh38
rs74424345 Chr.17: 9629101-9629101 on Build GRCh38
rs771479704 Chr.17: 9628882-9628882 on Build GRCh38
rs780939657 Chr.17: 9628802-9628802 on Build GRCh38
rs145951373 Chr.17: 9628801-9628801 on Build GRCh38
rs775516550 Chr.17: 9628847-9628847 on Build GRCh38
rs141229371 Chr.17: 9628677-9628677 on Build GRCh38
rs57960962 Chr.17: 9628711-9628711 on Build GRCh38
rs772186858 Chr.17: 9628841-9628841 on Build GRCh38
rs368229972 Chr.17: 9628916-9628916 on Build GRCh38
rs747984041 Chr.17: 9628809-9628809 on Build GRCh38
rs779903938 Chr.17: 9628805-9628805 on Build GRCh38
rs978073936 Chr.17: 9628876-9628876 on Build GRCh38
rs1044372568 Chr.17: 9629000-9629000 on Build GRCh38
rs189850829 Chr.17: 9629059-9629059 on Build GRCh38
rs150503527 Chr.17: 9628760-9628760 on Build GRCh38
rs758095633 Chr.17: 9628871-9628871 on Build GRCh38
rs948707016 Chr.17: 9628999-9628999 on Build GRCh38
rs775006460 Chr.17: 9628939-9628939 on Build GRCh38
rs141646758 Chr.17: 9628752-9628752 on Build GRCh38
rs746958003 Chr.17: 9628827-9628828 on Build GRCh38
rs895789657 Chr.17: 9628750-9628750 on Build GRCh38
rs776935857 Chr.17: 9628862-9628862 on Build GRCh38
rs756901981 Chr.17: 9628696-9628696 on Build GRCh38
rs943439397 Chr.17: 9628977-9628977 on Build GRCh38
rs777114820 Chr.17: 9628908-9628908 on Build GRCh38
rs201932551 Chr.17: 9628869-9628869 on Build GRCh38
rs757506984 Chr.17: 9628780-9628780 on Build GRCh38
rs761146293 Chr.17: 9628767-9628767 on Build GRCh38
rs369013852 Chr.17: 9628854-9628854 on Build GRCh38
rs1037780598 Chr.17: 9629039-9629039 on Build GRCh38
rs916538910 Chr.17: 9628985-9628985 on Build GRCh38
rs1052241941 Chr.17: 9628704-9628704 on Build GRCh38
rs371698292 Chr.17: 9628755-9628755 on Build GRCh38
rs1003594075 Chr.17: 9629119-9629119 on Build GRCh38
rs1053619531 Chr.17: 9629070-9629070 on Build GRCh38
rs778381409 Chr.17: 9628702-9628702 on Build GRCh38
rs760630676 Chr.17: 9628852-9628852 on Build GRCh38
rs746074979 Chr.17: 9628832-9628832 on Build GRCh38
rs374648753 Chr.17: 9628674-9628674 on Build GRCh38
rs113576151 Chr.17: 9628693-9628693 on Build GRCh38
rs377115961 Chr.17: 9628827-9628827 on Build GRCh38
rs764406343 Chr.17: 9628768-9628768 on Build GRCh38
rs920396220 Chr.17: 9628673-9628673 on Build GRCh38
rs780209016 Chr.17: 9628727-9628727 on Build GRCh38
rs75671790 Chr.17: 9628669-9628669 on Build GRCh38
rs754259989 Chr.17: 9628772-9628772 on Build GRCh38
rs920326419 Chr.17: 9628914-9628914 on Build GRCh38
rs891961033 Chr.17: 9629071-9629071 on Build GRCh38
rs117569464 Chr.17: 9628789-9628789 on Build GRCh38
rs904633446 Chr.17: 9628771-9628771 on Build GRCh38
rs920421832 Chr.17: 9629045-9629045 on Build GRCh38
rs77481032 Chr.17: 9628941-9628941 on Build GRCh38
rs987501971 Chr.17: 9628944-9628944 on Build GRCh38
rs889213613 Chr.17: 9628710-9628710 on Build GRCh38
rs746987913 Chr.17: 9628790-9628790 on Build GRCh38
rs764947992 Chr.17: 9628781-9628781 on Build GRCh38
rs774494211 Chr.17: 9628753-9628753 on Build GRCh38
rs762307224 Chr.17: 9628863-9628863 on Build GRCh38
rs1000244715 Chr.17: 9629066-9629066 on Build GRCh38
rs906460390 Chr.17: 9629107-9629107 on Build GRCh38
rs867819243 Chr.17: 9628708-9628708 on Build GRCh38

View Full Product Details