Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63787083-63787546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX42
Entrez Gene ID
11325
Gene Name
DEAD-box helicase 42
Gene Aliases
DDX42P, RHELP, RNAHP, SF3B8, SF3b125
UniGene
Hs.702010
Gene Chromosome Location
Chr.17: 63773856-63819317 on Build GRCh38
Transcripts
NM_007372.3 NM_203499.2 XM_006721657.1 XM_017024111.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203263 c.221+1G>A p.? Chr.17: 63787271-63787271 on Build GRCh38
6324304 c.70G>A p.G24R Chr.17: 63787119-63787119 on Build GRCh38
6081777 c.181A>C p.K61Q Chr.17: 63787230-63787230 on Build GRCh38
229308 c.124A>T p.T42S Chr.17: 63787173-63787173 on Build GRCh38
3820207 c.152C>G p.S51* Chr.17: 63787201-63787201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022168928 Chr.17: 63787485-63787485 on Build GRCh38
rs904619012 Chr.17: 63787502-63787502 on Build GRCh38
rs76225333 Chr.17: 63787150-63787150 on Build GRCh38
rs1001466188 Chr.17: 63787515-63787515 on Build GRCh38
rs759608225 Chr.17: 63787238-63787238 on Build GRCh38
rs980424986 Chr.17: 63787497-63787497 on Build GRCh38
rs545231981 Chr.17: 63787519-63787519 on Build GRCh38
rs369269240 Chr.17: 63787285-63787285 on Build GRCh38
rs1010406574 Chr.17: 63787476-63787476 on Build GRCh38
rs776428033 Chr.17: 63787455-63787455 on Build GRCh38
rs765072563 Chr.17: 63787296-63787296 on Build GRCh38
rs767718123 Chr.17: 63787106-63787106 on Build GRCh38
rs753595532 Chr.17: 63787109-63787109 on Build GRCh38
rs9747164 Chr.17: 63787427-63787427 on Build GRCh38
rs760639538 Chr.17: 63787352-63787352 on Build GRCh38
rs901946712 Chr.17: 63787368-63787368 on Build GRCh38
rs143333200 Chr.17: 63787206-63787206 on Build GRCh38
rs746463979 Chr.17: 63787156-63787156 on Build GRCh38
rs773702399 Chr.17: 63787202-63787202 on Build GRCh38
rs370838961 Chr.17: 63787320-63787320 on Build GRCh38
rs556556496 Chr.17: 63787393-63787393 on Build GRCh38
rs770246027 Chr.17: 63787195-63787195 on Build GRCh38
rs1049360304 Chr.17: 63787220-63787220 on Build GRCh38
rs1007571328 Chr.17: 63787366-63787366 on Build GRCh38
rs927659124 Chr.17: 63787518-63787518 on Build GRCh38
rs866118153 Chr.17: 63787248-63787248 on Build GRCh38
rs930883779 Chr.17: 63787211-63787211 on Build GRCh38
rs776502994 Chr.17: 63787146-63787146 on Build GRCh38
rs190569526 Chr.17: 63787436-63787436 on Build GRCh38
rs747726846 Chr.17: 63787175-63787175 on Build GRCh38
rs149040414 Chr.17: 63787451-63787451 on Build GRCh38
rs757034774 Chr.17: 63787113-63787113 on Build GRCh38
rs201879734 Chr.17: 63787121-63787121 on Build GRCh38
rs558165093 Chr.17: 63787145-63787145 on Build GRCh38
rs767864186 Chr.17: 63787179-63787181 on Build GRCh38
rs775377895 Chr.17: 63787326-63787326 on Build GRCh38
rs767661453 Chr.17: 63787245-63787245 on Build GRCh38
rs919462027 Chr.17: 63787204-63787204 on Build GRCh38
rs933541552 Chr.17: 63787350-63787350 on Build GRCh38
rs139915610 Chr.17: 63787194-63787194 on Build GRCh38
rs73331985 Chr.17: 63787496-63787496 on Build GRCh38
rs1031236989 Chr.17: 63787394-63787394 on Build GRCh38
rs562648904 Chr.17: 63787143-63787143 on Build GRCh38
rs998973019 Chr.17: 63787370-63787370 on Build GRCh38
rs754610104 Chr.17: 63787167-63787167 on Build GRCh38
rs371458465 Chr.17: 63787290-63787290 on Build GRCh38
rs201845056 Chr.17: 63787260-63787260 on Build GRCh38
rs1040452932 Chr.17: 63787367-63787367 on Build GRCh38
rs758317871 Chr.17: 63787137-63787137 on Build GRCh38
rs200639700 Chr.17: 63787203-63787203 on Build GRCh38
rs146726768 Chr.17: 63787221-63787221 on Build GRCh38
rs774581698 Chr.17: 63787233-63787233 on Build GRCh38
rs750408943 Chr.17: 63787132-63787132 on Build GRCh38
rs780858678 Chr.17: 63787170-63787170 on Build GRCh38

View Full Product Details