Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73252538-73253034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF4L
Entrez Gene ID
642843
Gene Name
cleavage and polyadenylation specific factor 4 like
Gene Aliases
-
UniGene
Hs.534707
Gene Chromosome Location
Chr.17: 73242343-73263778 on Build GRCh38
Transcripts
NM_001129885.1 XM_011525115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1385689 c.222G>A p.L74L Chr.17: 73252716-73252716 on Build GRCh38
4496231 c.281C>T p.P94L Chr.17: 73252657-73252657 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs76140197 Chr.17: 73252581-73252581 on Build GRCh38
rs909031991 Chr.17: 73252707-73252707 on Build GRCh38
rs774414124 Chr.17: 73252822-73252822 on Build GRCh38
rs1037605026 Chr.17: 73252865-73252865 on Build GRCh38
rs189860170 Chr.17: 73252989-73252989 on Build GRCh38
rs537583823 Chr.17: 73252999-73252999 on Build GRCh38
rs377495206 Chr.17: 73252949-73252949 on Build GRCh38
rs866142978 Chr.17: 73252700-73252700 on Build GRCh38
rs943037407 Chr.17: 73252783-73252783 on Build GRCh38
rs199550226 Chr.17: 73253005-73253005 on Build GRCh38
rs1033513916 Chr.17: 73252619-73252621 on Build GRCh38
rs869085637 Chr.17: 73253004-73253005 on Build GRCh38
rs5821956 Chr.17: 73253002-73253003 on Build GRCh38
rs775610275 Chr.17: 73252608-73252608 on Build GRCh38
rs1046031284 Chr.17: 73252790-73252790 on Build GRCh38
rs570065024 Chr.17: 73252992-73252992 on Build GRCh38
rs1012766431 Chr.17: 73252660-73252660 on Build GRCh38
rs975986869 Chr.17: 73252694-73252694 on Build GRCh38
rs541343168 Chr.17: 73252705-73252705 on Build GRCh38
rs903363454 Chr.17: 73252605-73252605 on Build GRCh38
rs529881132 Chr.17: 73252738-73252738 on Build GRCh38
rs377495286 Chr.17: 73252739-73252739 on Build GRCh38
rs397772914 Chr.17: 73253003-73253004 on Build GRCh38
rs763701465 Chr.17: 73252768-73252768 on Build GRCh38
rs1047561387 Chr.17: 73252835-73252835 on Build GRCh38
rs985111534 Chr.17: 73252932-73252932 on Build GRCh38
rs752417718 Chr.17: 73252682-73252682 on Build GRCh38
rs35498916 Chr.17: 73252662-73252663 on Build GRCh38
rs965997525 Chr.17: 73252686-73252686 on Build GRCh38
rs563575551 Chr.17: 73252815-73252815 on Build GRCh38
rs371092902 Chr.17: 73252758-73252758 on Build GRCh38
rs558875854 Chr.17: 73252594-73252594 on Build GRCh38
rs1008046529 Chr.17: 73252642-73252642 on Build GRCh38
rs8068398 Chr.17: 73252891-73252891 on Build GRCh38
rs1006058728 Chr.17: 73252617-73252617 on Build GRCh38
rs895145246 Chr.17: 73252655-73252655 on Build GRCh38
rs1054661445 Chr.17: 73252829-73252829 on Build GRCh38
rs374678176 Chr.17: 73252810-73252810 on Build GRCh38
rs151195750 Chr.17: 73252706-73252706 on Build GRCh38
rs192640053 Chr.17: 73252837-73252837 on Build GRCh38
rs5821955 Chr.17: 73252991-73252992 on Build GRCh38
rs373301369 Chr.17: 73252946-73252946 on Build GRCh38
rs763239761 Chr.17: 73252703-73252703 on Build GRCh38
rs373299697 Chr.17: 73252693-73252693 on Build GRCh38
rs970772093 Chr.17: 73252699-73252699 on Build GRCh38
rs906626078 Chr.17: 73252975-73252975 on Build GRCh38
rs369879623 Chr.17: 73252855-73252855 on Build GRCh38
rs909070124 Chr.17: 73252933-73252933 on Build GRCh38
rs933168806 Chr.17: 73252589-73252589 on Build GRCh38
rs759993609 Chr.17: 73252726-73252726 on Build GRCh38
rs776650194 Chr.17: 73252591-73252591 on Build GRCh38
rs543474279 Chr.17: 73252593-73252593 on Build GRCh38
rs749075573 Chr.17: 73252656-73252656 on Build GRCh38
rs1038763537 Chr.17: 73252564-73252564 on Build GRCh38
rs74909356 Chr.17: 73252638-73252638 on Build GRCh38
rs893289253 Chr.17: 73252830-73252830 on Build GRCh38
rs765285744 Chr.17: 73252678-73252678 on Build GRCh38
rs528053609 Chr.17: 73252856-73252856 on Build GRCh38
rs145442898 Chr.17: 73252824-73252824 on Build GRCh38
rs185025061 Chr.17: 73252948-73252948 on Build GRCh38
rs201954808 Chr.17: 73253001-73253001 on Build GRCh38
rs941991466 Chr.17: 73252842-73252842 on Build GRCh38
rs759491240 Chr.17: 73252877-73252877 on Build GRCh38
rs949035540 Chr.17: 73252993-73252993 on Build GRCh38
rs566336093 Chr.17: 73252902-73252902 on Build GRCh38
rs529862493 Chr.17: 73252757-73252757 on Build GRCh38

View Full Product Details