Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74523490-74524002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300LB
Entrez Gene ID
124599
Gene Name
CD300 molecule like family member b
Gene Aliases
CD300b, CLM-7, CLM7, CMRF35-A2, IREM-3, IREM3, TREM-5, TREM5
UniGene
Hs.313343
Gene Chromosome Location
Chr.17: 74521174-74531474 on Build GRCh38
Transcripts
NM_174892.3 XM_005257027.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3932720 c.482-1G>C p.? Chr.17: 74523652-74523652 on Build GRCh38
5972395 c.527T>C p.M176T Chr.17: 74523606-74523606 on Build GRCh38
394165 c.519C>A p.N173K Chr.17: 74523614-74523614 on Build GRCh38
4415940 c.482-8G>A p.? Chr.17: 74523659-74523659 on Build GRCh38
5722677 c.509C>T p.S170L Chr.17: 74523624-74523624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113638198 Chr.17: 74523801-74523801 on Build GRCh38
rs779617250 Chr.17: 74523634-74523634 on Build GRCh38
rs372672106 Chr.17: 74523699-74523699 on Build GRCh38
rs780940093 Chr.17: 74523547-74523547 on Build GRCh38
rs146364730 Chr.17: 74523676-74523676 on Build GRCh38
rs748147677 Chr.17: 74523549-74523549 on Build GRCh38
rs769906446 Chr.17: 74523658-74523658 on Build GRCh38
rs376742858 Chr.17: 74523724-74523724 on Build GRCh38
rs202114723 Chr.17: 74523682-74523682 on Build GRCh38
rs1014867683 Chr.17: 74523910-74523910 on Build GRCh38
rs759815150 Chr.17: 74523578-74523578 on Build GRCh38
rs767801454 Chr.17: 74523920-74523920 on Build GRCh38
rs372167958 Chr.17: 74523544-74523544 on Build GRCh38
rs751245293 Chr.17: 74523635-74523635 on Build GRCh38
rs375010301 Chr.17: 74523570-74523570 on Build GRCh38
rs367952780 Chr.17: 74523612-74523612 on Build GRCh38
rs1044901091 Chr.17: 74523807-74523807 on Build GRCh38
rs1024554846 Chr.17: 74523917-74523917 on Build GRCh38
rs779813963 Chr.17: 74523532-74523532 on Build GRCh38
rs967642698 Chr.17: 74523921-74523921 on Build GRCh38
rs956361437 Chr.17: 74523895-74523895 on Build GRCh38
rs760893352 Chr.17: 74523596-74523596 on Build GRCh38
rs764705121 Chr.17: 74523615-74523615 on Build GRCh38
rs768874346 Chr.17: 74523674-74523674 on Build GRCh38
rs1030795983 Chr.17: 74523880-74523880 on Build GRCh38
rs775529760 Chr.17: 74523668-74523668 on Build GRCh38
rs772487384 Chr.17: 74523581-74523581 on Build GRCh38
rs934061081 Chr.17: 74523739-74523739 on Build GRCh38
rs1038060087 Chr.17: 74523593-74523593 on Build GRCh38
rs761717778 Chr.17: 74523928-74523928 on Build GRCh38
rs200261260 Chr.17: 74523592-74523592 on Build GRCh38
rs766463307 Chr.17: 74523971-74523971 on Build GRCh38
rs137895820 Chr.17: 74523629-74523629 on Build GRCh38
rs142435917 Chr.17: 74523645-74523645 on Build GRCh38
rs143428568 Chr.17: 74523616-74523616 on Build GRCh38
rs762350959 Chr.17: 74523614-74523614 on Build GRCh38
rs1010996887 Chr.17: 74523908-74523908 on Build GRCh38
rs538373553 Chr.17: 74523793-74523793 on Build GRCh38
rs764543166 Chr.17: 74523606-74523606 on Build GRCh38
rs760991369 Chr.17: 74523671-74523671 on Build GRCh38
rs144492950 Chr.17: 74523974-74523974 on Build GRCh38
rs370650884 Chr.17: 74523703-74523703 on Build GRCh38
rs765802456 Chr.17: 74523691-74523691 on Build GRCh38
rs1472726 Chr.17: 74523659-74523659 on Build GRCh38
rs1047166540 Chr.17: 74523763-74523763 on Build GRCh38
rs370422556 Chr.17: 74523969-74523969 on Build GRCh38
rs964212993 Chr.17: 74523718-74523718 on Build GRCh38
rs776061321 Chr.17: 74523551-74523551 on Build GRCh38
rs754687733 Chr.17: 74523637-74523637 on Build GRCh38
rs772288726 Chr.17: 74523661-74523661 on Build GRCh38
rs1034859931 Chr.17: 74523830-74523830 on Build GRCh38
rs1003287927 Chr.17: 74523859-74523859 on Build GRCh38
rs781081246 Chr.17: 74523639-74523639 on Build GRCh38
rs977646919 Chr.17: 74523953-74523953 on Build GRCh38
rs868327740 Chr.17: 74523591-74523591 on Build GRCh38
rs760913699 Chr.17: 74523679-74523679 on Build GRCh38
rs773714408 Chr.17: 74523702-74523702 on Build GRCh38
rs975567799 Chr.17: 74523722-74523722 on Build GRCh38
rs557624854 Chr.17: 74523520-74523520 on Build GRCh38
rs745874819 Chr.17: 74523660-74523660 on Build GRCh38
rs771103106 Chr.17: 74523554-74523554 on Build GRCh38
rs958885412 Chr.17: 74523856-74523856 on Build GRCh38
rs1472725 Chr.17: 74523794-74523794 on Build GRCh38
rs370309053 Chr.17: 74523538-74523538 on Build GRCh38
rs376342343 Chr.17: 74523723-74523723 on Build GRCh38
rs754853592 Chr.17: 74523764-74523764 on Build GRCh38
rs540014956 Chr.17: 74523791-74523791 on Build GRCh38
rs115899842 Chr.17: 74523765-74523765 on Build GRCh38
rs994193869 Chr.17: 74523516-74523516 on Build GRCh38
rs1023360659 Chr.17: 74523644-74523644 on Build GRCh38
rs1050143068 Chr.17: 74523705-74523705 on Build GRCh38
rs1040272727 Chr.17: 74523726-74523726 on Build GRCh38
rs1047131076 Chr.17: 74523790-74523790 on Build GRCh38
rs139775917 Chr.17: 74523711-74523711 on Build GRCh38

View Full Product Details