Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74864699-74865109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDXR
Entrez Gene ID
2232
Gene Name
ferredoxin reductase
Gene Aliases
ADXR
UniGene
Hs.69745
Gene Chromosome Location
Chr.17: 74862497-74873031 on Build GRCh38
Transcripts
NM_001258012.3 NM_001258013.3 NM_001258014.3 NM_001258015.2 NM_001258016.3 NM_004110.5 NM_024417.4 NR_047576.2 XM_006721772.1 XM_011524527.1 XM_011524528.1 XM_011524529.1 XM_011524531.2 XM_011524532.2 XM_011524533.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4861489 c.548G>T p.G183V Chr.17: 74864861-74864861 on Build GRCh38
5364053 c.662T>C p.L221P Chr.17: 74864897-74864897 on Build GRCh38
6229044 c.700C>T p.R234W Chr.17: 74864859-74864859 on Build GRCh38
7267557 c.641C>T p.T214M Chr.17: 74864918-74864918 on Build GRCh38
7267559 c.491C>T p.T164M Chr.17: 74864918-74864918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368211441 Chr.17: 74865071-74865071 on Build GRCh38
rs144522648 Chr.17: 74865031-74865031 on Build GRCh38
rs541505879 Chr.17: 74865043-74865043 on Build GRCh38
rs748044388 Chr.17: 74864995-74864995 on Build GRCh38
rs1026772364 Chr.17: 74864744-74864744 on Build GRCh38
rs35102176 Chr.17: 74864935-74864935 on Build GRCh38
rs777429435 Chr.17: 74864819-74864819 on Build GRCh38
rs373972990 Chr.17: 74864782-74864782 on Build GRCh38
rs970119640 Chr.17: 74864747-74864747 on Build GRCh38
rs778378259 Chr.17: 74864882-74864882 on Build GRCh38
rs761802850 Chr.17: 74864926-74864926 on Build GRCh38
rs35692345 Chr.17: 74864903-74864903 on Build GRCh38
rs913053748 Chr.17: 74864777-74864777 on Build GRCh38
rs571528622 Chr.17: 74865002-74865002 on Build GRCh38
rs895478288 Chr.17: 74865065-74865065 on Build GRCh38
rs111758978 Chr.17: 74864808-74864808 on Build GRCh38
rs771829779 Chr.17: 74864838-74864838 on Build GRCh38
rs780295136 Chr.17: 74865056-74865056 on Build GRCh38
rs759614440 Chr.17: 74865001-74865001 on Build GRCh38
rs544497279 Chr.17: 74865070-74865070 on Build GRCh38
rs766423469 Chr.17: 74864927-74864927 on Build GRCh38
rs369527938 Chr.17: 74864773-74864773 on Build GRCh38
rs753048793 Chr.17: 74864874-74864874 on Build GRCh38
rs758648690 Chr.17: 74864794-74864794 on Build GRCh38
rs775293878 Chr.17: 74864907-74864907 on Build GRCh38
rs377370693 Chr.17: 74864966-74864966 on Build GRCh38
rs774427615 Chr.17: 74864930-74864930 on Build GRCh38
rs763977480 Chr.17: 74864845-74864845 on Build GRCh38
rs113660127 Chr.17: 74864830-74864830 on Build GRCh38
rs375906566 Chr.17: 74864973-74864973 on Build GRCh38
rs752745486 Chr.17: 74864964-74864964 on Build GRCh38
rs950215562 Chr.17: 74864868-74864868 on Build GRCh38
rs750435603 Chr.17: 74864790-74864790 on Build GRCh38
rs555332221 Chr.17: 74865044-74865044 on Build GRCh38
rs370653074 Chr.17: 74864841-74864841 on Build GRCh38
rs951876731 Chr.17: 74865000-74865000 on Build GRCh38
rs769433044 Chr.17: 74864829-74864829 on Build GRCh38
rs891135331 Chr.17: 74864849-74864849 on Build GRCh38
rs763067630 Chr.17: 74864858-74864858 on Build GRCh38
rs757769616 Chr.17: 74864959-74864959 on Build GRCh38
rs745565334 Chr.17: 74864899-74864899 on Build GRCh38
rs201431096 Chr.17: 74864856-74864856 on Build GRCh38
rs763768416 Chr.17: 74864777-74864778 on Build GRCh38
rs574307590 Chr.17: 74865013-74865013 on Build GRCh38
rs766889330 Chr.17: 74864979-74864980 on Build GRCh38
rs746030706 Chr.17: 74865035-74865035 on Build GRCh38
rs959959699 Chr.17: 74864762-74864762 on Build GRCh38
rs763373202 Chr.17: 74865050-74865051 on Build GRCh38
rs765315364 Chr.17: 74864783-74864783 on Build GRCh38
rs768414971 Chr.17: 74864917-74864917 on Build GRCh38
rs753137078 Chr.17: 74864936-74864936 on Build GRCh38
rs377283703 Chr.17: 74864981-74864981 on Build GRCh38
rs374574829 Chr.17: 74864883-74864883 on Build GRCh38
rs370448217 Chr.17: 74864972-74864972 on Build GRCh38
rs370506060 Chr.17: 74864779-74864779 on Build GRCh38
rs575089940 Chr.17: 74865057-74865057 on Build GRCh38
rs547136156 Chr.17: 74864815-74864815 on Build GRCh38
rs979478271 Chr.17: 74864788-74864788 on Build GRCh38
rs770035976 Chr.17: 74865054-74865054 on Build GRCh38
rs199826370 Chr.17: 74864918-74864918 on Build GRCh38
rs752616185 Chr.17: 74864781-74864781 on Build GRCh38
rs748770358 Chr.17: 74864828-74864828 on Build GRCh38
rs148950642 Chr.17: 74864943-74864943 on Build GRCh38
rs918807724 Chr.17: 74865026-74865026 on Build GRCh38
rs377677821 Chr.17: 74864859-74864859 on Build GRCh38
rs756723359 Chr.17: 74864879-74864879 on Build GRCh38
rs753323215 Chr.17: 74864813-74864813 on Build GRCh38
rs780948868 Chr.17: 74864983-74864983 on Build GRCh38
rs760610593 Chr.17: 74864843-74864843 on Build GRCh38
rs1035765959 Chr.17: 74864760-74864760 on Build GRCh38
rs756758083 Chr.17: 74864885-74864885 on Build GRCh38
rs778482062 Chr.17: 74864886-74864886 on Build GRCh38
rs781612378 Chr.17: 74864804-74864804 on Build GRCh38
rs768502863 Chr.17: 74864982-74864982 on Build GRCh38
rs764524114 Chr.17: 74864939-74864939 on Build GRCh38
rs201356542 Chr.17: 74864999-74864999 on Build GRCh38
rs564282507 Chr.17: 74864726-74864726 on Build GRCh38
rs761341065 Chr.17: 74864977-74864978 on Build GRCh38
rs950127560 Chr.17: 74865053-74865053 on Build GRCh38
rs369903298 Chr.17: 74864872-74864872 on Build GRCh38
rs944057459 Chr.17: 74865005-74865005 on Build GRCh38
rs988702605 Chr.17: 74864769-74864769 on Build GRCh38
rs746756041 Chr.17: 74864913-74864913 on Build GRCh38
rs375282282 Chr.17: 74864799-74864799 on Build GRCh38
rs34964564 Chr.17: 74864800-74864800 on Build GRCh38
rs751791983 Chr.17: 74864865-74864865 on Build GRCh38
rs1037312154 Chr.17: 74864855-74864855 on Build GRCh38
rs755334781 Chr.17: 74864802-74864802 on Build GRCh38
rs897358847 Chr.17: 74864722-74864722 on Build GRCh38
rs1996710 Chr.17: 74865023-74865023 on Build GRCh38
rs979755554 Chr.17: 74864757-74864757 on Build GRCh38
rs1035333750 Chr.17: 74864897-74864897 on Build GRCh38
rs944260508 Chr.17: 74864851-74864851 on Build GRCh38

View Full Product Details