Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75778728-75779217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H3F3B
Entrez Gene ID
3021
Gene Name
H3 histone, family 3B
Gene Aliases
H3.3B
UniGene
Hs.180877
Gene Chromosome Location
Chr.17: 75776434-75779935 on Build GRCh38
Transcripts
NM_005324.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6958942 c.109_128+8del28 p.? Chr.17: 75779039-75779066 on Build GRCh38
6954691 c.278_279insT p.Q94fs*4 Chr.17: 75778813-75778814 on Build GRCh38
6962429 c.25C>A p.R9S Chr.17: 75779150-75779150 on Build GRCh38
6961067 c.148C>T p.R50* Chr.17: 75778944-75778944 on Build GRCh38
6983003 c.18G>T p.Q6H Chr.17: 75779157-75779157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753840984 Chr.17: 75778984-75778984 on Build GRCh38
rs754901721 Chr.17: 75778798-75778798 on Build GRCh38
rs375326820 Chr.17: 75778973-75778973 on Build GRCh38
rs780930857 Chr.17: 75778765-75778765 on Build GRCh38
rs748325508 Chr.17: 75778997-75778997 on Build GRCh38
rs769438639 Chr.17: 75779185-75779185 on Build GRCh38
rs753788015 Chr.17: 75779028-75779028 on Build GRCh38
rs201602885 Chr.17: 75779100-75779100 on Build GRCh38
rs368183110 Chr.17: 75779193-75779193 on Build GRCh38
rs750282970 Chr.17: 75779037-75779037 on Build GRCh38
rs774850649 Chr.17: 75779022-75779022 on Build GRCh38
rs369011460 Chr.17: 75778777-75778777 on Build GRCh38
rs746366357 Chr.17: 75779151-75779151 on Build GRCh38
rs752295905 Chr.17: 75778978-75778978 on Build GRCh38
rs551047862 Chr.17: 75779160-75779160 on Build GRCh38
rs150028698 Chr.17: 75778825-75778825 on Build GRCh38
rs763519204 Chr.17: 75779026-75779026 on Build GRCh38
rs777402446 Chr.17: 75778911-75778911 on Build GRCh38
rs755150586 Chr.17: 75779038-75779038 on Build GRCh38
rs9895763 Chr.17: 75778796-75778796 on Build GRCh38
rs778281521 Chr.17: 75779008-75779008 on Build GRCh38
rs746136410 Chr.17: 75778774-75778774 on Build GRCh38
rs375253689 Chr.17: 75779012-75779012 on Build GRCh38
rs779570578 Chr.17: 75778752-75778752 on Build GRCh38
rs867543814 Chr.17: 75779083-75779083 on Build GRCh38
rs761814713 Chr.17: 75779199-75779199 on Build GRCh38
rs372771522 Chr.17: 75778779-75778779 on Build GRCh38
rs756599830 Chr.17: 75779014-75779015 on Build GRCh38
rs749775664 Chr.17: 75779070-75779070 on Build GRCh38
rs371984165 Chr.17: 75778993-75778993 on Build GRCh38
rs754123266 Chr.17: 75779031-75779031 on Build GRCh38
rs368010219 Chr.17: 75778990-75778990 on Build GRCh38
rs192068190 Chr.17: 75778768-75778768 on Build GRCh38
rs896000081 Chr.17: 75778900-75778900 on Build GRCh38
rs557779142 Chr.17: 75779191-75779191 on Build GRCh38
rs760174028 Chr.17: 75779025-75779025 on Build GRCh38
rs770184463 Chr.17: 75779004-75779004 on Build GRCh38
rs764805226 Chr.17: 75778759-75778759 on Build GRCh38
rs199901785 Chr.17: 75778797-75778797 on Build GRCh38
rs765331604 Chr.17: 75779112-75779112 on Build GRCh38
rs757047753 Chr.17: 75778985-75778985 on Build GRCh38
rs1000160446 Chr.17: 75778942-75778942 on Build GRCh38
rs369866281 Chr.17: 75778807-75778807 on Build GRCh38
rs750002250 Chr.17: 75778791-75778791 on Build GRCh38
rs879076699 Chr.17: 75778967-75778967 on Build GRCh38
rs74374260 Chr.17: 75779189-75779189 on Build GRCh38
rs61734773 Chr.17: 75779076-75779076 on Build GRCh38
rs746293947 Chr.17: 75779082-75779082 on Build GRCh38
rs993342434 Chr.17: 75779121-75779121 on Build GRCh38
rs771295937 Chr.17: 75778975-75778975 on Build GRCh38
rs773674895 Chr.17: 75778861-75778861 on Build GRCh38
rs772231568 Chr.17: 75778775-75778775 on Build GRCh38
rs746898890 Chr.17: 75778753-75778753 on Build GRCh38
rs958180165 Chr.17: 75779197-75779197 on Build GRCh38
rs761809884 Chr.17: 75779109-75779109 on Build GRCh38
rs766083180 Chr.17: 75778894-75778894 on Build GRCh38
rs11547396 Chr.17: 75779053-75779053 on Build GRCh38
rs780367196 Chr.17: 75778918-75778918 on Build GRCh38
rs772135674 Chr.17: 75779187-75779190 on Build GRCh38
rs545389058 Chr.17: 75779020-75779020 on Build GRCh38
rs188695621 Chr.17: 75779139-75779139 on Build GRCh38
rs781352768 Chr.17: 75778936-75778936 on Build GRCh38
rs756310531 Chr.17: 75779127-75779127 on Build GRCh38
rs11547392 Chr.17: 75778859-75778859 on Build GRCh38
rs762028119 Chr.17: 75778789-75778789 on Build GRCh38
rs375470878 Chr.17: 75779114-75779114 on Build GRCh38
rs374778385 Chr.17: 75778903-75778903 on Build GRCh38
rs762411753 Chr.17: 75778845-75778845 on Build GRCh38
rs748581652 Chr.17: 75778951-75778951 on Build GRCh38
rs199824594 Chr.17: 75778790-75778790 on Build GRCh38
rs777858573 Chr.17: 75779068-75779068 on Build GRCh38
rs115278451 Chr.17: 75779169-75779169 on Build GRCh38
rs755829204 Chr.17: 75778819-75778819 on Build GRCh38
rs797020985 Chr.17: 75779183-75779183 on Build GRCh38
rs202221480 Chr.17: 75778822-75778822 on Build GRCh38
rs373849930 Chr.17: 75778979-75778979 on Build GRCh38
rs765294615 Chr.17: 75778960-75778960 on Build GRCh38
rs768815367 Chr.17: 75778927-75778927 on Build GRCh38
rs765100412 Chr.17: 75779036-75779036 on Build GRCh38
rs11547397 Chr.17: 75779110-75779110 on Build GRCh38
rs147330413 Chr.17: 75779073-75779073 on Build GRCh38
rs770372843 Chr.17: 75778971-75778971 on Build GRCh38
rs764152280 Chr.17: 75778982-75778982 on Build GRCh38
rs764361933 Chr.17: 75779196-75779196 on Build GRCh38
rs762507759 Chr.17: 75778800-75778801 on Build GRCh38
rs1041834044 Chr.17: 75779027-75779027 on Build GRCh38
rs757779367 Chr.17: 75778750-75778750 on Build GRCh38
rs777042953 Chr.17: 75778781-75778781 on Build GRCh38
rs371386581 Chr.17: 75778891-75778891 on Build GRCh38
rs11547395 Chr.17: 75779125-75779125 on Build GRCh38
rs568882617 Chr.17: 75778906-75778906 on Build GRCh38
rs754425060 Chr.17: 75778896-75778896 on Build GRCh38
rs772467371 Chr.17: 75778831-75778831 on Build GRCh38
rs778883358 Chr.17: 75778988-75778988 on Build GRCh38
rs559347636 Chr.17: 75779032-75779032 on Build GRCh38
rs771242574 Chr.17: 75779019-75779019 on Build GRCh38
rs745994095 Chr.17: 75779185-75779186 on Build GRCh38
rs200368963 Chr.17: 75779067-75779067 on Build GRCh38
rs767833460 Chr.17: 75779052-75779052 on Build GRCh38
rs541360201 Chr.17: 75778849-75778849 on Build GRCh38
rs765468661 Chr.17: 75778846-75778846 on Build GRCh38
rs11547388 Chr.17: 75778885-75778885 on Build GRCh38
rs752871689 Chr.17: 75779055-75779055 on Build GRCh38
rs772338531 Chr.17: 75779088-75779088 on Build GRCh38
rs753988472 Chr.17: 75779136-75779136 on Build GRCh38
rs371726328 Chr.17: 75779198-75779198 on Build GRCh38
rs1033102099 Chr.17: 75778788-75778788 on Build GRCh38
rs367605635 Chr.17: 75778921-75778921 on Build GRCh38
rs748127055 Chr.17: 75778816-75778816 on Build GRCh38
rs758681498 Chr.17: 75778915-75778915 on Build GRCh38
rs200810602 Chr.17: 75778991-75778991 on Build GRCh38
rs779064600 Chr.17: 75779122-75779123 on Build GRCh38
rs768872612 Chr.17: 75778840-75778840 on Build GRCh38
rs11547391 Chr.17: 75779138-75779138 on Build GRCh38
rs776937727 Chr.17: 75778843-75778843 on Build GRCh38
rs745495299 Chr.17: 75778799-75778800 on Build GRCh38
rs778237161 Chr.17: 75779017-75779027 on Build GRCh38
rs762748626 Chr.17: 75779124-75779124 on Build GRCh38
rs747143516 Chr.17: 75778834-75778834 on Build GRCh38
rs967385289 Chr.17: 75778764-75778764 on Build GRCh38
rs144205696 Chr.17: 75779094-75779094 on Build GRCh38
rs757357510 Chr.17: 75779030-75779031 on Build GRCh38
rs368752239 Chr.17: 75779029-75779029 on Build GRCh38
rs2071654 Chr.17: 75778760-75778760 on Build GRCh38
rs746674587 Chr.17: 75779097-75779097 on Build GRCh38
rs951912780 Chr.17: 75778986-75778986 on Build GRCh38
rs574608131 Chr.17: 75778969-75778969 on Build GRCh38
rs776276812 Chr.17: 75779184-75779184 on Build GRCh38
rs972005035 Chr.17: 75778820-75778820 on Build GRCh38
rs759159728 Chr.17: 75779021-75779021 on Build GRCh38
rs766917543 Chr.17: 75778977-75778977 on Build GRCh38
rs775708260 Chr.17: 75778832-75778832 on Build GRCh38
rs753734696 Chr.17: 75778912-75778912 on Build GRCh38
rs879116715 Chr.17: 75778808-75778808 on Build GRCh38
rs937733974 Chr.17: 75778888-75778888 on Build GRCh38
rs760589830 Chr.17: 75778763-75778763 on Build GRCh38
rs370991523 Chr.17: 75779178-75779178 on Build GRCh38
rs1033399291 Chr.17: 75778954-75778954 on Build GRCh38
rs775518127 Chr.17: 75778974-75778974 on Build GRCh38
rs753228847 Chr.17: 75778991-75778996 on Build GRCh38
rs372492941 Chr.17: 75778762-75778762 on Build GRCh38
rs937677994 Chr.17: 75779010-75779010 on Build GRCh38
rs768462270 Chr.17: 75779180-75779180 on Build GRCh38
rs184828841 Chr.17: 75778976-75778976 on Build GRCh38
rs375775315 Chr.17: 75778776-75778776 on Build GRCh38

View Full Product Details