Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 48663631-48664129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTIF
Entrez Gene ID
9811
Gene Name
cap binding complex dependent translation initiation factor
Gene Aliases
Gm672, KIAA0427
UniGene
Hs.145230
Gene Chromosome Location
Chr.18: 48539056-48863217 on Build GRCh38
Transcripts
NM_001142397.1 NM_014772.2 XM_005258392.4 XM_006722583.3 XM_006722586.3 XM_006722587.3 XM_006722588.3 XM_006722589.3 XM_011526278.2 XM_011526279.2 XM_017026100.1 XM_017026101.1 XM_017026102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4444641 c.161C>T p.A54V Chr.18: 48663804-48663804 on Build GRCh38
1241338 c.287G>A p.G96D Chr.18: 48663786-48663786 on Build GRCh38
7270111 c.146C>T p.T49M Chr.18: 48663789-48663789 on Build GRCh38
5056059 c.257G>A p.G86D Chr.18: 48663756-48663756 on Build GRCh38
4487631 c.176C>T p.S59F Chr.18: 48663819-48663819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373648903 Chr.18: 48663712-48663712 on Build GRCh38
rs777454329 Chr.18: 48663863-48663863 on Build GRCh38
rs972871965 Chr.18: 48664102-48664102 on Build GRCh38
rs748746672 Chr.18: 48663736-48663736 on Build GRCh38
rs769981777 Chr.18: 48663973-48663973 on Build GRCh38
rs949457931 Chr.18: 48664006-48664006 on Build GRCh38
rs774058443 Chr.18: 48663752-48663752 on Build GRCh38
rs768129432 Chr.18: 48663716-48663716 on Build GRCh38
rs370286375 Chr.18: 48663711-48663711 on Build GRCh38
rs928515097 Chr.18: 48663669-48663669 on Build GRCh38
rs1051426495 Chr.18: 48664096-48664096 on Build GRCh38
rs915781784 Chr.18: 48663698-48663698 on Build GRCh38
rs1037883397 Chr.18: 48663725-48663725 on Build GRCh38
rs763060686 Chr.18: 48663788-48663788 on Build GRCh38
rs770945661 Chr.18: 48663775-48663775 on Build GRCh38
rs937154065 Chr.18: 48663970-48663970 on Build GRCh38
rs965160351 Chr.18: 48664098-48664098 on Build GRCh38
rs375182568 Chr.18: 48663893-48663893 on Build GRCh38
rs1007053144 Chr.18: 48664060-48664060 on Build GRCh38
rs764183797 Chr.18: 48663709-48663709 on Build GRCh38
rs896517008 Chr.18: 48663701-48663701 on Build GRCh38
rs890704523 Chr.18: 48663794-48663794 on Build GRCh38
rs775746916 Chr.18: 48663783-48663783 on Build GRCh38
rs1048260571 Chr.18: 48663663-48663663 on Build GRCh38
rs771238225 Chr.18: 48663728-48663728 on Build GRCh38
rs889993703 Chr.18: 48664050-48664050 on Build GRCh38
rs75687084 Chr.18: 48663829-48663829 on Build GRCh38
rs890152158 Chr.18: 48664097-48664097 on Build GRCh38
rs752264597 Chr.18: 48663853-48663853 on Build GRCh38
rs927133917 Chr.18: 48663960-48663960 on Build GRCh38
rs1042408029 Chr.18: 48664026-48664026 on Build GRCh38
rs374938044 Chr.18: 48663856-48663856 on Build GRCh38
rs745853920 Chr.18: 48663891-48663891 on Build GRCh38
rs754652767 Chr.18: 48663740-48663740 on Build GRCh38
rs1032540154 Chr.18: 48664049-48664049 on Build GRCh38
rs528388395 Chr.18: 48663949-48663949 on Build GRCh38
rs561738810 Chr.18: 48664066-48664066 on Build GRCh38
rs1055197020 Chr.18: 48663697-48663697 on Build GRCh38
rs965452407 Chr.18: 48663902-48663902 on Build GRCh38
rs143431042 Chr.18: 48663665-48663665 on Build GRCh38
rs954200111 Chr.18: 48663868-48663868 on Build GRCh38
rs79505651 Chr.18: 48663927-48663927 on Build GRCh38
rs541432086 Chr.18: 48663955-48663955 on Build GRCh38
rs902491588 Chr.18: 48664032-48664032 on Build GRCh38
rs369328929 Chr.18: 48663785-48663785 on Build GRCh38
rs555755626 Chr.18: 48663802-48663802 on Build GRCh38
rs779515229 Chr.18: 48663729-48663729 on Build GRCh38
rs918969282 Chr.18: 48664088-48664088 on Build GRCh38
rs544337905 Chr.18: 48663831-48663831 on Build GRCh38
rs777810338 Chr.18: 48663762-48663762 on Build GRCh38
rs771747837 Chr.18: 48663761-48663761 on Build GRCh38
rs576675021 Chr.18: 48663877-48663877 on Build GRCh38
rs750889472 Chr.18: 48663721-48663721 on Build GRCh38
rs938607759 Chr.18: 48663684-48663684 on Build GRCh38
rs770998796 Chr.18: 48663867-48663867 on Build GRCh38
rs367786142 Chr.18: 48663774-48663774 on Build GRCh38
rs896575426 Chr.18: 48663998-48663998 on Build GRCh38
rs193106168 Chr.18: 48663840-48663840 on Build GRCh38
rs918218430 Chr.18: 48663931-48663931 on Build GRCh38
rs201605930 Chr.18: 48663763-48663763 on Build GRCh38
rs1001103359 Chr.18: 48664036-48664036 on Build GRCh38
rs746763407 Chr.18: 48663866-48663866 on Build GRCh38
rs1055650751 Chr.18: 48663976-48663976 on Build GRCh38
rs762305421 Chr.18: 48663805-48663805 on Build GRCh38
rs991389289 Chr.18: 48663989-48663989 on Build GRCh38
rs1020167662 Chr.18: 48664089-48664089 on Build GRCh38
rs902773868 Chr.18: 48663787-48663787 on Build GRCh38
rs1021432284 Chr.18: 48663861-48663861 on Build GRCh38
rs539460174 Chr.18: 48663800-48663800 on Build GRCh38
rs941828589 Chr.18: 48664053-48664053 on Build GRCh38
rs941768814 Chr.18: 48663703-48663703 on Build GRCh38
rs1019359408 Chr.18: 48663892-48663892 on Build GRCh38
rs781219952 Chr.18: 48663869-48663869 on Build GRCh38
rs778682098 Chr.18: 48663847-48663847 on Build GRCh38
rs938106009 Chr.18: 48663692-48663692 on Build GRCh38
rs931236772 Chr.18: 48663942-48663942 on Build GRCh38
rs576126993 Chr.18: 48663901-48663901 on Build GRCh38
rs767743057 Chr.18: 48663813-48663813 on Build GRCh38
rs142723132 Chr.18: 48663757-48663757 on Build GRCh38
rs754383453 Chr.18: 48663841-48663841 on Build GRCh38
rs184269063 Chr.18: 48663914-48663914 on Build GRCh38

View Full Product Details