Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49837431-49837928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO5B
Entrez Gene ID
4645
Gene Name
myosin VB
Gene Aliases
-
UniGene
Hs.720076
Gene Chromosome Location
Chr.18: 49822786-50195081 on Build GRCh38
Transcripts
NM_001080467.2 NR_117096.1

Gene Symbol
SNHG22
Entrez Gene ID
103091864
Gene Name
small nucleolar RNA host gene 22
Gene Aliases
SCARNA17, SCARNA17HG
UniGene
Hs.646459
Gene Chromosome Location
Chr.18: 49814023-49851059 on Build GRCh38
Transcripts
NM_001080467.2 NR_117096.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6317549 c.4950C>A p.N1650K Chr.18: 49837705-49837705 on Build GRCh38
4072429 c.5072C>T p.A1691V Chr.18: 49837583-49837583 on Build GRCh38
1180643 c.4939G>A p.D1647N Chr.18: 49837716-49837716 on Build GRCh38
1750534 c.5094C>G p.L1698L Chr.18: 49837561-49837561 on Build GRCh38
1388945 c.4953A>G p.S1651S Chr.18: 49837702-49837702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772909385 Chr.18: 49837557-49837557 on Build GRCh38
rs201430138 Chr.18: 49837750-49837750 on Build GRCh38
rs77401395 Chr.18: 49837782-49837782 on Build GRCh38
rs778498719 Chr.18: 49837551-49837551 on Build GRCh38
rs1015340191 Chr.18: 49837587-49837587 on Build GRCh38
rs756487835 Chr.18: 49837602-49837602 on Build GRCh38
rs779955003 Chr.18: 49837742-49837742 on Build GRCh38
rs969988408 Chr.18: 49837653-49837653 on Build GRCh38
rs750995984 Chr.18: 49837832-49837832 on Build GRCh38
rs200556602 Chr.18: 49837535-49837535 on Build GRCh38
rs746850413 Chr.18: 49837638-49837638 on Build GRCh38
rs760972277 Chr.18: 49837754-49837754 on Build GRCh38
rs1037076353 Chr.18: 49837874-49837874 on Build GRCh38
rs756813138 Chr.18: 49837733-49837733 on Build GRCh38
rs776073123 Chr.18: 49837818-49837818 on Build GRCh38
rs541407556 Chr.18: 49837499-49837499 on Build GRCh38
rs764370265 Chr.18: 49837498-49837498 on Build GRCh38
rs773478649 Chr.18: 49837678-49837678 on Build GRCh38
rs890830778 Chr.18: 49837821-49837821 on Build GRCh38
rs75537257 Chr.18: 49837686-49837686 on Build GRCh38
rs755484254 Chr.18: 49837611-49837611 on Build GRCh38
rs772198023 Chr.18: 49837747-49837747 on Build GRCh38
rs760548998 Chr.18: 49837470-49837470 on Build GRCh38
rs774367800 Chr.18: 49837576-49837576 on Build GRCh38
rs987911570 Chr.18: 49837521-49837521 on Build GRCh38
rs866197728 Chr.18: 49837698-49837698 on Build GRCh38
rs200540643 Chr.18: 49837614-49837614 on Build GRCh38
rs200907103 Chr.18: 49837549-49837549 on Build GRCh38
rs771703731 Chr.18: 49837556-49837556 on Build GRCh38
rs1051156054 Chr.18: 49837761-49837761 on Build GRCh38
rs751792384 Chr.18: 49837805-49837805 on Build GRCh38
rs373021577 Chr.18: 49837561-49837562 on Build GRCh38
rs545924419 Chr.18: 49837857-49837857 on Build GRCh38
rs375780015 Chr.18: 49837740-49837740 on Build GRCh38
rs765900394 Chr.18: 49837816-49837816 on Build GRCh38
rs368237603 Chr.18: 49837664-49837664 on Build GRCh38
rs889378145 Chr.18: 49837764-49837764 on Build GRCh38
rs766861389 Chr.18: 49837680-49837680 on Build GRCh38
rs764095230 Chr.18: 49837496-49837497 on Build GRCh38
rs200915753 Chr.18: 49837819-49837819 on Build GRCh38
rs571997090 Chr.18: 49837473-49837473 on Build GRCh38
rs772242451 Chr.18: 49837674-49837674 on Build GRCh38
rs775421806 Chr.18: 49837567-49837567 on Build GRCh38
rs898263374 Chr.18: 49837899-49837899 on Build GRCh38
rs758022597 Chr.18: 49837632-49837632 on Build GRCh38
rs779757000 Chr.18: 49837848-49837848 on Build GRCh38
rs777084765 Chr.18: 49837722-49837722 on Build GRCh38
rs77840018 Chr.18: 49837561-49837561 on Build GRCh38
rs750276531 Chr.18: 49837842-49837842 on Build GRCh38
rs112417235 Chr.18: 49837593-49837593 on Build GRCh38
rs774456015 Chr.18: 49837652-49837652 on Build GRCh38
rs886053874 Chr.18: 49837817-49837817 on Build GRCh38
rs200090894 Chr.18: 49837839-49837839 on Build GRCh38
rs775582268 Chr.18: 49837474-49837474 on Build GRCh38
rs750762016 Chr.18: 49837511-49837511 on Build GRCh38
rs761973304 Chr.18: 49837698-49837699 on Build GRCh38
rs760147953 Chr.18: 49837695-49837695 on Build GRCh38
rs546240591 Chr.18: 49837739-49837739 on Build GRCh38
rs1033983398 Chr.18: 49837894-49837894 on Build GRCh38
rs372387496 Chr.18: 49837679-49837679 on Build GRCh38
rs780910211 Chr.18: 49837640-49837640 on Build GRCh38
rs748242455 Chr.18: 49837746-49837746 on Build GRCh38
rs772999913 Chr.18: 49837468-49837468 on Build GRCh38
rs149010847 Chr.18: 49837639-49837639 on Build GRCh38
rs768231362 Chr.18: 49837558-49837558 on Build GRCh38
rs375529250 Chr.18: 49837644-49837644 on Build GRCh38
rs767796633 Chr.18: 49837502-49837502 on Build GRCh38
rs751241323 Chr.18: 49837725-49837725 on Build GRCh38
rs1032363121 Chr.18: 49837822-49837822 on Build GRCh38
rs757908690 Chr.18: 49837718-49837718 on Build GRCh38
rs752193239 Chr.18: 49837709-49837709 on Build GRCh38
rs769679742 Chr.18: 49837646-49837646 on Build GRCh38
rs748864584 Chr.18: 49837548-49837548 on Build GRCh38
rs777173387 Chr.18: 49837783-49837783 on Build GRCh38
rs201466306 Chr.18: 49837701-49837701 on Build GRCh38
rs752212773 Chr.18: 49837626-49837626 on Build GRCh38
rs79271359 Chr.18: 49837560-49837560 on Build GRCh38
rs767872256 Chr.18: 49837815-49837816 on Build GRCh38
rs780999931 Chr.18: 49837734-49837734 on Build GRCh38
rs780336129 Chr.18: 49837885-49837885 on Build GRCh38
rs970576527 Chr.18: 49837884-49837884 on Build GRCh38
rs199837997 Chr.18: 49837585-49837585 on Build GRCh38
rs763012233 Chr.18: 49837493-49837493 on Build GRCh38
rs763684528 Chr.18: 49837828-49837828 on Build GRCh38
rs9950781 Chr.18: 49837607-49837607 on Build GRCh38
rs774568137 Chr.18: 49837696-49837697 on Build GRCh38
rs757415890 Chr.18: 49837500-49837500 on Build GRCh38
rs180877792 Chr.18: 49837869-49837869 on Build GRCh38
rs766862373 Chr.18: 49837604-49837604 on Build GRCh38
rs138128932 Chr.18: 49837547-49837547 on Build GRCh38
rs771235825 Chr.18: 49837780-49837780 on Build GRCh38
rs755900274 Chr.18: 49837847-49837847 on Build GRCh38
rs367710844 Chr.18: 49837637-49837637 on Build GRCh38

View Full Product Details