Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7678209-7678733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCEMP1
Entrez Gene ID
199675
Gene Name
mast cell expressed membrane protein 1
Gene Aliases
C19orf59
UniGene
Hs.709539
Gene Chromosome Location
Chr.19: 7677057-7679833 on Build GRCh38
Transcripts
NM_174918.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
440645 c.361G>C p.E121Q Chr.19: 7678388-7678388 on Build GRCh38
7411575 c.462T>G p.I154M Chr.19: 7678579-7678579 on Build GRCh38
1003345 c.397G>A p.E133K Chr.19: 7678514-7678514 on Build GRCh38
6242659 c.413G>A p.R138K Chr.19: 7678530-7678530 on Build GRCh38
4517800 c.425_426CC>TT p.S142F Chr.19: 7678542-7678543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762483146 Chr.19: 7678312-7678312 on Build GRCh38
rs760866330 Chr.19: 7678607-7678607 on Build GRCh38
rs766094609 Chr.19: 7678255-7678255 on Build GRCh38
rs769652201 Chr.19: 7678591-7678591 on Build GRCh38
rs199822833 Chr.19: 7678376-7678376 on Build GRCh38
rs773891462 Chr.19: 7678495-7678495 on Build GRCh38
rs199684838 Chr.19: 7678354-7678354 on Build GRCh38
rs376546618 Chr.19: 7678521-7678521 on Build GRCh38
rs936648983 Chr.19: 7678634-7678634 on Build GRCh38
rs767580417 Chr.19: 7678594-7678594 on Build GRCh38
rs764123797 Chr.19: 7678485-7678485 on Build GRCh38
rs12981176 Chr.19: 7678401-7678401 on Build GRCh38
rs12974784 Chr.19: 7678347-7678347 on Build GRCh38
rs780444468 Chr.19: 7678598-7678598 on Build GRCh38
rs754755999 Chr.19: 7678261-7678261 on Build GRCh38
rs754999223 Chr.19: 7678530-7678530 on Build GRCh38
rs764006691 Chr.19: 7678414-7678414 on Build GRCh38
rs770333214 Chr.19: 7678310-7678310 on Build GRCh38
rs747848411 Chr.19: 7678654-7678654 on Build GRCh38
rs372956590 Chr.19: 7678621-7678621 on Build GRCh38
rs755194931 Chr.19: 7678433-7678433 on Build GRCh38
rs146064936 Chr.19: 7678593-7678593 on Build GRCh38
rs764265206 Chr.19: 7678616-7678616 on Build GRCh38
rs770409681 Chr.19: 7678391-7678391 on Build GRCh38
rs77239495 Chr.19: 7678539-7678539 on Build GRCh38
rs1029922061 Chr.19: 7678481-7678481 on Build GRCh38
rs200077204 Chr.19: 7678508-7678508 on Build GRCh38
rs772156619 Chr.19: 7678274-7678274 on Build GRCh38
rs372401061 Chr.19: 7678272-7678272 on Build GRCh38
rs1039929808 Chr.19: 7678669-7678669 on Build GRCh38
rs186206778 Chr.19: 7678411-7678411 on Build GRCh38
rs756315122 Chr.19: 7678574-7678574 on Build GRCh38
rs773682928 Chr.19: 7678331-7678332 on Build GRCh38
rs758458851 Chr.19: 7678373-7678373 on Build GRCh38
rs376048245 Chr.19: 7678428-7678428 on Build GRCh38
rs138132511 Chr.19: 7678553-7678553 on Build GRCh38
rs147625914 Chr.19: 7678544-7678544 on Build GRCh38
rs532507573 Chr.19: 7678266-7678266 on Build GRCh38
rs768814799 Chr.19: 7678386-7678386 on Build GRCh38
rs139191722 Chr.19: 7678500-7678500 on Build GRCh38
rs576173771 Chr.19: 7678670-7678670 on Build GRCh38
rs767340251 Chr.19: 7678324-7678324 on Build GRCh38
rs377232049 Chr.19: 7678620-7678620 on Build GRCh38
rs772334591 Chr.19: 7678314-7678314 on Build GRCh38
rs766949553 Chr.19: 7678635-7678638 on Build GRCh38
rs373450354 Chr.19: 7678426-7678426 on Build GRCh38
rs753758784 Chr.19: 7678422-7678422 on Build GRCh38
rs955317321 Chr.19: 7678270-7678270 on Build GRCh38
rs74489689 Chr.19: 7678273-7678273 on Build GRCh38
rs12979223 Chr.19: 7678392-7678392 on Build GRCh38
rs750708775 Chr.19: 7678514-7678514 on Build GRCh38
rs766465194 Chr.19: 7678427-7678427 on Build GRCh38
rs756103034 Chr.19: 7678341-7678341 on Build GRCh38
rs146973085 Chr.19: 7678543-7678543 on Build GRCh38
rs748795543 Chr.19: 7678298-7678299 on Build GRCh38
rs368138357 Chr.19: 7678633-7678633 on Build GRCh38
rs368773876 Chr.19: 7678408-7678408 on Build GRCh38
rs771400396 Chr.19: 7678488-7678488 on Build GRCh38
rs752831986 Chr.19: 7678599-7678599 on Build GRCh38
rs973707826 Chr.19: 7678296-7678297 on Build GRCh38
rs531881786 Chr.19: 7678372-7678372 on Build GRCh38
rs984524186 Chr.19: 7678240-7678240 on Build GRCh38
rs372574521 Chr.19: 7678406-7678406 on Build GRCh38
rs780657987 Chr.19: 7678644-7678644 on Build GRCh38
rs774741125 Chr.19: 7678269-7678269 on Build GRCh38
rs373387576 Chr.19: 7678474-7678474 on Build GRCh38
rs138136167 Chr.19: 7678592-7678592 on Build GRCh38
rs374765264 Chr.19: 7678586-7678586 on Build GRCh38
rs779773805 Chr.19: 7678570-7678570 on Build GRCh38
rs771654814 Chr.19: 7678567-7678567 on Build GRCh38
rs12979248 Chr.19: 7678415-7678415 on Build GRCh38
rs773659150 Chr.19: 7678389-7678389 on Build GRCh38
rs984448748 Chr.19: 7678561-7678561 on Build GRCh38
rs983607945 Chr.19: 7678370-7678370 on Build GRCh38
rs748310557 Chr.19: 7678275-7678275 on Build GRCh38
rs267605762 Chr.19: 7678536-7678536 on Build GRCh38
rs147070011 Chr.19: 7678565-7678565 on Build GRCh38
rs12975052 Chr.19: 7678446-7678446 on Build GRCh38
rs778117035 Chr.19: 7678444-7678444 on Build GRCh38
rs142736681 Chr.19: 7678578-7678578 on Build GRCh38
rs757517206 Chr.19: 7678618-7678618 on Build GRCh38
rs749774425 Chr.19: 7678390-7678390 on Build GRCh38
rs1020205408 Chr.19: 7678492-7678492 on Build GRCh38
rs750404151 Chr.19: 7678523-7678523 on Build GRCh38
rs1041190356 Chr.19: 7678663-7678663 on Build GRCh38
rs12981161 Chr.19: 7678369-7678369 on Build GRCh38
rs765828480 Chr.19: 7678316-7678316 on Build GRCh38
rs751200770 Chr.19: 7678256-7678256 on Build GRCh38
rs759117586 Chr.19: 7678322-7678322 on Build GRCh38
rs747248512 Chr.19: 7678377-7678377 on Build GRCh38
rs756411400 Chr.19: 7678443-7678443 on Build GRCh38
rs1018061901 Chr.19: 7678510-7678510 on Build GRCh38
rs12971697 Chr.19: 7678358-7678358 on Build GRCh38
rs983693748 Chr.19: 7678568-7678568 on Build GRCh38
rs200784657 Chr.19: 7678407-7678407 on Build GRCh38
rs746642579 Chr.19: 7678572-7678572 on Build GRCh38
rs545267406 Chr.19: 7678695-7678695 on Build GRCh38
rs1036638862 Chr.19: 7678393-7678393 on Build GRCh38
rs12979224 Chr.19: 7678394-7678394 on Build GRCh38
rs201095007 Chr.19: 7678315-7678315 on Build GRCh38
rs995193297 Chr.19: 7678693-7678693 on Build GRCh38
rs368187849 Chr.19: 7678588-7678588 on Build GRCh38
rs12971681 Chr.19: 7678346-7678346 on Build GRCh38
rs748550906 Chr.19: 7678387-7678387 on Build GRCh38
rs768996217 Chr.19: 7678284-7678284 on Build GRCh38
rs1053860250 Chr.19: 7678438-7678438 on Build GRCh38
rs151233009 Chr.19: 7678297-7678298 on Build GRCh38
rs768309215 Chr.19: 7678638-7678638 on Build GRCh38
rs760942276 Chr.19: 7678344-7678344 on Build GRCh38
rs777902779 Chr.19: 7678367-7678367 on Build GRCh38
rs201106651 Chr.19: 7678342-7678342 on Build GRCh38
rs770175643 Chr.19: 7678388-7678388 on Build GRCh38
rs368600601 Chr.19: 7678629-7678629 on Build GRCh38
rs747206224 Chr.19: 7678281-7678281 on Build GRCh38
rs144579994 Chr.19: 7678513-7678513 on Build GRCh38
rs761784766 Chr.19: 7678519-7678519 on Build GRCh38
rs12979202 Chr.19: 7678359-7678359 on Build GRCh38
rs190620026 Chr.19: 7678674-7678674 on Build GRCh38
rs369386747 Chr.19: 7678441-7678441 on Build GRCh38
rs757755213 Chr.19: 7678563-7678563 on Build GRCh38
rs752599190 Chr.19: 7678337-7678337 on Build GRCh38
rs34383810 Chr.19: 7678527-7678528 on Build GRCh38
rs8111596 Chr.19: 7678502-7678502 on Build GRCh38
rs778100526 Chr.19: 7678549-7678549 on Build GRCh38
rs751167532 Chr.19: 7678319-7678319 on Build GRCh38
rs1028517934 Chr.19: 7678685-7678685 on Build GRCh38
rs780864358 Chr.19: 7678265-7678265 on Build GRCh38
rs767596828 Chr.19: 7678542-7678542 on Build GRCh38
rs562965704 Chr.19: 7678302-7678302 on Build GRCh38
rs950986435 Chr.19: 7678557-7678557 on Build GRCh38
rs945460592 Chr.19: 7678650-7678650 on Build GRCh38
rs780286191 Chr.19: 7678278-7678278 on Build GRCh38
rs373300914 Chr.19: 7678517-7678517 on Build GRCh38
rs11880394 Chr.19: 7678696-7678696 on Build GRCh38
rs8103654 Chr.19: 7678617-7678617 on Build GRCh38
rs770378055 Chr.19: 7678482-7678482 on Build GRCh38
rs926557981 Chr.19: 7678628-7678628 on Build GRCh38

View Full Product Details