Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8536225-8536719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO1F
Entrez Gene ID
4542
Gene Name
myosin IF
Gene Aliases
-
UniGene
Hs.465818
Gene Chromosome Location
Chr.19: 8520790-8577447 on Build GRCh38
Transcripts
NM_012335.3 XM_011528024.1 XM_011528025.1 XM_011528026.1 XM_011528027.1 XM_011528028.2 XM_017026821.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7421492 c.1953G>C p.Q651H Chr.19: 8536342-8536342 on Build GRCh38
4289030 c.1881C>T p.F627F Chr.19: 8536516-8536516 on Build GRCh38
6153342 c.2008G>A p.G670R Chr.19: 8536287-8536287 on Build GRCh38
6720261 c.1857C>T p.A619A Chr.19: 8536540-8536540 on Build GRCh38
4999831 c.1937G>T p.R646L Chr.19: 8536358-8536358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557720157 Chr.19: 8536323-8536323 on Build GRCh38
rs774066426 Chr.19: 8536372-8536372 on Build GRCh38
rs773488128 Chr.19: 8536406-8536406 on Build GRCh38
rs755109803 Chr.19: 8536450-8536450 on Build GRCh38
rs750703902 Chr.19: 8536279-8536279 on Build GRCh38
rs756136328 Chr.19: 8536683-8536683 on Build GRCh38
rs758730779 Chr.19: 8536351-8536351 on Build GRCh38
rs980092651 Chr.19: 8536662-8536662 on Build GRCh38
rs914659802 Chr.19: 8536654-8536654 on Build GRCh38
rs751355344 Chr.19: 8536246-8536246 on Build GRCh38
rs370105681 Chr.19: 8536392-8536392 on Build GRCh38
rs560090185 Chr.19: 8536606-8536606 on Build GRCh38
rs746792025 Chr.19: 8536514-8536514 on Build GRCh38
rs753914209 Chr.19: 8536269-8536269 on Build GRCh38
rs771930639 Chr.19: 8536378-8536378 on Build GRCh38
rs373627527 Chr.19: 8536404-8536404 on Build GRCh38
rs183420008 Chr.19: 8536466-8536466 on Build GRCh38
rs776718917 Chr.19: 8536515-8536515 on Build GRCh38
rs199981303 Chr.19: 8536546-8536546 on Build GRCh38
rs757654643 Chr.19: 8536568-8536568 on Build GRCh38
rs367762414 Chr.19: 8536445-8536445 on Build GRCh38
rs770548153 Chr.19: 8536367-8536367 on Build GRCh38
rs780031495 Chr.19: 8536496-8536496 on Build GRCh38
rs552152472 Chr.19: 8536245-8536245 on Build GRCh38
rs964640933 Chr.19: 8536442-8536442 on Build GRCh38
rs373362851 Chr.19: 8536302-8536302 on Build GRCh38
rs202195089 Chr.19: 8536373-8536373 on Build GRCh38
rs764376094 Chr.19: 8536564-8536564 on Build GRCh38
rs769328708 Chr.19: 8536313-8536313 on Build GRCh38
rs747364980 Chr.19: 8536303-8536303 on Build GRCh38
rs370361050 Chr.19: 8536451-8536451 on Build GRCh38
rs115875216 Chr.19: 8536473-8536473 on Build GRCh38
rs759362762 Chr.19: 8536330-8536330 on Build GRCh38
rs759457500 Chr.19: 8536253-8536253 on Build GRCh38
rs747892626 Chr.19: 8536680-8536680 on Build GRCh38
rs562451544 Chr.19: 8536651-8536651 on Build GRCh38
rs200744759 Chr.19: 8536366-8536366 on Build GRCh38
rs775694491 Chr.19: 8536635-8536635 on Build GRCh38
rs939532625 Chr.19: 8536478-8536478 on Build GRCh38
rs761041920 Chr.19: 8536552-8536552 on Build GRCh38
rs368622600 Chr.19: 8536479-8536479 on Build GRCh38
rs989309812 Chr.19: 8536328-8536328 on Build GRCh38
rs749867467 Chr.19: 8536584-8536584 on Build GRCh38
rs781175033 Chr.19: 8536674-8536674 on Build GRCh38
rs758642752 Chr.19: 8536281-8536281 on Build GRCh38
rs201325015 Chr.19: 8536534-8536534 on Build GRCh38
rs755378609 Chr.19: 8536359-8536359 on Build GRCh38
rs760648457 Chr.19: 8536346-8536346 on Build GRCh38
rs968861450 Chr.19: 8536523-8536523 on Build GRCh38
rs201138222 Chr.19: 8536319-8536319 on Build GRCh38
rs758133545 Chr.19: 8536592-8536592 on Build GRCh38
rs953917543 Chr.19: 8536550-8536550 on Build GRCh38
rs768607061 Chr.19: 8536621-8536621 on Build GRCh38
rs779097714 Chr.19: 8536276-8536276 on Build GRCh38
rs748169756 Chr.19: 8536625-8536625 on Build GRCh38
rs74768935 Chr.19: 8536452-8536452 on Build GRCh38
rs751842442 Chr.19: 8536436-8536436 on Build GRCh38
rs760740712 Chr.19: 8536400-8536400 on Build GRCh38
rs573248456 Chr.19: 8536358-8536358 on Build GRCh38
rs536899062 Chr.19: 8536339-8536339 on Build GRCh38
rs751282959 Chr.19: 8536666-8536666 on Build GRCh38
rs774505037 Chr.19: 8536464-8536464 on Build GRCh38
rs760846076 Chr.19: 8536637-8536637 on Build GRCh38
rs745445244 Chr.19: 8536374-8536374 on Build GRCh38
rs757855084 Chr.19: 8536657-8536657 on Build GRCh38
rs376655905 Chr.19: 8536311-8536311 on Build GRCh38
rs369101949 Chr.19: 8536257-8536257 on Build GRCh38
rs531360716 Chr.19: 8536684-8536684 on Build GRCh38
rs754372219 Chr.19: 8536565-8536565 on Build GRCh38
rs761619477 Chr.19: 8536516-8536516 on Build GRCh38
rs757436755 Chr.19: 8536271-8536271 on Build GRCh38
rs75948259 Chr.19: 8536467-8536467 on Build GRCh38
rs771598971 Chr.19: 8536485-8536485 on Build GRCh38
rs767816216 Chr.19: 8536683-8536683 on Build GRCh38
rs574535196 Chr.19: 8536543-8536543 on Build GRCh38
rs768467493 Chr.19: 8536449-8536449 on Build GRCh38
rs781088747 Chr.19: 8536624-8536624 on Build GRCh38
rs149369796 Chr.19: 8536650-8536650 on Build GRCh38
rs200911217 Chr.19: 8536322-8536322 on Build GRCh38
rs371339136 Chr.19: 8536347-8536347 on Build GRCh38
rs974830843 Chr.19: 8536449-8536449 on Build GRCh38
rs201646155 Chr.19: 8536350-8536350 on Build GRCh38
rs754657763 Chr.19: 8536671-8536671 on Build GRCh38
rs759522864 Chr.19: 8536629-8536629 on Build GRCh38
rs376377087 Chr.19: 8536540-8536540 on Build GRCh38
rs761736142 Chr.19: 8536631-8536674 on Build GRCh38
rs931464832 Chr.19: 8536434-8536434 on Build GRCh38
rs372596262 Chr.19: 8536472-8536472 on Build GRCh38
rs778532982 Chr.19: 8536580-8536580 on Build GRCh38
rs766306914 Chr.19: 8536418-8536418 on Build GRCh38
rs781374288 Chr.19: 8536364-8536364 on Build GRCh38
rs371681536 Chr.19: 8536626-8536626 on Build GRCh38
rs750003470 Chr.19: 8536655-8536655 on Build GRCh38
rs763107534 Chr.19: 8536410-8536410 on Build GRCh38
rs766616303 Chr.19: 8536354-8536354 on Build GRCh38
rs769105319 Chr.19: 8536308-8536308 on Build GRCh38
rs771853880 Chr.19: 8536318-8536318 on Build GRCh38
rs754129586 Chr.19: 8536642-8536642 on Build GRCh38
rs748591477 Chr.19: 8536312-8536312 on Build GRCh38
rs772082774 Chr.19: 8536632-8536632 on Build GRCh38
rs373423000 Chr.19: 8536365-8536365 on Build GRCh38
rs779675539 Chr.19: 8536658-8536658 on Build GRCh38
rs746597787 Chr.19: 8536611-8536611 on Build GRCh38
rs762522663 Chr.19: 8536244-8536244 on Build GRCh38
rs199663368 Chr.19: 8536338-8536338 on Build GRCh38
rs1031470726 Chr.19: 8536326-8536326 on Build GRCh38
rs771067950 Chr.19: 8536526-8536526 on Build GRCh38
rs532204636 Chr.19: 8536627-8536627 on Build GRCh38
rs764464219 Chr.19: 8536641-8536641 on Build GRCh38

View Full Product Details