Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10137749-10138258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT1
Entrez Gene ID
1786
Gene Name
DNA (cytosine-5-)-methyltransferase 1
Gene Aliases
ADCADN, AIM, CXXC9, DNMT, HSN1E, MCMT, m.HsaI
UniGene
Hs.202672
Gene Chromosome Location
Chr.19: 10133344-10195135 on Build GRCh38
Transcripts
NM_001130823.2 NM_001318730.1 NM_001318731.1 NM_001379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3148753 c.4201C>T p.R1401W Chr.19: 10137876-10137876 on Build GRCh38
4909597 c.4129G>A p.V1377M Chr.19: 10137948-10137948 on Build GRCh38
3148755 c.4133G>A p.R1378Q Chr.19: 10137944-10137944 on Build GRCh38
3528093 c.4150C>T p.L1384L Chr.19: 10137927-10137927 on Build GRCh38
5828755 c.4194G>A p.S1398S Chr.19: 10137931-10137931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759807018 Chr.19: 10137898-10137898 on Build GRCh38
rs552378111 Chr.19: 10138121-10138121 on Build GRCh38
rs921273595 Chr.19: 10138111-10138111 on Build GRCh38
rs1046789640 Chr.19: 10138098-10138098 on Build GRCh38
rs777085751 Chr.19: 10137940-10137940 on Build GRCh38
rs912708995 Chr.19: 10138092-10138092 on Build GRCh38
rs181952514 Chr.19: 10137799-10137799 on Build GRCh38
rs773693230 Chr.19: 10137982-10137982 on Build GRCh38
rs750205723 Chr.19: 10137812-10137813 on Build GRCh38
rs749741623 Chr.19: 10137985-10137985 on Build GRCh38
rs1046758700 Chr.19: 10137921-10137921 on Build GRCh38
rs763415829 Chr.19: 10138073-10138073 on Build GRCh38
rs539093112 Chr.19: 10138100-10138100 on Build GRCh38
rs959974113 Chr.19: 10137958-10137958 on Build GRCh38
rs1030941600 Chr.19: 10138038-10138038 on Build GRCh38
rs745946775 Chr.19: 10138083-10138084 on Build GRCh38
rs550868730 Chr.19: 10137952-10137952 on Build GRCh38
rs374484590 Chr.19: 10137800-10137800 on Build GRCh38
rs886054127 Chr.19: 10137921-10137921 on Build GRCh38
rs368682710 Chr.19: 10138209-10138209 on Build GRCh38
rs75357496 Chr.19: 10138213-10138213 on Build GRCh38
rs565037285 Chr.19: 10137777-10137777 on Build GRCh38
rs778374437 Chr.19: 10137946-10137946 on Build GRCh38
rs765191061 Chr.19: 10138055-10138055 on Build GRCh38
rs371795281 Chr.19: 10138048-10138048 on Build GRCh38
rs749889229 Chr.19: 10138059-10138059 on Build GRCh38
rs1055911134 Chr.19: 10138116-10138116 on Build GRCh38
rs766558026 Chr.19: 10137889-10137889 on Build GRCh38
rs745507446 Chr.19: 10137818-10137818 on Build GRCh38
rs374196844 Chr.19: 10137778-10137778 on Build GRCh38
rs771611874 Chr.19: 10138002-10138002 on Build GRCh38
rs1039977072 Chr.19: 10138107-10138107 on Build GRCh38
rs999843707 Chr.19: 10138217-10138217 on Build GRCh38
rs911023378 Chr.19: 10137810-10137810 on Build GRCh38
rs1018815357 Chr.19: 10138233-10138233 on Build GRCh38
rs748486662 Chr.19: 10137974-10137974 on Build GRCh38
rs988350711 Chr.19: 10138088-10138088 on Build GRCh38
rs770284928 Chr.19: 10137867-10137867 on Build GRCh38
rs1056996881 Chr.19: 10138185-10138185 on Build GRCh38
rs767793246 Chr.19: 10138021-10138021 on Build GRCh38
rs764380927 Chr.19: 10138126-10138126 on Build GRCh38
rs1012790442 Chr.19: 10138204-10138204 on Build GRCh38
rs976612002 Chr.19: 10138079-10138079 on Build GRCh38
rs138068477 Chr.19: 10138033-10138033 on Build GRCh38
rs367897930 Chr.19: 10137904-10137904 on Build GRCh38
rs149568166 Chr.19: 10138218-10138218 on Build GRCh38
rs922546844 Chr.19: 10138089-10138089 on Build GRCh38
rs774538556 Chr.19: 10138004-10138004 on Build GRCh38
rs757915237 Chr.19: 10138060-10138060 on Build GRCh38
rs887221164 Chr.19: 10138152-10138152 on Build GRCh38
rs759717920 Chr.19: 10138006-10138006 on Build GRCh38
rs1004689699 Chr.19: 10138168-10138168 on Build GRCh38
rs570668263 Chr.19: 10138125-10138125 on Build GRCh38
rs764029022 Chr.19: 10137928-10137928 on Build GRCh38
rs765505020 Chr.19: 10138071-10138071 on Build GRCh38
rs374047326 Chr.19: 10137870-10137870 on Build GRCh38
rs771761767 Chr.19: 10137820-10137820 on Build GRCh38
rs375225009 Chr.19: 10137932-10137932 on Build GRCh38
rs941095015 Chr.19: 10138108-10138108 on Build GRCh38
rs184125970 Chr.19: 10137979-10137979 on Build GRCh38
rs767263593 Chr.19: 10137800-10137802 on Build GRCh38
rs199506146 Chr.19: 10137827-10137827 on Build GRCh38
rs1022804378 Chr.19: 10138208-10138208 on Build GRCh38
rs753716290 Chr.19: 10138046-10138046 on Build GRCh38
rs758009053 Chr.19: 10137955-10137955 on Build GRCh38
rs773672940 Chr.19: 10138072-10138072 on Build GRCh38
rs756726375 Chr.19: 10137806-10137806 on Build GRCh38
rs757243017 Chr.19: 10137944-10137944 on Build GRCh38
rs895648436 Chr.19: 10138186-10138186 on Build GRCh38
rs942646868 Chr.19: 10137811-10137811 on Build GRCh38
rs1004025205 Chr.19: 10137874-10137874 on Build GRCh38
rs778466456 Chr.19: 10137812-10137812 on Build GRCh38
rs779701784 Chr.19: 10137961-10137961 on Build GRCh38
rs1048439128 Chr.19: 10137824-10137824 on Build GRCh38
rs748494068 Chr.19: 10137847-10137847 on Build GRCh38
rs755878403 Chr.19: 10137922-10137922 on Build GRCh38
rs767889968 Chr.19: 10137910-10137910 on Build GRCh38
rs530146603 Chr.19: 10138031-10138031 on Build GRCh38
rs570354490 Chr.19: 10138079-10138079 on Build GRCh38
rs775700127 Chr.19: 10138025-10138025 on Build GRCh38
rs368325985 Chr.19: 10137967-10137967 on Build GRCh38
rs752490174 Chr.19: 10137913-10137913 on Build GRCh38
rs774626723 Chr.19: 10137895-10137895 on Build GRCh38

View Full Product Details