Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10287399-10287896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICAM4
Entrez Gene ID
3386
Gene Name
intercellular adhesion molecule 4 (Landsteiner-Wiener blood group)
Gene Aliases
CD242, LW
UniGene
Hs.706750
Gene Chromosome Location
Chr.19: 10286967-10288584 on Build GRCh38
Transcripts
NM_001039132.2 NM_001544.4 NM_022377.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4073779 c.538C>A p.L180M Chr.19: 10287679-10287679 on Build GRCh38
710252 c.482G>T p.G161V Chr.19: 10287700-10287700 on Build GRCh38
6683527 c.394+2T>C p.? Chr.19: 10287408-10287408 on Build GRCh38
3403706 c.474C>G p.H158Q Chr.19: 10287692-10287692 on Build GRCh38
5493817 c.570C>T p.N190N Chr.19: 10287711-10287711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544219094 Chr.19: 10287710-10287710 on Build GRCh38
rs542450359 Chr.19: 10287501-10287501 on Build GRCh38
rs373009181 Chr.19: 10287514-10287514 on Build GRCh38
rs755583690 Chr.19: 10287715-10287715 on Build GRCh38
rs755759895 Chr.19: 10287566-10287566 on Build GRCh38
rs758847209 Chr.19: 10287685-10287685 on Build GRCh38
rs757827547 Chr.19: 10287438-10287452 on Build GRCh38
rs372453498 Chr.19: 10287686-10287686 on Build GRCh38
rs780710490 Chr.19: 10287591-10287591 on Build GRCh38
rs374457779 Chr.19: 10287535-10287535 on Build GRCh38
rs757624505 Chr.19: 10287450-10287450 on Build GRCh38
rs779310858 Chr.19: 10287567-10287567 on Build GRCh38
rs759533488 Chr.19: 10287629-10287629 on Build GRCh38
rs142109230 Chr.19: 10287789-10287789 on Build GRCh38
rs1021723224 Chr.19: 10287748-10287748 on Build GRCh38
rs377470882 Chr.19: 10287641-10287641 on Build GRCh38
rs1034069627 Chr.19: 10287673-10287673 on Build GRCh38
rs758686589 Chr.19: 10287687-10287687 on Build GRCh38
rs777970931 Chr.19: 10287856-10287856 on Build GRCh38
rs924104082 Chr.19: 10287489-10287489 on Build GRCh38
rs909829363 Chr.19: 10287497-10287497 on Build GRCh38
rs769960980 Chr.19: 10287604-10287604 on Build GRCh38
rs907035521 Chr.19: 10287649-10287649 on Build GRCh38
rs982593085 Chr.19: 10287493-10287493 on Build GRCh38
rs368901409 Chr.19: 10287496-10287496 on Build GRCh38
rs776924459 Chr.19: 10287512-10287512 on Build GRCh38
rs147458509 Chr.19: 10287554-10287554 on Build GRCh38
rs748372509 Chr.19: 10287603-10287603 on Build GRCh38
rs372395338 Chr.19: 10287418-10287418 on Build GRCh38
rs746352738 Chr.19: 10287456-10287456 on Build GRCh38
rs767069162 Chr.19: 10287549-10287549 on Build GRCh38
rs772921849 Chr.19: 10287529-10287529 on Build GRCh38
rs755494782 Chr.19: 10287835-10287835 on Build GRCh38
rs761626067 Chr.19: 10287829-10287829 on Build GRCh38
rs769629691 Chr.19: 10287523-10287523 on Build GRCh38
rs1034536330 Chr.19: 10287439-10287439 on Build GRCh38
rs780622559 Chr.19: 10287694-10287694 on Build GRCh38
rs780857348 Chr.19: 10287438-10287438 on Build GRCh38
rs879218941 Chr.19: 10287466-10287466 on Build GRCh38
rs751121948 Chr.19: 10287428-10287428 on Build GRCh38
rs36023325 Chr.19: 10287763-10287763 on Build GRCh38
rs570743605 Chr.19: 10287421-10287421 on Build GRCh38
rs775023987 Chr.19: 10287794-10287794 on Build GRCh38
rs976836360 Chr.19: 10287469-10287469 on Build GRCh38
rs747026880 Chr.19: 10287594-10287594 on Build GRCh38
rs760105445 Chr.19: 10287795-10287795 on Build GRCh38
rs758337091 Chr.19: 10287427-10287427 on Build GRCh38
rs776408886 Chr.19: 10287812-10287812 on Build GRCh38
rs576131531 Chr.19: 10287612-10287612 on Build GRCh38
rs951211455 Chr.19: 10287484-10287484 on Build GRCh38
rs766820266 Chr.19: 10287832-10287832 on Build GRCh38
rs1018903753 Chr.19: 10287788-10287788 on Build GRCh38
rs35165411 Chr.19: 10287530-10287530 on Build GRCh38
rs768844056 Chr.19: 10287595-10287595 on Build GRCh38
rs750773201 Chr.19: 10287578-10287578 on Build GRCh38
rs759055313 Chr.19: 10287544-10287544 on Build GRCh38
rs934200316 Chr.19: 10287490-10287490 on Build GRCh38
rs766979276 Chr.19: 10287631-10287631 on Build GRCh38
rs745781332 Chr.19: 10287773-10287773 on Build GRCh38
rs145697899 Chr.19: 10287723-10287723 on Build GRCh38
rs758932037 Chr.19: 10287585-10287585 on Build GRCh38
rs774288442 Chr.19: 10287626-10287626 on Build GRCh38
rs199887173 Chr.19: 10287423-10287423 on Build GRCh38
rs750731494 Chr.19: 10287679-10287679 on Build GRCh38
rs372936502 Chr.19: 10287819-10287819 on Build GRCh38
rs958384610 Chr.19: 10287447-10287447 on Build GRCh38
rs968273159 Chr.19: 10287463-10287463 on Build GRCh38
rs748201755 Chr.19: 10287733-10287733 on Build GRCh38
rs767944077 Chr.19: 10287836-10287836 on Build GRCh38
rs371076614 Chr.19: 10287531-10287531 on Build GRCh38
rs375526250 Chr.19: 10287487-10287487 on Build GRCh38
rs377604730 Chr.19: 10287842-10287842 on Build GRCh38
rs145323966 Chr.19: 10287754-10287754 on Build GRCh38
rs770792909 Chr.19: 10287620-10287620 on Build GRCh38
rs145682448 Chr.19: 10287430-10287430 on Build GRCh38
rs749477052 Chr.19: 10287859-10287859 on Build GRCh38
rs34385135 Chr.19: 10287455-10287455 on Build GRCh38
rs915579459 Chr.19: 10287582-10287582 on Build GRCh38
rs371910762 Chr.19: 10287658-10287658 on Build GRCh38
rs781614874 Chr.19: 10287540-10287541 on Build GRCh38
rs151158164 Chr.19: 10287805-10287805 on Build GRCh38
rs781196046 Chr.19: 10287599-10287599 on Build GRCh38
rs764431950 Chr.19: 10287420-10287420 on Build GRCh38
rs757516126 Chr.19: 10287860-10287860 on Build GRCh38
rs902919648 Chr.19: 10287651-10287651 on Build GRCh38
rs749028766 Chr.19: 10287762-10287762 on Build GRCh38
rs200471341 Chr.19: 10287650-10287650 on Build GRCh38
rs756322389 Chr.19: 10287745-10287745 on Build GRCh38
rs751034035 Chr.19: 10287536-10287536 on Build GRCh38
rs924234867 Chr.19: 10287820-10287820 on Build GRCh38
rs571059786 Chr.19: 10287838-10287838 on Build GRCh38
rs749551837 Chr.19: 10287443-10287443 on Build GRCh38
rs774167545 Chr.19: 10287764-10287764 on Build GRCh38
rs978196401 Chr.19: 10287483-10287483 on Build GRCh38
rs1038797401 Chr.19: 10287579-10287579 on Build GRCh38
rs898390193 Chr.19: 10287630-10287630 on Build GRCh38
rs375673380 Chr.19: 10287768-10287768 on Build GRCh38
rs540595516 Chr.19: 10287834-10287834 on Build GRCh38

View Full Product Details