Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39200537-39201040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCCRP1
Entrez Gene ID
342897
Gene Name
non-specific cytotoxic cell receptor protein 1 homolog (zebrafish)
Gene Aliases
FBXO50, NCCRP-1
UniGene
Hs.726934
Gene Chromosome Location
Chr.19: 39196964-39201882 on Build GRCh38
Transcripts
NM_001001414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4077915 c.719G>A p.G240D Chr.19: 39200647-39200647 on Build GRCh38
4077916 c.779G>A p.R260Q Chr.19: 39200707-39200707 on Build GRCh38
6449298 c.758G>A p.R253H Chr.19: 39200686-39200686 on Build GRCh38
3990006 c.796G>A p.D266N Chr.19: 39200724-39200724 on Build GRCh38
6479265 c.718G>C p.G240R Chr.19: 39200646-39200646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917119752 Chr.19: 39200709-39200709 on Build GRCh38
rs578023720 Chr.19: 39200758-39200758 on Build GRCh38
rs764518747 Chr.19: 39200940-39200940 on Build GRCh38
rs201348392 Chr.19: 39200779-39200779 on Build GRCh38
rs200152362 Chr.19: 39200732-39200732 on Build GRCh38
rs762093413 Chr.19: 39200803-39200803 on Build GRCh38
rs140808558 Chr.19: 39200568-39200568 on Build GRCh38
rs138922486 Chr.19: 39200723-39200723 on Build GRCh38
rs757283881 Chr.19: 39200646-39200646 on Build GRCh38
rs747046037 Chr.19: 39200586-39200586 on Build GRCh38
rs779986852 Chr.19: 39200724-39200724 on Build GRCh38
rs376755212 Chr.19: 39200613-39200613 on Build GRCh38
rs760186843 Chr.19: 39200707-39200707 on Build GRCh38
rs970352094 Chr.19: 39200943-39200943 on Build GRCh38
rs781302321 Chr.19: 39200718-39200718 on Build GRCh38
rs778542772 Chr.19: 39200733-39200733 on Build GRCh38
rs985047474 Chr.19: 39200882-39200882 on Build GRCh38
rs150676477 Chr.19: 39200979-39200979 on Build GRCh38
rs141616015 Chr.19: 39200969-39200969 on Build GRCh38
rs202157094 Chr.19: 39200598-39200598 on Build GRCh38
rs763308091 Chr.19: 39200925-39200925 on Build GRCh38
rs550666456 Chr.19: 39200566-39200566 on Build GRCh38
rs113281917 Chr.19: 39200905-39200905 on Build GRCh38
rs1011855765 Chr.19: 39200560-39200560 on Build GRCh38
rs903032567 Chr.19: 39200741-39200741 on Build GRCh38
rs371735275 Chr.19: 39200645-39200645 on Build GRCh38
rs767016876 Chr.19: 39200619-39200619 on Build GRCh38
rs776505034 Chr.19: 39200591-39200591 on Build GRCh38
rs371951876 Chr.19: 39200872-39200872 on Build GRCh38
rs775337353 Chr.19: 39200603-39200603 on Build GRCh38
rs776023888 Chr.19: 39200793-39200793 on Build GRCh38
rs113786921 Chr.19: 39200748-39200748 on Build GRCh38
rs763326657 Chr.19: 39200715-39200715 on Build GRCh38
rs749536977 Chr.19: 39200630-39200632 on Build GRCh38
rs938461131 Chr.19: 39200933-39200933 on Build GRCh38
rs1031422463 Chr.19: 39200810-39200810 on Build GRCh38
rs769710252 Chr.19: 39200653-39200653 on Build GRCh38
rs780196086 Chr.19: 39200584-39200585 on Build GRCh38
rs776552275 Chr.19: 39200895-39200895 on Build GRCh38
rs367555888 Chr.19: 39200616-39200616 on Build GRCh38
rs928399855 Chr.19: 39200908-39200908 on Build GRCh38
rs768654637 Chr.19: 39200685-39200712 on Build GRCh38
rs771837045 Chr.19: 39200751-39200751 on Build GRCh38
rs181179662 Chr.19: 39200976-39200976 on Build GRCh38
rs749048848 Chr.19: 39200663-39200663 on Build GRCh38
rs940386731 Chr.19: 39200686-39200686 on Build GRCh38
rs953716628 Chr.19: 39200856-39200856 on Build GRCh38
rs544369012 Chr.19: 39200716-39200716 on Build GRCh38
rs372470395 Chr.19: 39200717-39200717 on Build GRCh38
rs772826705 Chr.19: 39200706-39200706 on Build GRCh38
rs772853224 Chr.19: 39200752-39200752 on Build GRCh38
rs376752911 Chr.19: 39200652-39200652 on Build GRCh38
rs763794783 Chr.19: 39200608-39200608 on Build GRCh38
rs555600339 Chr.19: 39200658-39200658 on Build GRCh38
rs955782385 Chr.19: 39200831-39200831 on Build GRCh38
rs761440833 Chr.19: 39200673-39200673 on Build GRCh38
rs781215125 Chr.19: 39200647-39200647 on Build GRCh38
rs78285559 Chr.19: 39200567-39200567 on Build GRCh38
rs750500854 Chr.19: 39200719-39200719 on Build GRCh38
rs1031084289 Chr.19: 39200749-39200749 on Build GRCh38
rs909815229 Chr.19: 39200961-39200961 on Build GRCh38
rs759280206 Chr.19: 39200945-39200945 on Build GRCh38
rs138608219 Chr.19: 39200894-39200894 on Build GRCh38
rs770870612 Chr.19: 39200590-39200590 on Build GRCh38
rs370440185 Chr.19: 39200759-39200759 on Build GRCh38
rs199509879 Chr.19: 39200800-39200800 on Build GRCh38
rs934518909 Chr.19: 39201007-39201007 on Build GRCh38
rs754358994 Chr.19: 39200624-39200624 on Build GRCh38
rs747852321 Chr.19: 39200738-39200738 on Build GRCh38
rs764565333 Chr.19: 39200802-39200802 on Build GRCh38
rs142659092 Chr.19: 39200685-39200685 on Build GRCh38
rs1044410884 Chr.19: 39200561-39200561 on Build GRCh38
rs760512302 Chr.19: 39200710-39200710 on Build GRCh38
rs768209885 Chr.19: 39200668-39200668 on Build GRCh38
rs997896027 Chr.19: 39200786-39200786 on Build GRCh38
rs560285072 Chr.19: 39200941-39200941 on Build GRCh38
rs11672475 Chr.19: 39200576-39200576 on Build GRCh38
rs774008978 Chr.19: 39200671-39200671 on Build GRCh38
rs754266995 Chr.19: 39200565-39200565 on Build GRCh38
rs1007159656 Chr.19: 39200753-39200753 on Build GRCh38
rs867483239 Chr.19: 39200762-39200762 on Build GRCh38
rs367877232 Chr.19: 39200625-39200625 on Build GRCh38
rs765570355 Chr.19: 39200631-39200631 on Build GRCh38
rs762567731 Chr.19: 39200604-39200604 on Build GRCh38
rs11672479 Chr.19: 39200599-39200599 on Build GRCh38
rs377645422 Chr.19: 39200643-39200643 on Build GRCh38
rs774535300 Chr.19: 39200632-39200632 on Build GRCh38
rs144574051 Chr.19: 39200577-39200577 on Build GRCh38

View Full Product Details