Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39308022-39308527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRFN1
Entrez Gene ID
57622
Gene Name
leucine rich repeat and fibronectin type III domain containing 1
Gene Aliases
SALM2
UniGene
Hs.97860
Gene Chromosome Location
Chr.19: 39306817-39320867 on Build GRCh38
Transcripts
NM_020862.1 XM_005259102.4 XM_017027033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7375413 c.1684G>A p.E562K Chr.19: 39308121-39308121 on Build GRCh38
5474149 c.1529G>A p.R510H Chr.19: 39308276-39308276 on Build GRCh38
4430455 c.1740C>T p.A580A Chr.19: 39308065-39308065 on Build GRCh38
5574731 c.1687G>T p.V563L Chr.19: 39308118-39308118 on Build GRCh38
3223571 c.1513G>A p.V505I Chr.19: 39308292-39308292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773251875 Chr.19: 39308248-39308248 on Build GRCh38
rs749689228 Chr.19: 39308243-39308243 on Build GRCh38
rs1017576746 Chr.19: 39308482-39308482 on Build GRCh38
rs929481940 Chr.19: 39308338-39308338 on Build GRCh38
rs1052960227 Chr.19: 39308402-39308402 on Build GRCh38
rs777414968 Chr.19: 39308222-39308222 on Build GRCh38
rs777928614 Chr.19: 39308367-39308367 on Build GRCh38
rs746307312 Chr.19: 39308101-39308101 on Build GRCh38
rs973684154 Chr.19: 39308496-39308496 on Build GRCh38
rs760650091 Chr.19: 39308170-39308170 on Build GRCh38
rs963629336 Chr.19: 39308490-39308490 on Build GRCh38
rs199820258 Chr.19: 39308241-39308241 on Build GRCh38
rs191440882 Chr.19: 39308453-39308453 on Build GRCh38
rs770120908 Chr.19: 39308106-39308106 on Build GRCh38
rs879184184 Chr.19: 39308494-39308494 on Build GRCh38
rs919389763 Chr.19: 39308276-39308276 on Build GRCh38
rs753635789 Chr.19: 39308174-39308174 on Build GRCh38
rs371594150 Chr.19: 39308437-39308437 on Build GRCh38
rs890514191 Chr.19: 39308459-39308459 on Build GRCh38
rs755458726 Chr.19: 39308435-39308435 on Build GRCh38
rs556422711 Chr.19: 39308372-39308372 on Build GRCh38
rs538074300 Chr.19: 39308324-39308324 on Build GRCh38
rs757928159 Chr.19: 39308092-39308092 on Build GRCh38
rs762798779 Chr.19: 39308396-39308396 on Build GRCh38
rs753089255 Chr.19: 39308447-39308447 on Build GRCh38
rs1005464936 Chr.19: 39308133-39308133 on Build GRCh38
rs904451759 Chr.19: 39308375-39308375 on Build GRCh38
rs760567296 Chr.19: 39308256-39308256 on Build GRCh38
rs763863575 Chr.19: 39308411-39308411 on Build GRCh38
rs550097387 Chr.19: 39308077-39308077 on Build GRCh38
rs759386064 Chr.19: 39308381-39308381 on Build GRCh38
rs778065170 Chr.19: 39308450-39308450 on Build GRCh38
rs61730568 Chr.19: 39308113-39308113 on Build GRCh38
rs544649712 Chr.19: 39308458-39308458 on Build GRCh38
rs779239580 Chr.19: 39308345-39308345 on Build GRCh38
rs916986393 Chr.19: 39308365-39308365 on Build GRCh38
rs767469727 Chr.19: 39308045-39308045 on Build GRCh38
rs766193622 Chr.19: 39308068-39308068 on Build GRCh38
rs776379708 Chr.19: 39308266-39308266 on Build GRCh38
rs551770525 Chr.19: 39308140-39308140 on Build GRCh38
rs747130303 Chr.19: 39308368-39308368 on Build GRCh38
rs775401485 Chr.19: 39308392-39308392 on Build GRCh38
rs748366703 Chr.19: 39308246-39308246 on Build GRCh38
rs1022917953 Chr.19: 39308413-39308413 on Build GRCh38
rs770928915 Chr.19: 39308261-39308261 on Build GRCh38
rs781117727 Chr.19: 39308481-39308481 on Build GRCh38
rs536686948 Chr.19: 39308086-39308086 on Build GRCh38
rs768957461 Chr.19: 39308138-39308138 on Build GRCh38
rs748420076 Chr.19: 39308350-39308350 on Build GRCh38
rs745866127 Chr.19: 39308489-39308489 on Build GRCh38
rs200709133 Chr.19: 39308096-39308096 on Build GRCh38
rs750303056 Chr.19: 39308052-39308052 on Build GRCh38
rs776635148 Chr.19: 39308376-39308376 on Build GRCh38
rs369371960 Chr.19: 39308434-39308434 on Build GRCh38
rs751023716 Chr.19: 39308231-39308231 on Build GRCh38
rs758709899 Chr.19: 39308449-39308449 on Build GRCh38
rs888363423 Chr.19: 39308122-39308122 on Build GRCh38
rs555643766 Chr.19: 39308244-39308244 on Build GRCh38
rs772248340 Chr.19: 39308354-39308354 on Build GRCh38
rs767519157 Chr.19: 39308159-39308159 on Build GRCh38
rs372557671 Chr.19: 39308292-39308292 on Build GRCh38
rs760498025 Chr.19: 39308170-39308171 on Build GRCh38
rs1017881129 Chr.19: 39308205-39308205 on Build GRCh38
rs779112582 Chr.19: 39308245-39308245 on Build GRCh38
rs769767520 Chr.19: 39308391-39308391 on Build GRCh38
rs755908691 Chr.19: 39308053-39308053 on Build GRCh38
rs762680130 Chr.19: 39308274-39308274 on Build GRCh38
rs752335634 Chr.19: 39308204-39308204 on Build GRCh38
rs756616593 Chr.19: 39308234-39308234 on Build GRCh38
rs867760333 Chr.19: 39308225-39308225 on Build GRCh38
rs375124004 Chr.19: 39308304-39308304 on Build GRCh38
rs948564903 Chr.19: 39308099-39308099 on Build GRCh38
rs756674066 Chr.19: 39308303-39308303 on Build GRCh38
rs375598950 Chr.19: 39308270-39308270 on Build GRCh38
rs35681395 Chr.19: 39308075-39308076 on Build GRCh38
rs766936007 Chr.19: 39308427-39308427 on Build GRCh38
rs778326094 Chr.19: 39308081-39308081 on Build GRCh38
rs1044015972 Chr.19: 39308374-39308374 on Build GRCh38
rs368555768 Chr.19: 39308277-39308277 on Build GRCh38
rs766405954 Chr.19: 39308188-39308190 on Build GRCh38
rs753490764 Chr.19: 39308075-39308075 on Build GRCh38
rs752462602 Chr.19: 39308273-39308273 on Build GRCh38
rs773817761 Chr.19: 39308418-39308418 on Build GRCh38
rs764964582 Chr.19: 39308197-39308197 on Build GRCh38
rs775520420 Chr.19: 39308498-39308498 on Build GRCh38
rs566825325 Chr.19: 39308221-39308221 on Build GRCh38
rs746474635 Chr.19: 39308150-39308150 on Build GRCh38
rs772199296 Chr.19: 39308247-39308247 on Build GRCh38
rs866085342 Chr.19: 39308456-39308456 on Build GRCh38
rs1036913589 Chr.19: 39308155-39308155 on Build GRCh38
rs761894549 Chr.19: 39308141-39308141 on Build GRCh38
rs761391871 Chr.19: 39308423-39308423 on Build GRCh38
rs769885912 Chr.19: 39308495-39308495 on Build GRCh38
rs759210358 Chr.19: 39308184-39308184 on Build GRCh38
rs934324238 Chr.19: 39308054-39308054 on Build GRCh38
rs973568625 Chr.19: 39308224-39308224 on Build GRCh38

View Full Product Details