Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 51911423-51911964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB3B
Entrez Gene ID
5865
Gene Name
RAB3B, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.123072
Gene Chromosome Location
Chr.1: 51907956-51990764 on Build GRCh38
Transcripts
NM_002867.3

SNP Detail

SNP ID Chromosome Location
rs144922366 Chr.1: 51911771-51911771 on Build GRCh38
rs1035386656 Chr.1: 51911841-51911841 on Build GRCh38
rs922910712 Chr.1: 51911532-51911532 on Build GRCh38
rs781768727 Chr.1: 51911529-51911530 on Build GRCh38
rs530283336 Chr.1: 51911638-51911638 on Build GRCh38
rs963287182 Chr.1: 51911860-51911860 on Build GRCh38
rs1037538290 Chr.1: 51911557-51911557 on Build GRCh38
rs548198656 Chr.1: 51911654-51911654 on Build GRCh38
rs907774671 Chr.1: 51911539-51911539 on Build GRCh38
rs41293263 Chr.1: 51911855-51911855 on Build GRCh38
rs993419405 Chr.1: 51911743-51911743 on Build GRCh38
rs751314682 Chr.1: 51911906-51911906 on Build GRCh38
rs1049853303 Chr.1: 51911630-51911630 on Build GRCh38
rs925557628 Chr.1: 51911877-51911877 on Build GRCh38
rs543213388 Chr.1: 51911887-51911888 on Build GRCh38
rs964443521 Chr.1: 51911845-51911845 on Build GRCh38
rs1055288438 Chr.1: 51911913-51911913 on Build GRCh38
rs994177554 Chr.1: 51911930-51911930 on Build GRCh38
rs765374335 Chr.1: 51911615-51911615 on Build GRCh38
rs1001209441 Chr.1: 51911790-51911790 on Build GRCh38
rs1013622638 Chr.1: 51911629-51911629 on Build GRCh38
rs975706741 Chr.1: 51911492-51911492 on Build GRCh38
rs959812285 Chr.1: 51911849-51911849 on Build GRCh38
rs560079087 Chr.1: 51911692-51911692 on Build GRCh38
rs952010570 Chr.1: 51911777-51911777 on Build GRCh38
rs756992850 Chr.1: 51911923-51911923 on Build GRCh38
rs748554498 Chr.1: 51911484-51911484 on Build GRCh38
rs973735561 Chr.1: 51911873-51911873 on Build GRCh38
rs904027543 Chr.1: 51911544-51911544 on Build GRCh38
rs370446493 Chr.1: 51911637-51911637 on Build GRCh38
rs970206907 Chr.1: 51911773-51911773 on Build GRCh38
rs1012279768 Chr.1: 51911738-51911738 on Build GRCh38
rs187467736 Chr.1: 51911489-51911489 on Build GRCh38
rs921764219 Chr.1: 51911600-51911600 on Build GRCh38
rs921759587 Chr.1: 51911915-51911915 on Build GRCh38
rs527815977 Chr.1: 51911702-51911702 on Build GRCh38
rs764228776 Chr.1: 51911884-51911884 on Build GRCh38
rs892038993 Chr.1: 51911723-51911723 on Build GRCh38
rs371975407 Chr.1: 51911733-51911733 on Build GRCh38
rs1051619709 Chr.1: 51911925-51911925 on Build GRCh38
rs975781916 Chr.1: 51911848-51911848 on Build GRCh38
rs936847355 Chr.1: 51911588-51911588 on Build GRCh38
rs895215813 Chr.1: 51911618-51911618 on Build GRCh38
rs907575363 Chr.1: 51911856-51911856 on Build GRCh38
rs983104132 Chr.1: 51911538-51911538 on Build GRCh38
rs891107507 Chr.1: 51911662-51911662 on Build GRCh38
rs912636993 Chr.1: 51911926-51911926 on Build GRCh38
rs984621794 Chr.1: 51911825-51911825 on Build GRCh38
rs941978798 Chr.1: 51911547-51911547 on Build GRCh38

View Full Product Details