Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51009103-51009610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLK9
Entrez Gene ID
284366
Gene Name
kallikrein related peptidase 9
Gene Aliases
KLK-L3, KLKL3
UniGene
Hs.448942
Gene Chromosome Location
Chr.19: 51002513-51009634 on Build GRCh38
Transcripts
NM_012315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7374239 c.179C>T p.T60I Chr.19: 51009204-51009204 on Build GRCh38
5981596 c.29T>G p.L10R Chr.19: 51009519-51009519 on Build GRCh38
7348858 c.199C>G p.P67A Chr.19: 51009184-51009184 on Build GRCh38
1291004 c.194G>A p.R65H Chr.19: 51009189-51009189 on Build GRCh38
5856483 c.145G>A p.G49R Chr.19: 51009238-51009238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976086419 Chr.19: 51009218-51009218 on Build GRCh38
rs773264106 Chr.19: 51009155-51009155 on Build GRCh38
rs781576826 Chr.19: 51009147-51009147 on Build GRCh38
rs752120360 Chr.19: 51009333-51009333 on Build GRCh38
rs772772059 Chr.19: 51009358-51009358 on Build GRCh38
rs558922127 Chr.19: 51009353-51009353 on Build GRCh38
rs990710207 Chr.19: 51009265-51009265 on Build GRCh38
rs201034459 Chr.19: 51009582-51009582 on Build GRCh38
rs755325343 Chr.19: 51009138-51009138 on Build GRCh38
rs199945979 Chr.19: 51009318-51009318 on Build GRCh38
rs1012180946 Chr.19: 51009312-51009312 on Build GRCh38
rs201875196 Chr.19: 51009491-51009491 on Build GRCh38
rs75072873 Chr.19: 51009446-51009446 on Build GRCh38
rs576163713 Chr.19: 51009294-51009294 on Build GRCh38
rs750087926 Chr.19: 51009297-51009297 on Build GRCh38
rs572415127 Chr.19: 51009169-51009169 on Build GRCh38
rs986790958 Chr.19: 51009404-51009404 on Build GRCh38
rs749577323 Chr.19: 51009156-51009156 on Build GRCh38
rs911233815 Chr.19: 51009432-51009432 on Build GRCh38
rs775351827 Chr.19: 51009268-51009268 on Build GRCh38
rs771450313 Chr.19: 51009273-51009274 on Build GRCh38
rs914956107 Chr.19: 51009135-51009135 on Build GRCh38
rs771821787 Chr.19: 51009259-51009259 on Build GRCh38
rs923667666 Chr.19: 51009549-51009549 on Build GRCh38
rs768168789 Chr.19: 51009402-51009402 on Build GRCh38
rs567713358 Chr.19: 51009450-51009450 on Build GRCh38
rs978033148 Chr.19: 51009488-51009488 on Build GRCh38
rs766343476 Chr.19: 51009585-51009585 on Build GRCh38
rs762650669 Chr.19: 51009305-51009305 on Build GRCh38
rs759271424 Chr.19: 51009178-51009178 on Build GRCh38
rs766068135 Chr.19: 51009371-51009371 on Build GRCh38
rs903685505 Chr.19: 51009509-51009509 on Build GRCh38
rs375285822 Chr.19: 51009583-51009583 on Build GRCh38
rs772345573 Chr.19: 51009330-51009330 on Build GRCh38
rs746080624 Chr.19: 51009252-51009252 on Build GRCh38
rs751123063 Chr.19: 51009377-51009377 on Build GRCh38
rs114957460 Chr.19: 51009319-51009319 on Build GRCh38
rs922183313 Chr.19: 51009223-51009223 on Build GRCh38
rs770948361 Chr.19: 51009249-51009249 on Build GRCh38
rs758363909 Chr.19: 51009485-51009485 on Build GRCh38
rs370785528 Chr.19: 51009563-51009563 on Build GRCh38
rs764804237 Chr.19: 51009295-51009295 on Build GRCh38
rs761626230 Chr.19: 51009510-51009518 on Build GRCh38
rs762613848 Chr.19: 51009381-51009381 on Build GRCh38
rs543176018 Chr.19: 51009311-51009311 on Build GRCh38
rs118001778 Chr.19: 51009190-51009190 on Build GRCh38
rs759175914 Chr.19: 51009561-51009561 on Build GRCh38
rs762109744 Chr.19: 51009574-51009574 on Build GRCh38
rs895103261 Chr.19: 51009168-51009168 on Build GRCh38
rs748654152 Chr.19: 51009150-51009150 on Build GRCh38
rs535116738 Chr.19: 51009515-51009515 on Build GRCh38
rs142241854 Chr.19: 51009280-51009280 on Build GRCh38
rs367832421 Chr.19: 51009209-51009209 on Build GRCh38
rs1056092378 Chr.19: 51009569-51009569 on Build GRCh38
rs746847427 Chr.19: 51009338-51009338 on Build GRCh38
rs748657584 Chr.19: 51009224-51009224 on Build GRCh38
rs368772123 Chr.19: 51009558-51009558 on Build GRCh38
rs748850775 Chr.19: 51009547-51009547 on Build GRCh38
rs769483742 Chr.19: 51009356-51009356 on Build GRCh38
rs765008197 Chr.19: 51009208-51009208 on Build GRCh38
rs920077852 Chr.19: 51009470-51009470 on Build GRCh38
rs886159548 Chr.19: 51009499-51009499 on Build GRCh38
rs778274403 Chr.19: 51009310-51009310 on Build GRCh38
rs750531806 Chr.19: 51009482-51009482 on Build GRCh38
rs368005269 Chr.19: 51009586-51009586 on Build GRCh38
rs756411328 Chr.19: 51009307-51009307 on Build GRCh38
rs775339995 Chr.19: 51009182-51009182 on Build GRCh38
rs747396579 Chr.19: 51009136-51009136 on Build GRCh38
rs761058524 Chr.19: 51009388-51009388 on Build GRCh38
rs754402433 Chr.19: 51009474-51009474 on Build GRCh38
rs1015301367 Chr.19: 51009210-51009210 on Build GRCh38
rs781595948 Chr.19: 51009217-51009217 on Build GRCh38
rs753107467 Chr.19: 51009306-51009306 on Build GRCh38
rs769811274 Chr.19: 51009151-51009151 on Build GRCh38
rs765799306 Chr.19: 51009575-51009575 on Build GRCh38
rs557702718 Chr.19: 51009250-51009250 on Build GRCh38
rs756639038 Chr.19: 51009230-51009230 on Build GRCh38
rs942944016 Chr.19: 51009189-51009189 on Build GRCh38
rs778184487 Chr.19: 51009231-51009231 on Build GRCh38
rs997451038 Chr.19: 51009355-51009355 on Build GRCh38
rs767313381 Chr.19: 51009180-51009180 on Build GRCh38
rs760649897 Chr.19: 51009187-51009187 on Build GRCh38
rs1042416991 Chr.19: 51009146-51009146 on Build GRCh38
rs371619053 Chr.19: 51009389-51009389 on Build GRCh38
rs760322248 Chr.19: 51009269-51009269 on Build GRCh38
rs202182260 Chr.19: 51009321-51009321 on Build GRCh38
rs747206849 Chr.19: 51009487-51009487 on Build GRCh38
rs757757976 Chr.19: 51009476-51009476 on Build GRCh38
rs748161036 Chr.19: 51009504-51009504 on Build GRCh38
rs570185388 Chr.19: 51009417-51009417 on Build GRCh38
rs552038744 Chr.19: 51009411-51009411 on Build GRCh38
rs781117231 Chr.19: 51009494-51009494 on Build GRCh38
rs770306784 Chr.19: 51009550-51009550 on Build GRCh38
rs375009841 Chr.19: 51009347-51009347 on Build GRCh38
rs772962792 Chr.19: 51009516-51009516 on Build GRCh38
rs754864302 Chr.19: 51009489-51009489 on Build GRCh38
rs58876874 Chr.19: 51009366-51009366 on Build GRCh38
rs766765626 Chr.19: 51009235-51009235 on Build GRCh38
rs780357953 Chr.19: 51009486-51009486 on Build GRCh38
rs768731474 Chr.19: 51009279-51009279 on Build GRCh38
rs149100992 Chr.19: 51009216-51009216 on Build GRCh38
rs771144624 Chr.19: 51009159-51009159 on Build GRCh38
rs779223736 Chr.19: 51009481-51009481 on Build GRCh38
rs749383199 Chr.19: 51009233-51009233 on Build GRCh38
rs373246049 Chr.19: 51009440-51009440 on Build GRCh38
rs776986326 Chr.19: 51009567-51009567 on Build GRCh38

View Full Product Details