Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52015790-52016293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF614
Entrez Gene ID
80110
Gene Name
zinc finger protein 614
Gene Aliases
-
UniGene
Hs.292336
Gene Chromosome Location
Chr.19: 52013324-52028427 on Build GRCh38
Transcripts
NM_025040.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1000316 c.1445C>T p.P482L Chr.19: 52016153-52016153 on Build GRCh38
7166278 c.1398G>C p.Q466H Chr.19: 52016200-52016200 on Build GRCh38
6085534 c.1312A>G p.T438A Chr.19: 52016286-52016286 on Build GRCh38
1000315 c.1690G>T p.E564* Chr.19: 52015908-52015908 on Build GRCh38
7168656 c.1747C>T p.P583S Chr.19: 52015851-52015851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772956705 Chr.19: 52016238-52016238 on Build GRCh38
rs749804253 Chr.19: 52015969-52015969 on Build GRCh38
rs927021321 Chr.19: 52016117-52016117 on Build GRCh38
rs542117629 Chr.19: 52015836-52015836 on Build GRCh38
rs778684503 Chr.19: 52015839-52015839 on Build GRCh38
rs1050506097 Chr.19: 52016194-52016194 on Build GRCh38
rs757184052 Chr.19: 52015901-52015901 on Build GRCh38
rs768611015 Chr.19: 52016012-52016013 on Build GRCh38
rs762724681 Chr.19: 52016146-52016146 on Build GRCh38
rs752186759 Chr.19: 52016179-52016179 on Build GRCh38
rs748616123 Chr.19: 52015858-52015858 on Build GRCh38
rs199963700 Chr.19: 52015834-52015834 on Build GRCh38
rs759212450 Chr.19: 52016167-52016167 on Build GRCh38
rs148053965 Chr.19: 52016208-52016208 on Build GRCh38
rs762182085 Chr.19: 52016142-52016145 on Build GRCh38
rs778323349 Chr.19: 52015957-52015957 on Build GRCh38
rs868492152 Chr.19: 52015851-52015851 on Build GRCh38
rs373361966 Chr.19: 52016203-52016203 on Build GRCh38
rs779585934 Chr.19: 52015938-52015938 on Build GRCh38
rs774282844 Chr.19: 52016089-52016090 on Build GRCh38
rs200147759 Chr.19: 52015846-52015846 on Build GRCh38
rs994457280 Chr.19: 52016007-52016007 on Build GRCh38
rs773413455 Chr.19: 52016148-52016149 on Build GRCh38
rs564581984 Chr.19: 52016002-52016002 on Build GRCh38
rs767643393 Chr.19: 52016145-52016146 on Build GRCh38
rs755280234 Chr.19: 52016225-52016225 on Build GRCh38
rs750791115 Chr.19: 52016189-52016189 on Build GRCh38
rs746571951 Chr.19: 52015944-52015944 on Build GRCh38
rs542386134 Chr.19: 52016065-52016065 on Build GRCh38
rs769282366 Chr.19: 52015860-52015860 on Build GRCh38
rs757672975 Chr.19: 52015946-52015946 on Build GRCh38
rs368979195 Chr.19: 52015877-52015877 on Build GRCh38
rs779342561 Chr.19: 52016185-52016185 on Build GRCh38
rs201885469 Chr.19: 52016119-52016119 on Build GRCh38
rs182429124 Chr.19: 52016098-52016098 on Build GRCh38
rs758020339 Chr.19: 52015929-52015929 on Build GRCh38
rs780489737 Chr.19: 52016124-52016124 on Build GRCh38
rs749711962 Chr.19: 52015868-52015868 on Build GRCh38
rs889145328 Chr.19: 52015926-52015927 on Build GRCh38
rs200985913 Chr.19: 52016140-52016140 on Build GRCh38
rs770668249 Chr.19: 52016265-52016265 on Build GRCh38
rs867901644 Chr.19: 52016069-52016069 on Build GRCh38
rs771059044 Chr.19: 52015875-52015875 on Build GRCh38
rs766428658 Chr.19: 52016267-52016269 on Build GRCh38
rs746059952 Chr.19: 52015997-52015997 on Build GRCh38
rs368277614 Chr.19: 52015960-52015960 on Build GRCh38
rs376539129 Chr.19: 52016234-52016234 on Build GRCh38
rs758623266 Chr.19: 52016193-52016193 on Build GRCh38
rs766169981 Chr.19: 52016151-52016151 on Build GRCh38
rs375760839 Chr.19: 52016120-52016120 on Build GRCh38
rs746264178 Chr.19: 52016122-52016122 on Build GRCh38
rs749214686 Chr.19: 52015998-52015999 on Build GRCh38
rs775321396 Chr.19: 52015911-52015912 on Build GRCh38
rs747239289 Chr.19: 52016125-52016125 on Build GRCh38
rs376960134 Chr.19: 52015861-52015861 on Build GRCh38
rs761668334 Chr.19: 52016061-52016061 on Build GRCh38
rs767502096 Chr.19: 52015814-52015814 on Build GRCh38
rs751064925 Chr.19: 52016088-52016088 on Build GRCh38
rs146501709 Chr.19: 52016084-52016084 on Build GRCh38
rs373635517 Chr.19: 52015831-52015831 on Build GRCh38
rs775740971 Chr.19: 52015893-52015893 on Build GRCh38
rs768709180 Chr.19: 52016038-52016038 on Build GRCh38
rs1016847149 Chr.19: 52015952-52015952 on Build GRCh38
rs750032179 Chr.19: 52015928-52015928 on Build GRCh38
rs764943767 Chr.19: 52015920-52015920 on Build GRCh38
rs772321022 Chr.19: 52016123-52016123 on Build GRCh38
rs369319461 Chr.19: 52016049-52016049 on Build GRCh38
rs771272193 Chr.19: 52015984-52015984 on Build GRCh38
rs767587556 Chr.19: 52015890-52015890 on Build GRCh38
rs760696985 Chr.19: 52016182-52016196 on Build GRCh38
rs143573470 Chr.19: 52016182-52016182 on Build GRCh38
rs145338687 Chr.19: 52015843-52015843 on Build GRCh38
rs377286967 Chr.19: 52015904-52015904 on Build GRCh38
rs750226700 Chr.19: 52016081-52016081 on Build GRCh38
rs766977365 Chr.19: 52016170-52016170 on Build GRCh38
rs762646626 Chr.19: 52016082-52016082 on Build GRCh38
rs766575145 Chr.19: 52016270-52016270 on Build GRCh38
rs752624933 Chr.19: 52015818-52015818 on Build GRCh38
rs563826556 Chr.19: 52016017-52016017 on Build GRCh38
rs773459814 Chr.19: 52016134-52016134 on Build GRCh38
rs774570178 Chr.19: 52015876-52015876 on Build GRCh38
rs764172255 Chr.19: 52015898-52015898 on Build GRCh38
rs749265951 Chr.19: 52016259-52016259 on Build GRCh38
rs780697736 Chr.19: 52016104-52016104 on Build GRCh38
rs369045441 Chr.19: 52016139-52016139 on Build GRCh38
rs375579380 Chr.19: 52016018-52016018 on Build GRCh38
rs768796873 Chr.19: 52016133-52016133 on Build GRCh38
rs756140114 Chr.19: 52015823-52015823 on Build GRCh38
rs879049514 Chr.19: 52016172-52016172 on Build GRCh38
rs73934727 Chr.19: 52016221-52016221 on Build GRCh38
rs115605807 Chr.19: 52016164-52016164 on Build GRCh38
rs367753832 Chr.19: 52016023-52016023 on Build GRCh38
rs773544718 Chr.19: 52015864-52015864 on Build GRCh38
rs769766064 Chr.19: 52016236-52016236 on Build GRCh38
rs549653554 Chr.19: 52016235-52016235 on Build GRCh38
rs756601400 Chr.19: 52015948-52015948 on Build GRCh38
rs142351310 Chr.19: 52015896-52015896 on Build GRCh38
rs765567946 Chr.19: 52015985-52015985 on Build GRCh38
rs538034597 Chr.19: 52016000-52016000 on Build GRCh38
rs753672766 Chr.19: 52015899-52015899 on Build GRCh38
rs757738536 Chr.19: 52016113-52016113 on Build GRCh38
rs372930850 Chr.19: 52016110-52016110 on Build GRCh38
rs946330118 Chr.19: 52016144-52016144 on Build GRCh38
rs201085926 Chr.19: 52016239-52016239 on Build GRCh38
rs147676609 Chr.19: 52015850-52015850 on Build GRCh38

View Full Product Details