Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52385394-52385892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF880
Entrez Gene ID
400713
Gene Name
zinc finger protein 880
Gene Aliases
-
UniGene
Hs.502314
Gene Chromosome Location
Chr.19: 52369900-52393740 on Build GRCh38
Transcripts
NM_001145434.1 XM_017026812.1

SNP Detail

SNP ID Chromosome Location
rs115603968 Chr.19: 52385620-52385620 on Build GRCh38
rs192886392 Chr.19: 52385844-52385844 on Build GRCh38
rs983059869 Chr.19: 52385604-52385604 on Build GRCh38
rs572779806 Chr.19: 52385692-52385692 on Build GRCh38
rs770830624 Chr.19: 52385868-52385868 on Build GRCh38
rs934579036 Chr.19: 52385718-52385718 on Build GRCh38
rs141602552 Chr.19: 52385725-52385725 on Build GRCh38
rs369722694 Chr.19: 52385760-52385760 on Build GRCh38
rs761903256 Chr.19: 52385612-52385612 on Build GRCh38
rs764407636 Chr.19: 52385585-52385585 on Build GRCh38
rs542400885 Chr.19: 52385636-52385636 on Build GRCh38
rs371978669 Chr.19: 52385563-52385564 on Build GRCh38
rs770844403 Chr.19: 52385669-52385669 on Build GRCh38
rs751445590 Chr.19: 52385690-52385690 on Build GRCh38
rs1006422533 Chr.19: 52385608-52385608 on Build GRCh38
rs947126621 Chr.19: 52385708-52385708 on Build GRCh38
rs936018206 Chr.19: 52385706-52385706 on Build GRCh38
rs766257620 Chr.19: 52385652-52385652 on Build GRCh38
rs996839228 Chr.19: 52385584-52385584 on Build GRCh38
rs996334718 Chr.19: 52385845-52385845 on Build GRCh38
rs978582170 Chr.19: 52385714-52385714 on Build GRCh38
rs555939742 Chr.19: 52385436-52385436 on Build GRCh38
rs578203972 Chr.19: 52385530-52385530 on Build GRCh38
rs775869997 Chr.19: 52385458-52385458 on Build GRCh38
rs924406412 Chr.19: 52385717-52385717 on Build GRCh38
rs900627566 Chr.19: 52385583-52385583 on Build GRCh38
rs925208204 Chr.19: 52385702-52385702 on Build GRCh38
rs1052866160 Chr.19: 52385518-52385518 on Build GRCh38
rs991885142 Chr.19: 52385660-52385660 on Build GRCh38
rs753913990 Chr.19: 52385602-52385602 on Build GRCh38
rs951732753 Chr.19: 52385595-52385595 on Build GRCh38
rs1019527298 Chr.19: 52385563-52385563 on Build GRCh38
rs532295660 Chr.19: 52385828-52385828 on Build GRCh38
rs895719181 Chr.19: 52385843-52385843 on Build GRCh38
rs891871123 Chr.19: 52385534-52385534 on Build GRCh38
rs56955135 Chr.19: 52385588-52385588 on Build GRCh38
rs1009102258 Chr.19: 52385561-52385561 on Build GRCh38
rs550313139 Chr.19: 52385838-52385838 on Build GRCh38
rs1057080553 Chr.19: 52385735-52385735 on Build GRCh38
rs904537401 Chr.19: 52385423-52385423 on Build GRCh38
rs754779033 Chr.19: 52385691-52385691 on Build GRCh38
rs1053099248 Chr.19: 52385710-52385710 on Build GRCh38
rs761035660 Chr.19: 52385468-52385468 on Build GRCh38
rs979778635 Chr.19: 52385693-52385693 on Build GRCh38
rs916408096 Chr.19: 52385661-52385661 on Build GRCh38
rs954438714 Chr.19: 52385635-52385635 on Build GRCh38
rs746746429 Chr.19: 52385781-52385781 on Build GRCh38
rs557593685 Chr.19: 52385616-52385616 on Build GRCh38
rs560937002 Chr.19: 52385663-52385663 on Build GRCh38
rs962307936 Chr.19: 52385615-52385615 on Build GRCh38

View Full Product Details