Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53351038-53351550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF845
Entrez Gene ID
91664
Gene Name
zinc finger protein 845
Gene Aliases
-
UniGene
Hs.720672
Gene Chromosome Location
Chr.19: 53333719-53356119 on Build GRCh38
Transcripts
NM_001321522.1 NM_001321523.1 NM_001321524.1 NM_138374.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4498947 c.530C>T p.S177F Chr.19: 53351205-53351205 on Build GRCh38
7112950 c.868C>G p.H290D Chr.19: 53351543-53351543 on Build GRCh38
5646755 c.493G>A p.E165K Chr.19: 53351168-53351168 on Build GRCh38
1000787 c.438G>A p.S146S Chr.19: 53351113-53351113 on Build GRCh38
6264679 c.825T>G p.D275E Chr.19: 53351500-53351500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780670436 Chr.19: 53351428-53351428 on Build GRCh38
rs772421071 Chr.19: 53351201-53351201 on Build GRCh38
rs747348279 Chr.19: 53351310-53351310 on Build GRCh38
rs764664402 Chr.19: 53351128-53351128 on Build GRCh38
rs750958161 Chr.19: 53351274-53351274 on Build GRCh38
rs771328693 Chr.19: 53351324-53351324 on Build GRCh38
rs772949975 Chr.19: 53351442-53351442 on Build GRCh38
rs1000140526 Chr.19: 53351132-53351132 on Build GRCh38
rs61752582 Chr.19: 53351304-53351304 on Build GRCh38
rs780394233 Chr.19: 53351501-53351501 on Build GRCh38
rs779155365 Chr.19: 53351190-53351190 on Build GRCh38
rs765060460 Chr.19: 53351456-53351456 on Build GRCh38
rs768426345 Chr.19: 53351346-53351349 on Build GRCh38
rs781150772 Chr.19: 53351087-53351087 on Build GRCh38
rs187114554 Chr.19: 53351500-53351500 on Build GRCh38
rs757875961 Chr.19: 53351353-53351353 on Build GRCh38
rs61743334 Chr.19: 53351098-53351098 on Build GRCh38
rs61743337 Chr.19: 53351112-53351112 on Build GRCh38
rs762512207 Chr.19: 53351137-53351137 on Build GRCh38
rs751207012 Chr.19: 53351143-53351143 on Build GRCh38
rs752894826 Chr.19: 53351303-53351303 on Build GRCh38
rs556181514 Chr.19: 53351434-53351434 on Build GRCh38
rs190734350 Chr.19: 53351393-53351393 on Build GRCh38
rs767355902 Chr.19: 53351253-53351253 on Build GRCh38
rs759394097 Chr.19: 53351386-53351386 on Build GRCh38
rs73057056 Chr.19: 53351248-53351248 on Build GRCh38
rs746386737 Chr.19: 53351330-53351330 on Build GRCh38
rs947088854 Chr.19: 53351375-53351375 on Build GRCh38
rs545998523 Chr.19: 53351328-53351328 on Build GRCh38
rs764184372 Chr.19: 53351472-53351472 on Build GRCh38
rs10415799 Chr.19: 53351144-53351144 on Build GRCh38
rs182006841 Chr.19: 53351460-53351460 on Build GRCh38
rs780207028 Chr.19: 53351172-53351172 on Build GRCh38
rs767849535 Chr.19: 53351482-53351482 on Build GRCh38
rs370007608 Chr.19: 53351101-53351101 on Build GRCh38
rs747588639 Chr.19: 53351436-53351436 on Build GRCh38
rs138639565 Chr.19: 53351241-53351241 on Build GRCh38
rs757571917 Chr.19: 53351403-53351403 on Build GRCh38
rs749454079 Chr.19: 53351104-53351104 on Build GRCh38
rs145709331 Chr.19: 53351237-53351237 on Build GRCh38
rs758054251 Chr.19: 53351063-53351063 on Build GRCh38
rs759430402 Chr.19: 53351454-53351454 on Build GRCh38
rs770198839 Chr.19: 53351425-53351425 on Build GRCh38
rs751815256 Chr.19: 53351395-53351395 on Build GRCh38
rs879243538 Chr.19: 53351092-53351092 on Build GRCh38
rs898332524 Chr.19: 53351488-53351488 on Build GRCh38
rs576404205 Chr.19: 53351307-53351307 on Build GRCh38
rs778869993 Chr.19: 53351299-53351299 on Build GRCh38
rs756211629 Chr.19: 53351100-53351100 on Build GRCh38
rs777359043 Chr.19: 53351066-53351066 on Build GRCh38
rs752716776 Chr.19: 53351459-53351459 on Build GRCh38
rs755035226 Chr.19: 53351297-53351297 on Build GRCh38
rs371446024 Chr.19: 53351452-53351452 on Build GRCh38
rs768918565 Chr.19: 53351110-53351110 on Build GRCh38
rs1051007931 Chr.19: 53351077-53351077 on Build GRCh38
rs1025306060 Chr.19: 53351163-53351163 on Build GRCh38
rs753857286 Chr.19: 53351279-53351279 on Build GRCh38
rs568966992 Chr.19: 53351182-53351182 on Build GRCh38
rs762283354 Chr.19: 53351234-53351234 on Build GRCh38
rs771838725 Chr.19: 53351352-53351352 on Build GRCh38
rs543460835 Chr.19: 53351113-53351113 on Build GRCh38
rs750286058 Chr.19: 53351455-53351455 on Build GRCh38
rs368740048 Chr.19: 53351443-53351443 on Build GRCh38
rs774797916 Chr.19: 53351132-53351133 on Build GRCh38
rs747437987 Chr.19: 53351218-53351218 on Build GRCh38
rs768158079 Chr.19: 53351335-53351335 on Build GRCh38
rs781553410 Chr.19: 53351406-53351406 on Build GRCh38
rs774009128 Chr.19: 53351247-53351247 on Build GRCh38
rs763776334 Chr.19: 53351140-53351140 on Build GRCh38
rs773740031 Chr.19: 53351339-53351339 on Build GRCh38
rs749356222 Chr.19: 53351180-53351180 on Build GRCh38
rs751725606 Chr.19: 53351476-53351476 on Build GRCh38
rs889712165 Chr.19: 53351429-53351429 on Build GRCh38
rs760505583 Chr.19: 53351361-53351361 on Build GRCh38
rs753749173 Chr.19: 53351371-53351371 on Build GRCh38
rs541535088 Chr.19: 53351343-53351343 on Build GRCh38
rs549418994 Chr.19: 53351083-53351083 on Build GRCh38
rs752292123 Chr.19: 53351062-53351062 on Build GRCh38
rs1053047557 Chr.19: 53351424-53351424 on Build GRCh38
rs756123874 Chr.19: 53351147-53351147 on Build GRCh38
rs370565475 Chr.19: 53351191-53351191 on Build GRCh38
rs773567389 Chr.19: 53351211-53351211 on Build GRCh38
rs923099116 Chr.19: 53351337-53351337 on Build GRCh38
rs756030870 Chr.19: 53351271-53351271 on Build GRCh38
rs758518640 Chr.19: 53351394-53351394 on Build GRCh38
rs571802419 Chr.19: 53351115-53351115 on Build GRCh38
rs750240008 Chr.19: 53351257-53351257 on Build GRCh38
rs751351529 Chr.19: 53351068-53351068 on Build GRCh38
rs766297418 Chr.19: 53351366-53351366 on Build GRCh38
rs867562170 Chr.19: 53351090-53351090 on Build GRCh38
rs746322112 Chr.19: 53351409-53351409 on Build GRCh38
rs374984657 Chr.19: 53351493-53351493 on Build GRCh38
rs367669285 Chr.19: 53351513-53351513 on Build GRCh38
rs771738910 Chr.19: 53351441-53351441 on Build GRCh38
rs757922002 Chr.19: 53351520-53351520 on Build GRCh38
rs539111747 Chr.19: 53351133-53351133 on Build GRCh38
rs910456413 Chr.19: 53351356-53351356 on Build GRCh38
rs757477372 Chr.19: 53351477-53351477 on Build GRCh38
rs1006266902 Chr.19: 53351523-53351523 on Build GRCh38
rs762958046 Chr.19: 53351159-53351159 on Build GRCh38
rs373977075 Chr.19: 53351181-53351181 on Build GRCh38
rs771410312 Chr.19: 53351224-53351224 on Build GRCh38

View Full Product Details