Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53242769-53243288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100507564
Entrez Gene ID
100507564
Gene Name
uncharacterized LOC100507564
Gene Aliases
-
UniGene
Hs.592502
Gene Chromosome Location
Chr.1: 53238610-53242783 on Build GRCh38
Transcripts
NR_038953.1 NM_001018054.2 NM_004631.4 NM_017522.4 NM_033300.3

Gene Symbol
LRP8
Entrez Gene ID
7804
Gene Name
LDL receptor related protein 8
Gene Aliases
APOER2, HSZ75190, LRP-8, MCI1
UniGene
Hs.280387
Gene Chromosome Location
Chr.1: 53242364-53328149 on Build GRCh38
Transcripts
NR_038953.1 NM_001018054.2 NM_004631.4 NM_017522.4 NM_033300.3

SNP Detail

SNP ID Chromosome Location
rs796527635 Chr.1: 53242858-53242859 on Build GRCh38
rs898673084 Chr.1: 53243012-53243012 on Build GRCh38
rs574103941 Chr.1: 53243126-53243126 on Build GRCh38
rs760399892 Chr.1: 53242837-53242840 on Build GRCh38
rs931880192 Chr.1: 53243156-53243156 on Build GRCh38
rs995760302 Chr.1: 53243027-53243027 on Build GRCh38
rs61660881 Chr.1: 53242862-53242862 on Build GRCh38
rs1018686926 Chr.1: 53242819-53242819 on Build GRCh38
rs979355235 Chr.1: 53243235-53243235 on Build GRCh38
rs57520830 Chr.1: 53242860-53242860 on Build GRCh38
rs956352346 Chr.1: 53242818-53242818 on Build GRCh38
rs563640118 Chr.1: 53243001-53243001 on Build GRCh38
rs969108053 Chr.1: 53243214-53243214 on Build GRCh38
rs1008765392 Chr.1: 53243168-53243168 on Build GRCh38
rs56722919 Chr.1: 53242858-53242859 on Build GRCh38
rs938903746 Chr.1: 53242987-53242987 on Build GRCh38
rs940657204 Chr.1: 53243052-53243052 on Build GRCh38
rs907975865 Chr.1: 53243022-53243022 on Build GRCh38
rs1056560960 Chr.1: 53242871-53242871 on Build GRCh38
rs746220586 Chr.1: 53242839-53242840 on Build GRCh38
rs1018570545 Chr.1: 53243172-53243172 on Build GRCh38
rs550886281 Chr.1: 53243217-53243217 on Build GRCh38
rs565138237 Chr.1: 53243177-53243177 on Build GRCh38
rs3737984 Chr.1: 53243178-53243178 on Build GRCh38
rs987960006 Chr.1: 53242855-53242855 on Build GRCh38
rs896607653 Chr.1: 53242872-53242872 on Build GRCh38
rs1034643345 Chr.1: 53243237-53243237 on Build GRCh38
rs771071197 Chr.1: 53243152-53243152 on Build GRCh38
rs927437375 Chr.1: 53242918-53242918 on Build GRCh38
rs938017788 Chr.1: 53242864-53242864 on Build GRCh38
rs569291231 Chr.1: 53243255-53243255 on Build GRCh38
rs914785148 Chr.1: 53242857-53242860 on Build GRCh38
rs557051853 Chr.1: 53243241-53243241 on Build GRCh38
rs144308373 Chr.1: 53242837-53242838 on Build GRCh38
rs202125021 Chr.1: 53242858-53242858 on Build GRCh38
rs1037177095 Chr.1: 53242898-53242898 on Build GRCh38
rs56876335 Chr.1: 53242859-53242860 on Build GRCh38
rs543749145 Chr.1: 53242897-53242897 on Build GRCh38
rs778529390 Chr.1: 53243259-53243259 on Build GRCh38
rs542732682 Chr.1: 53243127-53243127 on Build GRCh38
rs1050452741 Chr.1: 53243153-53243153 on Build GRCh38
rs867220780 Chr.1: 53242991-53242991 on Build GRCh38
rs559291822 Chr.1: 53242872-53242873 on Build GRCh38
rs980264178 Chr.1: 53242873-53242873 on Build GRCh38
rs528667571 Chr.1: 53243162-53243162 on Build GRCh38
rs373357546 Chr.1: 53243050-53243050 on Build GRCh38
rs201213593 Chr.1: 53242856-53242856 on Build GRCh38
rs960452763 Chr.1: 53243250-53243250 on Build GRCh38
rs965841288 Chr.1: 53242842-53242842 on Build GRCh38
rs11206126 Chr.1: 53243174-53243174 on Build GRCh38
rs1009751145 Chr.1: 53243175-53243175 on Build GRCh38
rs754561374 Chr.1: 53243181-53243181 on Build GRCh38
rs559831905 Chr.1: 53243160-53243160 on Build GRCh38
rs1037684347 Chr.1: 53243142-53243142 on Build GRCh38

View Full Product Details