Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53767684-53768186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDC1
Entrez Gene ID
55706
Gene Name
NDC1 transmembrane nucleoporin
Gene Aliases
NET3, TMEM48
UniGene
Hs.476525
Gene Chromosome Location
Chr.1: 53765460-53838552 on Build GRCh38
Transcripts
NM_001168551.1 NM_018087.4 NR_033142.1 XM_011541766.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
303413 c.1984C>A p.H662N Chr.1: 53768011-53768011 on Build GRCh38
3805569 c.2019A>C p.K673N Chr.1: 53767976-53767976 on Build GRCh38
1320988 c.1978G>A p.A660T Chr.1: 53768017-53768017 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1003999734 Chr.1: 53767882-53767882 on Build GRCh38
rs374774558 Chr.1: 53768005-53768005 on Build GRCh38
rs1025653612 Chr.1: 53767945-53767945 on Build GRCh38
rs368324431 Chr.1: 53767924-53767924 on Build GRCh38
rs1030316263 Chr.1: 53767805-53767805 on Build GRCh38
rs761340819 Chr.1: 53768046-53768046 on Build GRCh38
rs535338305 Chr.1: 53767851-53767851 on Build GRCh38
rs781682508 Chr.1: 53767902-53767902 on Build GRCh38
rs750011192 Chr.1: 53768045-53768046 on Build GRCh38
rs766919396 Chr.1: 53768056-53768056 on Build GRCh38
rs534511439 Chr.1: 53767992-53767992 on Build GRCh38
rs936534217 Chr.1: 53768096-53768096 on Build GRCh38
rs372305558 Chr.1: 53767950-53767950 on Build GRCh38
rs1053602745 Chr.1: 53768115-53768115 on Build GRCh38
rs975617400 Chr.1: 53767994-53767994 on Build GRCh38
rs749993294 Chr.1: 53768065-53768065 on Build GRCh38
rs750355287 Chr.1: 53767933-53767933 on Build GRCh38
rs776971958 Chr.1: 53768024-53768024 on Build GRCh38
rs1009597643 Chr.1: 53767770-53767770 on Build GRCh38
rs923568760 Chr.1: 53767714-53767714 on Build GRCh38
rs759948741 Chr.1: 53768026-53768026 on Build GRCh38
rs771465194 Chr.1: 53768016-53768016 on Build GRCh38
rs751982339 Chr.1: 53767856-53767856 on Build GRCh38
rs763801270 Chr.1: 53767922-53767922 on Build GRCh38
rs371861925 Chr.1: 53768001-53768001 on Build GRCh38
rs1029795542 Chr.1: 53768011-53768011 on Build GRCh38
rs71637818 Chr.1: 53767838-53767838 on Build GRCh38
rs759870110 Chr.1: 53767925-53767925 on Build GRCh38
rs772431063 Chr.1: 53767926-53767928 on Build GRCh38
rs557981379 Chr.1: 53768111-53768111 on Build GRCh38
rs892408230 Chr.1: 53768145-53768145 on Build GRCh38
rs368528310 Chr.1: 53767963-53767963 on Build GRCh38
rs954832872 Chr.1: 53767821-53767821 on Build GRCh38
rs753350561 Chr.1: 53767767-53767767 on Build GRCh38
rs987597225 Chr.1: 53768037-53768037 on Build GRCh38
rs761063573 Chr.1: 53768015-53768015 on Build GRCh38
rs763676532 Chr.1: 53768035-53768035 on Build GRCh38
rs756728968 Chr.1: 53767927-53767927 on Build GRCh38
rs781146916 Chr.1: 53767930-53767930 on Build GRCh38
rs1053740560 Chr.1: 53767751-53767751 on Build GRCh38
rs764849013 Chr.1: 53767967-53767970 on Build GRCh38
rs754470935 Chr.1: 53767835-53767835 on Build GRCh38
rs866565956 Chr.1: 53767864-53767864 on Build GRCh38
rs776169981 Chr.1: 53767950-53767951 on Build GRCh38
rs935275251 Chr.1: 53767755-53767755 on Build GRCh38
rs1000791382 Chr.1: 53767739-53767739 on Build GRCh38
rs41311158 Chr.1: 53767869-53767869 on Build GRCh38
rs201665807 Chr.1: 53768036-53768036 on Build GRCh38
rs756360674 Chr.1: 53767979-53767979 on Build GRCh38
rs923649231 Chr.1: 53768133-53768133 on Build GRCh38
rs150627210 Chr.1: 53767961-53767961 on Build GRCh38
rs868742838 Chr.1: 53768009-53768009 on Build GRCh38
rs756130200 Chr.1: 53767948-53767948 on Build GRCh38
rs761194658 Chr.1: 53767962-53767965 on Build GRCh38
rs780888336 Chr.1: 53767814-53767814 on Build GRCh38

View Full Product Details