Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46580955-46581455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGF
Entrez Gene ID
5281
Gene Name
phosphatidylinositol glycan anchor biosynthesis class F
Gene Aliases
-
UniGene
Hs.468415
Gene Chromosome Location
Chr.2: 46581274-46617112 on Build GRCh38
Transcripts
NM_002643.3 NM_173074.2 XM_011532908.2 NM_012249.3 XM_005264229.2 XM_011532726.2 XM_011532728.2 XM_017003716.1

Gene Symbol
RHOQ
Entrez Gene ID
23433
Gene Name
ras homolog family member Q
Gene Aliases
ARHQ, HEL-S-42, RASL7A, TC10, TC10A
UniGene
Hs.709193
Gene Chromosome Location
Chr.2: 46542409-46584688 on Build GRCh38
Transcripts
NM_002643.3 NM_173074.2 XM_011532908.2 NM_012249.3 XM_005264229.2 XM_011532726.2 XM_011532728.2 XM_017003716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
21941 c.500C>T p.T167I Chr.2: 46580965-46580965 on Build GRCh38
5662235 c.570A>T p.V190V Chr.2: 46581035-46581035 on Build GRCh38
6158543 c.529G>T p.E177* Chr.2: 46580994-46580994 on Build GRCh38
1408217 c.499A>G p.T167A Chr.2: 46580964-46580964 on Build GRCh38
6766270 c.550A>G p.T184A Chr.2: 46581015-46581015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936670450 Chr.2: 46581376-46581376 on Build GRCh38
rs1026360459 Chr.2: 46581222-46581222 on Build GRCh38
rs1028423552 Chr.2: 46581046-46581046 on Build GRCh38
rs531888006 Chr.2: 46581230-46581230 on Build GRCh38
rs770383518 Chr.2: 46581377-46581378 on Build GRCh38
rs926374965 Chr.2: 46581235-46581235 on Build GRCh38
rs202122445 Chr.2: 46581364-46581366 on Build GRCh38
rs141544914 Chr.2: 46581079-46581079 on Build GRCh38
rs765355378 Chr.2: 46581123-46581123 on Build GRCh38
rs778047457 Chr.2: 46581023-46581023 on Build GRCh38
rs17038378 Chr.2: 46581105-46581105 on Build GRCh38
rs889864427 Chr.2: 46581379-46581379 on Build GRCh38
rs750352612 Chr.2: 46581055-46581055 on Build GRCh38
rs763105097 Chr.2: 46581134-46581134 on Build GRCh38
rs554944769 Chr.2: 46581428-46581428 on Build GRCh38
rs149388763 Chr.2: 46581105-46581106 on Build GRCh38
rs972966095 Chr.2: 46581216-46581216 on Build GRCh38
rs945238006 Chr.2: 46581355-46581355 on Build GRCh38
rs1001335304 Chr.2: 46581034-46581034 on Build GRCh38
rs748792572 Chr.2: 46581042-46581042 on Build GRCh38
rs3088236 Chr.2: 46581421-46581421 on Build GRCh38
rs55654131 Chr.2: 46581029-46581029 on Build GRCh38
rs374937930 Chr.2: 46581159-46581159 on Build GRCh38
rs758830720 Chr.2: 46581014-46581014 on Build GRCh38
rs752951746 Chr.2: 46581008-46581008 on Build GRCh38
rs1804166 Chr.2: 46581427-46581427 on Build GRCh38
rs536547507 Chr.2: 46581238-46581238 on Build GRCh38
rs759592600 Chr.2: 46581121-46581121 on Build GRCh38
rs755258293 Chr.2: 46580981-46580981 on Build GRCh38
rs770951853 Chr.2: 46581113-46581113 on Build GRCh38
rs538794415 Chr.2: 46581168-46581168 on Build GRCh38
rs779649857 Chr.2: 46581033-46581033 on Build GRCh38
rs903826517 Chr.2: 46581375-46581375 on Build GRCh38
rs553695421 Chr.2: 46581172-46581172 on Build GRCh38
rs757051621 Chr.2: 46581126-46581126 on Build GRCh38
rs1008299856 Chr.2: 46581384-46581384 on Build GRCh38
rs549446550 Chr.2: 46581144-46581144 on Build GRCh38
rs951763230 Chr.2: 46581222-46581222 on Build GRCh38
rs1019706064 Chr.2: 46581385-46581386 on Build GRCh38
rs897331078 Chr.2: 46581402-46581402 on Build GRCh38
rs917765102 Chr.2: 46581338-46581338 on Build GRCh38
rs771994356 Chr.2: 46581100-46581100 on Build GRCh38
rs765427531 Chr.2: 46580993-46580993 on Build GRCh38
rs149232659 Chr.2: 46581164-46581164 on Build GRCh38
rs747796684 Chr.2: 46581080-46581080 on Build GRCh38
rs367889567 Chr.2: 46581120-46581120 on Build GRCh38
rs61759911 Chr.2: 46581016-46581016 on Build GRCh38
rs746415852 Chr.2: 46581332-46581332 on Build GRCh38
rs55694648 Chr.2: 46581158-46581158 on Build GRCh38
rs35227200 Chr.2: 46581108-46581108 on Build GRCh38
rs549897439 Chr.2: 46581417-46581417 on Build GRCh38
rs773601492 Chr.2: 46581104-46581105 on Build GRCh38
rs376417203 Chr.2: 46581105-46581105 on Build GRCh38
rs773082737 Chr.2: 46581108-46581108 on Build GRCh38
rs572259148 Chr.2: 46581226-46581226 on Build GRCh38
rs566917162 Chr.2: 46581028-46581028 on Build GRCh38
rs752869496 Chr.2: 46581129-46581129 on Build GRCh38
rs760708597 Chr.2: 46581112-46581112 on Build GRCh38
rs780730059 Chr.2: 46581143-46581143 on Build GRCh38
rs991986127 Chr.2: 46581252-46581252 on Build GRCh38

View Full Product Details