Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46699291-46699784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS5
Entrez Gene ID
9655
Gene Name
suppressor of cytokine signaling 5
Gene Aliases
CIS6, CISH6, Cish5, SOCS-5
UniGene
Hs.468426
Gene Chromosome Location
Chr.2: 46698960-46762788 on Build GRCh38
Transcripts
NM_014011.4 NM_144949.2

SNP Detail

SNP ID Chromosome Location
rs895327220 Chr.2: 46699715-46699715 on Build GRCh38
rs936841944 Chr.2: 46699423-46699423 on Build GRCh38
rs1050413108 Chr.2: 46699472-46699472 on Build GRCh38
rs917991039 Chr.2: 46699349-46699349 on Build GRCh38
rs987425399 Chr.2: 46699610-46699610 on Build GRCh38
rs7580441 Chr.2: 46699574-46699574 on Build GRCh38
rs149904494 Chr.2: 46699608-46699608 on Build GRCh38
rs901543988 Chr.2: 46699739-46699739 on Build GRCh38
rs369756135 Chr.2: 46699764-46699764 on Build GRCh38
rs775156440 Chr.2: 46699763-46699763 on Build GRCh38
rs375435039 Chr.2: 46699600-46699600 on Build GRCh38
rs911464757 Chr.2: 46699657-46699657 on Build GRCh38
rs948279436 Chr.2: 46699724-46699724 on Build GRCh38
rs567152063 Chr.2: 46699405-46699405 on Build GRCh38
rs978162700 Chr.2: 46699641-46699641 on Build GRCh38
rs951861526 Chr.2: 46699548-46699548 on Build GRCh38
rs1042502361 Chr.2: 46699393-46699393 on Build GRCh38
rs183910345 Chr.2: 46699753-46699753 on Build GRCh38
rs912516073 Chr.2: 46699624-46699624 on Build GRCh38
rs901510705 Chr.2: 46699530-46699530 on Build GRCh38
rs35219928 Chr.2: 46699712-46699713 on Build GRCh38
rs571357103 Chr.2: 46699508-46699508 on Build GRCh38
rs1053984181 Chr.2: 46699672-46699672 on Build GRCh38
rs1006037295 Chr.2: 46699556-46699556 on Build GRCh38
rs936804391 Chr.2: 46699669-46699669 on Build GRCh38
rs543147992 Chr.2: 46699655-46699655 on Build GRCh38
rs576968120 Chr.2: 46699748-46699748 on Build GRCh38
rs945403430 Chr.2: 46699391-46699391 on Build GRCh38
rs921838736 Chr.2: 46699340-46699340 on Build GRCh38
rs944341143 Chr.2: 46699662-46699662 on Build GRCh38
rs1007630785 Chr.2: 46699506-46699506 on Build GRCh38
rs556002639 Chr.2: 46699503-46699503 on Build GRCh38
rs924060407 Chr.2: 46699651-46699651 on Build GRCh38
rs538817286 Chr.2: 46699526-46699526 on Build GRCh38
rs978147963 Chr.2: 46699637-46699637 on Build GRCh38
rs973236379 Chr.2: 46699337-46699337 on Build GRCh38
rs543832887 Chr.2: 46699640-46699640 on Build GRCh38
rs537919609 Chr.2: 46699498-46699498 on Build GRCh38
rs1046643755 Chr.2: 46699738-46699738 on Build GRCh38
rs554056781 Chr.2: 46699542-46699542 on Build GRCh38
rs959356194 Chr.2: 46699577-46699577 on Build GRCh38
rs34092154 Chr.2: 46699545-46699545 on Build GRCh38
rs1049887372 Chr.2: 46699652-46699652 on Build GRCh38
rs3738890 Chr.2: 46699580-46699580 on Build GRCh38
rs548527818 Chr.2: 46699392-46699392 on Build GRCh38
rs963181787 Chr.2: 46699316-46699316 on Build GRCh38
rs538886774 Chr.2: 46699542-46699543 on Build GRCh38
rs1040080181 Chr.2: 46699516-46699516 on Build GRCh38
rs193120142 Chr.2: 46699747-46699747 on Build GRCh38
rs547340009 Chr.2: 46699691-46699691 on Build GRCh38
rs1014662390 Chr.2: 46699315-46699315 on Build GRCh38
rs1041684817 Chr.2: 46699687-46699687 on Build GRCh38
rs993175111 Chr.2: 46699539-46699539 on Build GRCh38

View Full Product Details