Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 48983020-48983528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSHR
Entrez Gene ID
2492
Gene Name
follicle stimulating hormone receptor
Gene Aliases
FSHR1, FSHRO, LGR1, ODG1
UniGene
Hs.1428
Gene Chromosome Location
Chr.2: 48953161-49154527 on Build GRCh38
Transcripts
NM_000145.3 NM_181446.2 XM_011532733.2 XM_011532734.2 XM_011532735.2 XM_011532736.2 XM_011532740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4426117 c.525-1G>A p.? Chr.2: 48983167-48983167 on Build GRCh38
5702121 c.574_575GG>AT p.G192I Chr.2: 48983116-48983117 on Build GRCh38
4900451 c.547C>T p.Q183* Chr.2: 48983144-48983144 on Build GRCh38
4094530 c.556C>T p.H186Y Chr.2: 48983135-48983135 on Build GRCh38
4553390 c.589G>A p.E197K Chr.2: 48983102-48983102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs72875918 Chr.2: 48983203-48983203 on Build GRCh38
rs1012298762 Chr.2: 48983441-48983441 on Build GRCh38
rs184321544 Chr.2: 48983309-48983309 on Build GRCh38
rs893967947 Chr.2: 48983423-48983423 on Build GRCh38
rs973064249 Chr.2: 48983490-48983490 on Build GRCh38
rs982666103 Chr.2: 48983353-48983353 on Build GRCh38
rs748177242 Chr.2: 48983118-48983118 on Build GRCh38
rs766486126 Chr.2: 48983480-48983480 on Build GRCh38
rs115166339 Chr.2: 48983296-48983296 on Build GRCh38
rs1045860658 Chr.2: 48983445-48983445 on Build GRCh38
rs113141348 Chr.2: 48983245-48983245 on Build GRCh38
rs150508772 Chr.2: 48983097-48983097 on Build GRCh38
rs561783189 Chr.2: 48983255-48983255 on Build GRCh38
rs998481169 Chr.2: 48983470-48983470 on Build GRCh38
rs778691800 Chr.2: 48983076-48983076 on Build GRCh38
rs1014946792 Chr.2: 48983468-48983468 on Build GRCh38
rs902688892 Chr.2: 48983127-48983127 on Build GRCh38
rs1046935846 Chr.2: 48983180-48983180 on Build GRCh38
rs910996749 Chr.2: 48983503-48983503 on Build GRCh38
rs745603080 Chr.2: 48983071-48983071 on Build GRCh38
rs182216404 Chr.2: 48983306-48983306 on Build GRCh38
rs771449297 Chr.2: 48983210-48983210 on Build GRCh38
rs189566420 Chr.2: 48983219-48983219 on Build GRCh38
rs1037185623 Chr.2: 48983050-48983050 on Build GRCh38
rs371985775 Chr.2: 48983495-48983495 on Build GRCh38
rs373416539 Chr.2: 48983079-48983079 on Build GRCh38
rs554503980 Chr.2: 48983465-48983465 on Build GRCh38
rs750383167 Chr.2: 48983178-48983178 on Build GRCh38
rs906529828 Chr.2: 48983462-48983462 on Build GRCh38
rs961261633 Chr.2: 48983477-48983477 on Build GRCh38
rs371078381 Chr.2: 48983193-48983193 on Build GRCh38
rs1041265534 Chr.2: 48983319-48983319 on Build GRCh38
rs776939419 Chr.2: 48983217-48983217 on Build GRCh38
rs908506216 Chr.2: 48983365-48983365 on Build GRCh38
rs756728839 Chr.2: 48983502-48983502 on Build GRCh38
rs761234323 Chr.2: 48983123-48983123 on Build GRCh38
rs121909658 Chr.2: 48983125-48983125 on Build GRCh38
rs878929842 Chr.2: 48983251-48983251 on Build GRCh38
rs769052575 Chr.2: 48983113-48983113 on Build GRCh38
rs115192603 Chr.2: 48983501-48983501 on Build GRCh38
rs568267774 Chr.2: 48983191-48983191 on Build GRCh38
rs376490789 Chr.2: 48983179-48983179 on Build GRCh38
rs760503988 Chr.2: 48983349-48983349 on Build GRCh38
rs751015336 Chr.2: 48983159-48983159 on Build GRCh38
rs763684148 Chr.2: 48983148-48983148 on Build GRCh38
rs964189374 Chr.2: 48983305-48983305 on Build GRCh38
rs970724642 Chr.2: 48983204-48983204 on Build GRCh38
rs1011149705 Chr.2: 48983496-48983496 on Build GRCh38
rs889454073 Chr.2: 48983083-48983083 on Build GRCh38
rs1023758229 Chr.2: 48983460-48983460 on Build GRCh38
rs879062919 Chr.2: 48983258-48983258 on Build GRCh38
rs116271943 Chr.2: 48983250-48983250 on Build GRCh38
rs772569509 Chr.2: 48983119-48983119 on Build GRCh38
rs777027304 Chr.2: 48983130-48983130 on Build GRCh38
rs767115183 Chr.2: 48983177-48983177 on Build GRCh38
rs780813554 Chr.2: 48983387-48983387 on Build GRCh38
rs755961250 Chr.2: 48983082-48983082 on Build GRCh38
rs367812667 Chr.2: 48983469-48983469 on Build GRCh38
rs902800077 Chr.2: 48983355-48983355 on Build GRCh38
rs757178633 Chr.2: 48983062-48983062 on Build GRCh38
rs759567268 Chr.2: 48983150-48983150 on Build GRCh38
rs1038197459 Chr.2: 48983409-48983409 on Build GRCh38
rs572212951 Chr.2: 48983181-48983181 on Build GRCh38
rs374759120 Chr.2: 48983095-48983095 on Build GRCh38
rs777868940 Chr.2: 48983202-48983202 on Build GRCh38
rs149847853 Chr.2: 48983099-48983099 on Build GRCh38
rs1000197541 Chr.2: 48983363-48983363 on Build GRCh38
rs756731404 Chr.2: 48983164-48983164 on Build GRCh38
rs774444032 Chr.2: 48983116-48983116 on Build GRCh38
rs1031223111 Chr.2: 48983263-48983263 on Build GRCh38
rs779012746 Chr.2: 48983195-48983195 on Build GRCh38
rs894260294 Chr.2: 48983205-48983205 on Build GRCh38
rs184839217 Chr.2: 48983201-48983201 on Build GRCh38
rs748458068 Chr.2: 48983196-48983196 on Build GRCh38
rs751159089 Chr.2: 48983298-48983298 on Build GRCh38
rs1053934001 Chr.2: 48983398-48983398 on Build GRCh38
rs752519073 Chr.2: 48983220-48983220 on Build GRCh38
rs762544680 Chr.2: 48983133-48983133 on Build GRCh38
rs751053368 Chr.2: 48983059-48983059 on Build GRCh38
rs780996962 Chr.2: 48983069-48983069 on Build GRCh38
rs961187898 Chr.2: 48983248-48983248 on Build GRCh38
rs1013069678 Chr.2: 48983207-48983207 on Build GRCh38
rs773576455 Chr.2: 48983121-48983121 on Build GRCh38

View Full Product Details