Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112585945-112586411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHCHD5
Entrez Gene ID
84269
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 5
Gene Aliases
C2orf9, MIC14
UniGene
Hs.375707
Gene Chromosome Location
Chr.2: 112583606-112589040 on Build GRCh38
Transcripts
NM_001304353.1 NM_001304354.1 NM_032309.3 XM_005263824.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6311585 c.243G>T p.E81D Chr.2: 112586299-112586299 on Build GRCh38
6153674 c.77A>G p.K26R Chr.2: 112586048-112586048 on Build GRCh38
6424544 c.79C>T p.P27S Chr.2: 112586050-112586050 on Build GRCh38
6346693 c.37G>T p.G13C Chr.2: 112586008-112586008 on Build GRCh38
5584787 c.309+5G>A p.? Chr.2: 112586370-112586370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764035755 Chr.2: 112586177-112586177 on Build GRCh38
rs748040543 Chr.2: 112586334-112586334 on Build GRCh38
rs751278369 Chr.2: 112586323-112586323 on Build GRCh38
rs751457352 Chr.2: 112585984-112585984 on Build GRCh38
rs774566764 Chr.2: 112586302-112586302 on Build GRCh38
rs767512684 Chr.2: 112586069-112586069 on Build GRCh38
rs576226346 Chr.2: 112586289-112586289 on Build GRCh38
rs775381994 Chr.2: 112586220-112586220 on Build GRCh38
rs201426009 Chr.2: 112585980-112585980 on Build GRCh38
rs921232149 Chr.2: 112586067-112586067 on Build GRCh38
rs781155807 Chr.2: 112586333-112586333 on Build GRCh38
rs370320631 Chr.2: 112586170-112586170 on Build GRCh38
rs140990791 Chr.2: 112586111-112586111 on Build GRCh38
rs750317614 Chr.2: 112586107-112586107 on Build GRCh38
rs1054016534 Chr.2: 112586235-112586235 on Build GRCh38
rs554568362 Chr.2: 112586265-112586265 on Build GRCh38
rs771978425 Chr.2: 112586148-112586148 on Build GRCh38
rs945627873 Chr.2: 112586221-112586221 on Build GRCh38
rs756891206 Chr.2: 112586209-112586209 on Build GRCh38
rs367909749 Chr.2: 112586211-112586211 on Build GRCh38
rs371284172 Chr.2: 112586371-112586371 on Build GRCh38
rs199535317 Chr.2: 112586193-112586193 on Build GRCh38
rs13006946 Chr.2: 112586315-112586315 on Build GRCh38
rs759995967 Chr.2: 112586304-112586304 on Build GRCh38
rs771038507 Chr.2: 112586340-112586340 on Build GRCh38
rs768453068 Chr.2: 112586285-112586285 on Build GRCh38
rs753637909 Chr.2: 112586316-112586316 on Build GRCh38
rs770119028 Chr.2: 112586248-112586260 on Build GRCh38
rs754760269 Chr.2: 112586324-112586324 on Build GRCh38
rs766271879 Chr.2: 112585982-112585982 on Build GRCh38
rs148443714 Chr.2: 112586338-112586338 on Build GRCh38
rs746315531 Chr.2: 112586021-112586021 on Build GRCh38
rs146873532 Chr.2: 112586043-112586043 on Build GRCh38
rs79064914 Chr.2: 112586309-112586309 on Build GRCh38
rs183662926 Chr.2: 112586186-112586186 on Build GRCh38
rs1024124466 Chr.2: 112586311-112586311 on Build GRCh38
rs779350982 Chr.2: 112586102-112586102 on Build GRCh38
rs757607382 Chr.2: 112586003-112586003 on Build GRCh38
rs1020098918 Chr.2: 112586058-112586058 on Build GRCh38
rs138729239 Chr.2: 112586210-112586210 on Build GRCh38
rs760719955 Chr.2: 112586076-112586076 on Build GRCh38
rs752810776 Chr.2: 112586074-112586074 on Build GRCh38
rs200306063 Chr.2: 112585979-112585979 on Build GRCh38
rs760577216 Chr.2: 112586122-112586122 on Build GRCh38
rs765834895 Chr.2: 112586372-112586372 on Build GRCh38
rs751960267 Chr.2: 112586201-112586201 on Build GRCh38
rs201779223 Chr.2: 112586351-112586351 on Build GRCh38
rs754654898 Chr.2: 112586369-112586369 on Build GRCh38
rs549517862 Chr.2: 112586369-112586370 on Build GRCh38
rs776556947 Chr.2: 112586240-112586240 on Build GRCh38
rs766796445 Chr.2: 112586200-112586200 on Build GRCh38
rs147407793 Chr.2: 112586310-112586310 on Build GRCh38
rs867391606 Chr.2: 112586239-112586239 on Build GRCh38
rs770663974 Chr.2: 112586052-112586052 on Build GRCh38
rs777363594 Chr.2: 112586123-112586123 on Build GRCh38
rs956662508 Chr.2: 112586036-112586036 on Build GRCh38
rs952520714 Chr.2: 112586306-112586306 on Build GRCh38
rs375517499 Chr.2: 112586282-112586282 on Build GRCh38
rs753326723 Chr.2: 112586270-112586270 on Build GRCh38
rs759710950 Chr.2: 112586343-112586343 on Build GRCh38
rs767527276 Chr.2: 112585995-112585995 on Build GRCh38
rs774147047 Chr.2: 112586062-112586062 on Build GRCh38
rs965271120 Chr.2: 112586065-112586065 on Build GRCh38
rs746876122 Chr.2: 112586284-112586284 on Build GRCh38
rs779454543 Chr.2: 112586009-112586009 on Build GRCh38
rs773045038 Chr.2: 112585976-112585976 on Build GRCh38
rs781227246 Chr.2: 112586286-112586286 on Build GRCh38
rs139481240 Chr.2: 112586341-112586341 on Build GRCh38
rs761107530 Chr.2: 112586354-112586354 on Build GRCh38
rs759492737 Chr.2: 112585994-112585994 on Build GRCh38
rs984808482 Chr.2: 112586191-112586191 on Build GRCh38
rs536960920 Chr.2: 112586037-112586037 on Build GRCh38
rs146213927 Chr.2: 112586292-112586292 on Build GRCh38
rs754210979 Chr.2: 112586000-112586000 on Build GRCh38
rs777080847 Chr.2: 112586204-112586204 on Build GRCh38
rs755436931 Chr.2: 112586264-112586264 on Build GRCh38
rs766038997 Chr.2: 112586132-112586132 on Build GRCh38
rs749444681 Chr.2: 112586339-112586339 on Build GRCh38
rs771251763 Chr.2: 112586301-112586301 on Build GRCh38
rs367672047 Chr.2: 112586345-112586345 on Build GRCh38
rs750880051 Chr.2: 112586103-112586103 on Build GRCh38
rs760701851 Chr.2: 112586160-112586160 on Build GRCh38
rs767574663 Chr.2: 112586098-112586098 on Build GRCh38
rs576592512 Chr.2: 112586252-112586252 on Build GRCh38
rs188390128 Chr.2: 112586244-112586244 on Build GRCh38
rs112498899 Chr.2: 112586115-112586115 on Build GRCh38
rs201729327 Chr.2: 112586042-112586042 on Build GRCh38
rs763201336 Chr.2: 112586299-112586299 on Build GRCh38
rs748529806 Chr.2: 112586218-112586218 on Build GRCh38
rs762697784 Chr.2: 112586051-112586051 on Build GRCh38
rs146594772 Chr.2: 112586066-112586066 on Build GRCh38
rs573555238 Chr.2: 112586272-112586272 on Build GRCh38
rs769258733 Chr.2: 112586141-112586147 on Build GRCh38
rs758871972 Chr.2: 112586195-112586195 on Build GRCh38
rs538755730 Chr.2: 112586099-112586099 on Build GRCh38
rs1043049648 Chr.2: 112586228-112586228 on Build GRCh38
rs771817041 Chr.2: 112586219-112586219 on Build GRCh38
rs557994395 Chr.2: 112586012-112586012 on Build GRCh38
rs773438751 Chr.2: 112586296-112586296 on Build GRCh38
rs199612227 Chr.2: 112586033-112586033 on Build GRCh38
rs755522629 Chr.2: 112586202-112586202 on Build GRCh38
rs750792036 Chr.2: 112586194-112586194 on Build GRCh38
rs780843084 Chr.2: 112586035-112586035 on Build GRCh38
rs777324354 Chr.2: 112586141-112586142 on Build GRCh38
rs373521228 Chr.2: 112586143-112586143 on Build GRCh38
rs751958893 Chr.2: 112586254-112586254 on Build GRCh38
rs752726453 Chr.2: 112585996-112585996 on Build GRCh38
rs764348810 Chr.2: 112586097-112586097 on Build GRCh38
rs376967395 Chr.2: 112586151-112586151 on Build GRCh38
rs542514193 Chr.2: 112586273-112586273 on Build GRCh38
rs200686053 Chr.2: 112586307-112586307 on Build GRCh38
rs199607921 Chr.2: 112585999-112585999 on Build GRCh38
rs768701822 Chr.2: 112586239-112586239 on Build GRCh38
rs770574466 Chr.2: 112586135-112586135 on Build GRCh38
rs370720308 Chr.2: 112586251-112586251 on Build GRCh38
rs764638182 Chr.2: 112586358-112586358 on Build GRCh38
rs746867076 Chr.2: 112586231-112586231 on Build GRCh38

View Full Product Details