Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 113074916-113075429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1F10
Entrez Gene ID
84639
Gene Name
interleukin 1 family member 10 (theta)
Gene Aliases
FIL1-theta, FKSG75, IL-1HY2, IL-38, IL1-theta, IL1HY2
UniGene
Hs.306974
Gene Chromosome Location
Chr.2: 113067970-113075850 on Build GRCh38
Transcripts
NM_032556.5 NM_173161.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6087359 c.285G>A p.E95E Chr.2: 113075190-113075190 on Build GRCh38
6685069 c.292C>T p.R98C Chr.2: 113075197-113075197 on Build GRCh38
5601633 c.324C>T p.S108S Chr.2: 113075229-113075229 on Build GRCh38
3565482 c.459G>A p.*153* Chr.2: 113075364-113075364 on Build GRCh38
5596868 c.330C>T p.F110F Chr.2: 113075235-113075235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143167366 Chr.2: 113075325-113075325 on Build GRCh38
rs1021789690 Chr.2: 113075058-113075058 on Build GRCh38
rs28928299 Chr.2: 113074951-113074951 on Build GRCh38
rs1019836239 Chr.2: 113075012-113075012 on Build GRCh38
rs572330401 Chr.2: 113075090-113075090 on Build GRCh38
rs992245163 Chr.2: 113075105-113075105 on Build GRCh38
rs745485687 Chr.2: 113075370-113075370 on Build GRCh38
rs545065486 Chr.2: 113075051-113075051 on Build GRCh38
rs147863918 Chr.2: 113075253-113075253 on Build GRCh38
rs979175731 Chr.2: 113075087-113075087 on Build GRCh38
rs968910607 Chr.2: 113075080-113075080 on Build GRCh38
rs956141478 Chr.2: 113075062-113075062 on Build GRCh38
rs777879022 Chr.2: 113075378-113075378 on Build GRCh38
rs748868234 Chr.2: 113075272-113075272 on Build GRCh38
rs765324777 Chr.2: 113075397-113075397 on Build GRCh38
rs769214678 Chr.2: 113075238-113075238 on Build GRCh38
rs758709836 Chr.2: 113075217-113075217 on Build GRCh38
rs866046706 Chr.2: 113075152-113075152 on Build GRCh38
rs939921003 Chr.2: 113075146-113075146 on Build GRCh38
rs771970029 Chr.2: 113075271-113075271 on Build GRCh38
rs769267615 Chr.2: 113075117-113075117 on Build GRCh38
rs188134180 Chr.2: 113075071-113075071 on Build GRCh38
rs770351924 Chr.2: 113075252-113075252 on Build GRCh38
rs140273950 Chr.2: 113075332-113075332 on Build GRCh38
rs767451803 Chr.2: 113075166-113075166 on Build GRCh38
rs754697184 Chr.2: 113075377-113075377 on Build GRCh38
rs765267198 Chr.2: 113075299-113075299 on Build GRCh38
rs919752517 Chr.2: 113075189-113075189 on Build GRCh38
rs995536136 Chr.2: 113074944-113074944 on Build GRCh38
rs147026346 Chr.2: 113075162-113075162 on Build GRCh38
rs145244749 Chr.2: 113075200-113075200 on Build GRCh38
rs770673479 Chr.2: 113075158-113075158 on Build GRCh38
rs774013490 Chr.2: 113075256-113075256 on Build GRCh38
rs141416419 Chr.2: 113075046-113075046 on Build GRCh38
rs761890626 Chr.2: 113075292-113075292 on Build GRCh38
rs560179853 Chr.2: 113075195-113075195 on Build GRCh38
rs937660072 Chr.2: 113075099-113075099 on Build GRCh38
rs747481457 Chr.2: 113075236-113075236 on Build GRCh38
rs755376466 Chr.2: 113075026-113075026 on Build GRCh38
rs909545130 Chr.2: 113075245-113075245 on Build GRCh38
rs13005572 Chr.2: 113075404-113075404 on Build GRCh38
rs926370472 Chr.2: 113075097-113075097 on Build GRCh38
rs956509397 Chr.2: 113075025-113075025 on Build GRCh38
rs756742319 Chr.2: 113075333-113075333 on Build GRCh38
rs751975083 Chr.2: 113075320-113075320 on Build GRCh38
rs866624660 Chr.2: 113075255-113075255 on Build GRCh38
rs370412858 Chr.2: 113075138-113075138 on Build GRCh38
rs761701313 Chr.2: 113075395-113075395 on Build GRCh38
rs375634508 Chr.2: 113075373-113075373 on Build GRCh38
rs182666752 Chr.2: 113074957-113074957 on Build GRCh38
rs760744538 Chr.2: 113075183-113075183 on Build GRCh38
rs748772821 Chr.2: 113075251-113075251 on Build GRCh38
rs140754412 Chr.2: 113075230-113075230 on Build GRCh38
rs777133739 Chr.2: 113075249-113075249 on Build GRCh38
rs772250494 Chr.2: 113075202-113075202 on Build GRCh38
rs375615196 Chr.2: 113075229-113075229 on Build GRCh38
rs556293374 Chr.2: 113074971-113074971 on Build GRCh38
rs778336458 Chr.2: 113075360-113075360 on Build GRCh38
rs762506375 Chr.2: 113075142-113075142 on Build GRCh38
rs542845382 Chr.2: 113075198-113075198 on Build GRCh38
rs768342890 Chr.2: 113075381-113075381 on Build GRCh38
rs138974369 Chr.2: 113075197-113075197 on Build GRCh38
rs753554063 Chr.2: 113075292-113075306 on Build GRCh38
rs764239995 Chr.2: 113075190-113075190 on Build GRCh38
rs759284855 Chr.2: 113075265-113075265 on Build GRCh38
rs1022901137 Chr.2: 113074949-113074949 on Build GRCh38
rs780390899 Chr.2: 113075318-113075318 on Build GRCh38
rs1033466501 Chr.2: 113075052-113075052 on Build GRCh38
rs527283732 Chr.2: 113075380-113075380 on Build GRCh38
rs531447533 Chr.2: 113075269-113075269 on Build GRCh38
rs368098427 Chr.2: 113075144-113075144 on Build GRCh38
rs760687262 Chr.2: 113075275-113075275 on Build GRCh38
rs376370944 Chr.2: 113075043-113075043 on Build GRCh38
rs989087887 Chr.2: 113075055-113075055 on Build GRCh38
rs763146014 Chr.2: 113075399-113075399 on Build GRCh38
rs1037205330 Chr.2: 113075308-113075308 on Build GRCh38
rs989309747 Chr.2: 113075088-113075088 on Build GRCh38
rs758654880 Chr.2: 113075310-113075310 on Build GRCh38
rs764152714 Chr.2: 113075279-113075279 on Build GRCh38
rs766529555 Chr.2: 113075401-113075401 on Build GRCh38
rs753735602 Chr.2: 113075280-113075280 on Build GRCh38
rs141551428 Chr.2: 113075171-113075171 on Build GRCh38
rs747532846 Chr.2: 113075103-113075103 on Build GRCh38
rs1019074451 Chr.2: 113075094-113075094 on Build GRCh38
rs548109033 Chr.2: 113075393-113075393 on Build GRCh38
rs551485444 Chr.2: 113075322-113075322 on Build GRCh38
rs142349803 Chr.2: 113075157-113075157 on Build GRCh38
rs779090494 Chr.2: 113075163-113075163 on Build GRCh38
rs750619017 Chr.2: 113075300-113075300 on Build GRCh38
rs796894997 Chr.2: 113075098-113075098 on Build GRCh38

View Full Product Details