Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196706999-196707536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP19
Entrez Gene ID
84984
Gene Name
centrosomal protein 19
Gene Aliases
C3orf34, MOSPGF
UniGene
Hs.282800
Gene Chromosome Location
Chr.3: 196706273-196712294 on Build GRCh38
Transcripts
NM_032898.4 XM_005269370.4 XM_011513246.2

SNP Detail

SNP ID Chromosome Location
rs958940532 Chr.3: 196707274-196707274 on Build GRCh38
rs140467347 Chr.3: 196707088-196707088 on Build GRCh38
rs572178094 Chr.3: 196707467-196707467 on Build GRCh38
rs775256774 Chr.3: 196707031-196707031 on Build GRCh38
rs767568481 Chr.3: 196707030-196707031 on Build GRCh38
rs965516745 Chr.3: 196707416-196707416 on Build GRCh38
rs879902561 Chr.3: 196707276-196707276 on Build GRCh38
rs537895079 Chr.3: 196707100-196707100 on Build GRCh38
rs894293550 Chr.3: 196707310-196707310 on Build GRCh38
rs921404664 Chr.3: 196707186-196707186 on Build GRCh38
rs12635551 Chr.3: 196707134-196707134 on Build GRCh38
rs932923848 Chr.3: 196707188-196707188 on Build GRCh38
rs1035539706 Chr.3: 196707278-196707278 on Build GRCh38
rs778700755 Chr.3: 196707493-196707494 on Build GRCh38
rs7647102 Chr.3: 196707113-196707113 on Build GRCh38
rs777655889 Chr.3: 196707356-196707356 on Build GRCh38
rs945364004 Chr.3: 196707227-196707227 on Build GRCh38
rs561890647 Chr.3: 196707420-196707420 on Build GRCh38
rs67479689 Chr.3: 196707494-196707498 on Build GRCh38
rs369649851 Chr.3: 196707496-196707500 on Build GRCh38
rs2686597 Chr.3: 196707250-196707250 on Build GRCh38
rs542112335 Chr.3: 196707154-196707154 on Build GRCh38
rs982586844 Chr.3: 196707099-196707099 on Build GRCh38
rs1048037423 Chr.3: 196707318-196707318 on Build GRCh38
rs374104352 Chr.3: 196707029-196707030 on Build GRCh38
rs755835866 Chr.3: 196707327-196707327 on Build GRCh38
rs137912040 Chr.3: 196707447-196707447 on Build GRCh38
rs543838784 Chr.3: 196707412-196707412 on Build GRCh38
rs564268574 Chr.3: 196707286-196707286 on Build GRCh38
rs553967861 Chr.3: 196707199-196707199 on Build GRCh38
rs548472759 Chr.3: 196707453-196707453 on Build GRCh38
rs982857372 Chr.3: 196707397-196707397 on Build GRCh38
rs376778944 Chr.3: 196707299-196707299 on Build GRCh38
rs1003598054 Chr.3: 196707273-196707273 on Build GRCh38
rs545971698 Chr.3: 196707251-196707251 on Build GRCh38
rs144560244 Chr.3: 196707330-196707330 on Build GRCh38
rs893837009 Chr.3: 196707219-196707219 on Build GRCh38
rs150165990 Chr.3: 196707093-196707093 on Build GRCh38
rs1004115393 Chr.3: 196707036-196707036 on Build GRCh38
rs970564146 Chr.3: 196707073-196707074 on Build GRCh38
rs1033972221 Chr.3: 196707366-196707366 on Build GRCh38
rs71161952 Chr.3: 196707049-196707049 on Build GRCh38
rs899573315 Chr.3: 196707277-196707277 on Build GRCh38
rs888084666 Chr.3: 196707320-196707320 on Build GRCh38
rs115474023 Chr.3: 196707070-196707070 on Build GRCh38
rs952451688 Chr.3: 196707383-196707383 on Build GRCh38
rs947603200 Chr.3: 196707187-196707187 on Build GRCh38
rs1035485251 Chr.3: 196707030-196707030 on Build GRCh38
rs572271935 Chr.3: 196707217-196707217 on Build GRCh38
rs1026672448 Chr.3: 196707091-196707091 on Build GRCh38
rs747716829 Chr.3: 196707324-196707324 on Build GRCh38
rs1053945255 Chr.3: 196707212-196707212 on Build GRCh38
rs921456962 Chr.3: 196707430-196707430 on Build GRCh38
rs1042518188 Chr.3: 196707231-196707231 on Build GRCh38
rs932809109 Chr.3: 196707442-196707442 on Build GRCh38
rs915478361 Chr.3: 196707455-196707455 on Build GRCh38

View Full Product Details