Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197532371-197532881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDH1
Entrez Gene ID
622
Gene Name
3-hydroxybutyrate dehydrogenase, type 1
Gene Aliases
BDH, SDR9C1
UniGene
Hs.274539
Gene Chromosome Location
Chr.3: 197509783-197573323 on Build GRCh38
Transcripts
NM_004051.4 NM_203314.2 NM_203315.2 XM_005269352.3 XM_011513067.2 XM_017007007.1 XM_017007008.1 XM_017007009.1 XM_017007010.1 XM_017007011.1 XM_017007012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400268 c.267+4A>G p.? Chr.3: 197532408-197532408 on Build GRCh38
6511927 c.223C>T p.L75L Chr.3: 197532456-197532456 on Build GRCh38
1753129 c.267+1G>A p.? Chr.3: 197532411-197532411 on Build GRCh38
4433369 c.261G>A p.L87L Chr.3: 197532418-197532418 on Build GRCh38
5398978 c.205T>C p.F69L Chr.3: 197532474-197532474 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764544382 Chr.3: 197532465-197532465 on Build GRCh38
rs527716653 Chr.3: 197532557-197532557 on Build GRCh38
rs770485161 Chr.3: 197532711-197532711 on Build GRCh38
rs755498143 Chr.3: 197532634-197532634 on Build GRCh38
rs781722562 Chr.3: 197532665-197532665 on Build GRCh38
rs541287538 Chr.3: 197532470-197532470 on Build GRCh38
rs201384972 Chr.3: 197532397-197532397 on Build GRCh38
rs2303981 Chr.3: 197532734-197532734 on Build GRCh38
rs773056463 Chr.3: 197532447-197532447 on Build GRCh38
rs751774782 Chr.3: 197532537-197532537 on Build GRCh38
rs780034347 Chr.3: 197532526-197532526 on Build GRCh38
rs570655773 Chr.3: 197532676-197532676 on Build GRCh38
rs376688866 Chr.3: 197532459-197532459 on Build GRCh38
rs967010509 Chr.3: 197532627-197532627 on Build GRCh38
rs1028030344 Chr.3: 197532514-197532514 on Build GRCh38
rs552140822 Chr.3: 197532624-197532624 on Build GRCh38
rs761310890 Chr.3: 197532632-197532632 on Build GRCh38
rs35643475 Chr.3: 197532544-197532545 on Build GRCh38
rs375898307 Chr.3: 197532409-197532409 on Build GRCh38
rs372692279 Chr.3: 197532403-197532403 on Build GRCh38
rs780438105 Chr.3: 197532469-197532469 on Build GRCh38
rs777745340 Chr.3: 197532560-197532560 on Build GRCh38
rs535794193 Chr.3: 197532813-197532813 on Build GRCh38
rs774470286 Chr.3: 197532532-197532532 on Build GRCh38
rs371395444 Chr.3: 197532432-197532432 on Build GRCh38
rs748062244 Chr.3: 197532426-197532426 on Build GRCh38
rs749223774 Chr.3: 197532563-197532563 on Build GRCh38
rs751901468 Chr.3: 197532565-197532565 on Build GRCh38
rs1048608945 Chr.3: 197532411-197532411 on Build GRCh38
rs1016160819 Chr.3: 197532484-197532484 on Build GRCh38
rs768257000 Chr.3: 197532536-197532536 on Build GRCh38
rs531644829 Chr.3: 197532722-197532722 on Build GRCh38
rs759331947 Chr.3: 197532464-197532464 on Build GRCh38
rs948673690 Chr.3: 197532704-197532704 on Build GRCh38
rs750913959 Chr.3: 197532475-197532475 on Build GRCh38
rs568603163 Chr.3: 197532744-197532744 on Build GRCh38
rs970683936 Chr.3: 197532765-197532765 on Build GRCh38
rs774023746 Chr.3: 197532460-197532460 on Build GRCh38
rs774066108 Chr.3: 197532574-197532574 on Build GRCh38
rs17850831 Chr.3: 197532471-197532471 on Build GRCh38
rs953679402 Chr.3: 197532516-197532516 on Build GRCh38
rs1003465156 Chr.3: 197532781-197532781 on Build GRCh38
rs901308434 Chr.3: 197532797-197532797 on Build GRCh38
rs748540843 Chr.3: 197532683-197532683 on Build GRCh38
rs888711486 Chr.3: 197532446-197532446 on Build GRCh38
rs998314950 Chr.3: 197532837-197532837 on Build GRCh38
rs916008520 Chr.3: 197532700-197532700 on Build GRCh38
rs770471195 Chr.3: 197532570-197532570 on Build GRCh38
rs1015615863 Chr.3: 197532717-197532717 on Build GRCh38
rs1037324222 Chr.3: 197532787-197532787 on Build GRCh38
rs978064996 Chr.3: 197532633-197532633 on Build GRCh38
rs1048895479 Chr.3: 197532732-197532732 on Build GRCh38
rs771227447 Chr.3: 197532402-197532402 on Build GRCh38
rs112167837 Chr.3: 197532468-197532468 on Build GRCh38
rs115448096 Chr.3: 197532502-197532502 on Build GRCh38
rs747869623 Chr.3: 197532556-197532556 on Build GRCh38
rs781404342 Chr.3: 197532545-197532545 on Build GRCh38
rs780271933 Chr.3: 197532402-197532402 on Build GRCh38
rs367568868 Chr.3: 197532427-197532427 on Build GRCh38
rs997887092 Chr.3: 197532504-197532504 on Build GRCh38
rs1022188699 Chr.3: 197532561-197532561 on Build GRCh38
rs755250586 Chr.3: 197532539-197532539 on Build GRCh38
rs781031088 Chr.3: 197532420-197532420 on Build GRCh38
rs1033629380 Chr.3: 197532798-197532798 on Build GRCh38

View Full Product Details