Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 72128506-72129004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPFFR2
Entrez Gene ID
10886
Gene Name
neuropeptide FF receptor 2
Gene Aliases
GPR74, HLWAR77, NPFF2, NPGPR
UniGene
Hs.99231
Gene Chromosome Location
Chr.4: 72031804-72148201 on Build GRCh38
Transcripts
NM_001144756.1 NM_004885.2 NM_053036.2 XM_011531554.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6101017 c.428A>T p.Q143L Chr.4: 72128713-72128713 on Build GRCh38
3605618 c.357G>A p.W119* Chr.4: 72128642-72128642 on Build GRCh38
3605619 c.491G>A p.G164E Chr.4: 72128776-72128776 on Build GRCh38
6878094 c.489G>A p.M163I Chr.4: 72128774-72128774 on Build GRCh38
3975014 c.630A>T p.I210I Chr.4: 72128915-72128915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776768663 Chr.4: 72128877-72128877 on Build GRCh38
rs186834987 Chr.4: 72128879-72128879 on Build GRCh38
rs759418089 Chr.4: 72128570-72128570 on Build GRCh38
rs746585454 Chr.4: 72128628-72128628 on Build GRCh38
rs756907832 Chr.4: 72128920-72128920 on Build GRCh38
rs747933686 Chr.4: 72128946-72128946 on Build GRCh38
rs754399081 Chr.4: 72128680-72128680 on Build GRCh38
rs777610698 Chr.4: 72128722-72128722 on Build GRCh38
rs146896505 Chr.4: 72128631-72128631 on Build GRCh38
rs147754873 Chr.4: 72128591-72128591 on Build GRCh38
rs745827351 Chr.4: 72128751-72128751 on Build GRCh38
rs780648190 Chr.4: 72128592-72128592 on Build GRCh38
rs1000923410 Chr.4: 72128683-72128683 on Build GRCh38
rs768469873 Chr.4: 72128856-72128856 on Build GRCh38
rs202241077 Chr.4: 72128886-72128886 on Build GRCh38
rs530842627 Chr.4: 72128686-72128686 on Build GRCh38
rs772258262 Chr.4: 72128782-72128782 on Build GRCh38
rs769673831 Chr.4: 72128764-72128764 on Build GRCh38
rs143711179 Chr.4: 72128769-72128769 on Build GRCh38
rs544646763 Chr.4: 72128660-72128660 on Build GRCh38
rs780012575 Chr.4: 72128850-72128850 on Build GRCh38
rs200268318 Chr.4: 72128810-72128810 on Build GRCh38
rs201113574 Chr.4: 72128968-72128968 on Build GRCh38
rs148970744 Chr.4: 72128699-72128699 on Build GRCh38
rs773777332 Chr.4: 72128834-72128834 on Build GRCh38
rs750468091 Chr.4: 72128667-72128667 on Build GRCh38
rs369681038 Chr.4: 72128966-72128966 on Build GRCh38
rs571035519 Chr.4: 72128818-72128818 on Build GRCh38
rs758885989 Chr.4: 72128817-72128817 on Build GRCh38
rs779093966 Chr.4: 72128607-72128607 on Build GRCh38
rs879240894 Chr.4: 72128976-72128976 on Build GRCh38
rs565275727 Chr.4: 72128612-72128612 on Build GRCh38
rs903832565 Chr.4: 72128953-72128953 on Build GRCh38
rs764203910 Chr.4: 72128575-72128575 on Build GRCh38
rs767146218 Chr.4: 72128922-72128922 on Build GRCh38
rs201045074 Chr.4: 72128676-72128676 on Build GRCh38
rs758998304 Chr.4: 72128604-72128604 on Build GRCh38
rs1042658806 Chr.4: 72128952-72128952 on Build GRCh38
rs771178484 Chr.4: 72128959-72128959 on Build GRCh38
rs550550481 Chr.4: 72128562-72128563 on Build GRCh38
rs751203458 Chr.4: 72128911-72128911 on Build GRCh38
rs747268905 Chr.4: 72128867-72128867 on Build GRCh38
rs77812923 Chr.4: 72128808-72128808 on Build GRCh38
rs776682784 Chr.4: 72128643-72128643 on Build GRCh38
rs1034526356 Chr.4: 72128796-72128796 on Build GRCh38
rs748287240 Chr.4: 72128554-72128554 on Build GRCh38
rs749256682 Chr.4: 72128938-72128938 on Build GRCh38
rs767592796 Chr.4: 72128588-72128588 on Build GRCh38
rs772836749 Chr.4: 72128627-72128627 on Build GRCh38
rs765823673 Chr.4: 72128884-72128884 on Build GRCh38
rs778656835 Chr.4: 72128944-72128944 on Build GRCh38
rs766484120 Chr.4: 72128561-72128561 on Build GRCh38
rs749031170 Chr.4: 72128854-72128854 on Build GRCh38
rs756659620 Chr.4: 72128822-72128822 on Build GRCh38
rs10004420 Chr.4: 72128579-72128579 on Build GRCh38
rs771116511 Chr.4: 72128875-72128875 on Build GRCh38
rs961589280 Chr.4: 72128780-72128780 on Build GRCh38
rs201647370 Chr.4: 72128646-72128646 on Build GRCh38
rs762773059 Chr.4: 72128773-72128773 on Build GRCh38
rs754719916 Chr.4: 72128942-72128942 on Build GRCh38
rs148112426 Chr.4: 72128805-72128805 on Build GRCh38
rs376456096 Chr.4: 72128906-72128906 on Build GRCh38
rs376287922 Chr.4: 72128735-72128735 on Build GRCh38
rs931047970 Chr.4: 72128568-72128568 on Build GRCh38
rs774620913 Chr.4: 72128576-72128578 on Build GRCh38
rs763339976 Chr.4: 72128890-72128890 on Build GRCh38
rs773444312 Chr.4: 72128802-72128802 on Build GRCh38
rs760474427 Chr.4: 72128755-72128757 on Build GRCh38
rs745432920 Chr.4: 72128838-72128838 on Build GRCh38
rs780735926 Chr.4: 72128826-72128826 on Build GRCh38
rs764581122 Chr.4: 72128891-72128891 on Build GRCh38
rs760075657 Chr.4: 72128809-72128809 on Build GRCh38
rs775114444 Chr.4: 72128718-72128718 on Build GRCh38
rs368846467 Chr.4: 72128611-72128611 on Build GRCh38
rs369957151 Chr.4: 72128539-72128539 on Build GRCh38
rs747807101 Chr.4: 72128618-72128618 on Build GRCh38
rs200033565 Chr.4: 72128692-72128692 on Build GRCh38
rs375838374 Chr.4: 72128541-72128541 on Build GRCh38
rs766483841 Chr.4: 72128807-72128807 on Build GRCh38
rs762005002 Chr.4: 72128661-72128661 on Build GRCh38
rs749938246 Chr.4: 72128924-72128924 on Build GRCh38
rs751291330 Chr.4: 72128819-72128819 on Build GRCh38
rs779902004 Chr.4: 72128930-72128930 on Build GRCh38
rs866940563 Chr.4: 72128623-72128623 on Build GRCh38
rs763956496 Chr.4: 72128815-72128816 on Build GRCh38
rs770499746 Chr.4: 72128740-72128740 on Build GRCh38
rs142617960 Chr.4: 72128620-72128620 on Build GRCh38
rs753765078 Chr.4: 72128916-72128916 on Build GRCh38
rs776773943 Chr.4: 72128961-72128961 on Build GRCh38
rs534362383 Chr.4: 72128882-72128882 on Build GRCh38
rs930992262 Chr.4: 72128815-72128815 on Build GRCh38
rs555150610 Chr.4: 72128557-72128557 on Build GRCh38
rs758120257 Chr.4: 72128711-72128711 on Build GRCh38
rs770931726 Chr.4: 72128569-72128569 on Build GRCh38
rs371873043 Chr.4: 72128816-72128816 on Build GRCh38
rs773354770 Chr.4: 72128857-72128857 on Build GRCh38
rs767391382 Chr.4: 72128611-72128611 on Build GRCh38
rs1024572100 Chr.4: 72128868-72128868 on Build GRCh38
rs759739994 Chr.4: 72128580-72128581 on Build GRCh38
rs1028378472 Chr.4: 72128981-72128981 on Build GRCh38
rs755594947 Chr.4: 72128925-72128925 on Build GRCh38
rs750555908 Chr.4: 72128589-72128589 on Build GRCh38
rs938148114 Chr.4: 72128859-72128859 on Build GRCh38
rs767790869 Chr.4: 72128665-72128665 on Build GRCh38
rs147934711 Chr.4: 72128675-72128675 on Build GRCh38
rs769666553 Chr.4: 72128650-72128650 on Build GRCh38
rs144971184 Chr.4: 72128844-72128844 on Build GRCh38
rs573209191 Chr.4: 72128562-72128562 on Build GRCh38
rs780956076 Chr.4: 72128746-72128746 on Build GRCh38

View Full Product Details