Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 82423943-82424453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPDL
Entrez Gene ID
9987
Gene Name
heterogeneous nuclear ribonucleoprotein D like
Gene Aliases
HNRNP, HNRPDL, JKTBP, JKTBP2, LGMD1G, laAUF1
UniGene
Hs.527105
Gene Chromosome Location
Chr.4: 82422564-82430225 on Build GRCh38
Transcripts
NM_001207000.1 NM_031372.3 NR_003249.2

SNP Detail

SNP ID Chromosome Location
rs72909808 Chr.4: 82424324-82424324 on Build GRCh38
rs1031562855 Chr.4: 82424019-82424022 on Build GRCh38
rs980727092 Chr.4: 82424043-82424047 on Build GRCh38
rs1014836288 Chr.4: 82424286-82424286 on Build GRCh38
rs1026965245 Chr.4: 82424266-82424266 on Build GRCh38
rs1013750152 Chr.4: 82424178-82424179 on Build GRCh38
rs369770953 Chr.4: 82424076-82424076 on Build GRCh38
rs774770092 Chr.4: 82424251-82424251 on Build GRCh38
rs924977705 Chr.4: 82424383-82424383 on Build GRCh38
rs985825006 Chr.4: 82424369-82424369 on Build GRCh38
rs997720 Chr.4: 82423993-82423993 on Build GRCh38
rs1001001656 Chr.4: 82424017-82424017 on Build GRCh38
rs567257660 Chr.4: 82424068-82424071 on Build GRCh38
rs1052687865 Chr.4: 82424158-82424158 on Build GRCh38
rs184276024 Chr.4: 82424202-82424202 on Build GRCh38
rs923043483 Chr.4: 82424098-82424098 on Build GRCh38
rs12644560 Chr.4: 82424234-82424234 on Build GRCh38
rs532153923 Chr.4: 82424125-82424125 on Build GRCh38
rs536468839 Chr.4: 82423989-82423989 on Build GRCh38
rs969414716 Chr.4: 82424036-82424036 on Build GRCh38
rs527471113 Chr.4: 82424338-82424338 on Build GRCh38
rs59154495 Chr.4: 82424003-82424003 on Build GRCh38
rs540060474 Chr.4: 82424084-82424084 on Build GRCh38
rs4572868 Chr.4: 82424157-82424157 on Build GRCh38
rs763569751 Chr.4: 82424175-82424175 on Build GRCh38
rs770160835 Chr.4: 82424313-82424313 on Build GRCh38
rs551179434 Chr.4: 82423983-82423983 on Build GRCh38
rs1034277755 Chr.4: 82424047-82424047 on Build GRCh38
rs953108756 Chr.4: 82424351-82424352 on Build GRCh38
rs936947555 Chr.4: 82424150-82424150 on Build GRCh38
rs551269134 Chr.4: 82424290-82424290 on Build GRCh38
rs1040104471 Chr.4: 82423988-82423988 on Build GRCh38
rs952673991 Chr.4: 82424268-82424268 on Build GRCh38
rs188226035 Chr.4: 82424298-82424298 on Build GRCh38
rs904132019 Chr.4: 82424141-82424141 on Build GRCh38
rs544135967 Chr.4: 82424054-82424054 on Build GRCh38
rs962226516 Chr.4: 82424310-82424310 on Build GRCh38
rs560144338 Chr.4: 82424107-82424107 on Build GRCh38
rs914116703 Chr.4: 82424411-82424411 on Build GRCh38
rs115114284 Chr.4: 82424281-82424281 on Build GRCh38
rs997721 Chr.4: 82424139-82424139 on Build GRCh38
rs892660176 Chr.4: 82424164-82424164 on Build GRCh38
rs753007304 Chr.4: 82424360-82424360 on Build GRCh38
rs1053153883 Chr.4: 82424394-82424394 on Build GRCh38
rs1025480547 Chr.4: 82424200-82424200 on Build GRCh38
rs892615672 Chr.4: 82424021-82424021 on Build GRCh38
rs901533927 Chr.4: 82423991-82423991 on Build GRCh38
rs558576841 Chr.4: 82424024-82424024 on Build GRCh38
rs756511691 Chr.4: 82423968-82423968 on Build GRCh38
rs567877399 Chr.4: 82423985-82423985 on Build GRCh38
rs951916217 Chr.4: 82424053-82424053 on Build GRCh38
rs538979251 Chr.4: 82424009-82424009 on Build GRCh38
rs575178993 Chr.4: 82424028-82424028 on Build GRCh38
rs113676360 Chr.4: 82424386-82424386 on Build GRCh38
rs749500260 Chr.4: 82424353-82424353 on Build GRCh38
rs151020164 Chr.4: 82424078-82424078 on Build GRCh38
rs562254859 Chr.4: 82424330-82424330 on Build GRCh38

View Full Product Details