Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 112087332-112087846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM229B
Entrez Gene ID
619208
Gene Name
family with sequence similarity 229 member B
Gene Aliases
C6orf225
UniGene
Hs.591340
Gene Chromosome Location
Chr.6: 112087471-112102790 on Build GRCh38
Transcripts
NM_001033564.2 XM_017011174.1 XM_017011175.1 NM_016262.4 XM_011535875.2 XM_017010918.1

Gene Symbol
TUBE1
Entrez Gene ID
51175
Gene Name
tubulin epsilon 1
Gene Aliases
TUBE, dJ142L7.2
UniGene
Hs.34851
Gene Chromosome Location
Chr.6: 112070657-112087548 on Build GRCh38
Transcripts
NM_001033564.2 XM_017011174.1 XM_017011175.1 NM_016262.4 XM_011535875.2 XM_017010918.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7210699 c.3G>A p.M1I Chr.6: 112087432-112087432 on Build GRCh38
7240579 c.25+3A>G p.? Chr.6: 112087407-112087407 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs140996744 Chr.6: 112087763-112087763 on Build GRCh38
rs199513778 Chr.6: 112087425-112087425 on Build GRCh38
rs587689817 Chr.6: 112087362-112087362 on Build GRCh38
rs894623940 Chr.6: 112087470-112087470 on Build GRCh38
rs1057243564 Chr.6: 112087457-112087457 on Build GRCh38
rs1013205132 Chr.6: 112087490-112087490 on Build GRCh38
rs587668145 Chr.6: 112087581-112087581 on Build GRCh38
rs587623185 Chr.6: 112087449-112087449 on Build GRCh38
rs954788763 Chr.6: 112087596-112087596 on Build GRCh38
rs782485988 Chr.6: 112087475-112087475 on Build GRCh38
rs183381227 Chr.6: 112087814-112087814 on Build GRCh38
rs192322895 Chr.6: 112087805-112087805 on Build GRCh38
rs1019467252 Chr.6: 112087618-112087618 on Build GRCh38
rs6568714 Chr.6: 112087407-112087407 on Build GRCh38
rs782704182 Chr.6: 112087434-112087434 on Build GRCh38
rs187692223 Chr.6: 112087671-112087671 on Build GRCh38
rs587594929 Chr.6: 112087499-112087499 on Build GRCh38
rs771138356 Chr.6: 112087454-112087454 on Build GRCh38
rs200986359 Chr.6: 112087378-112087378 on Build GRCh38
rs768611246 Chr.6: 112087551-112087551 on Build GRCh38
rs781862050 Chr.6: 112087532-112087532 on Build GRCh38
rs147330886 Chr.6: 112087436-112087436 on Build GRCh38
rs904737943 Chr.6: 112087505-112087505 on Build GRCh38
rs782670287 Chr.6: 112087479-112087479 on Build GRCh38
rs200357896 Chr.6: 112087423-112087423 on Build GRCh38
rs1046680469 Chr.6: 112087437-112087437 on Build GRCh38
rs587642779 Chr.6: 112087406-112087406 on Build GRCh38
rs182369129 Chr.6: 112087456-112087456 on Build GRCh38
rs587604370 Chr.6: 112087717-112087717 on Build GRCh38
rs372441271 Chr.6: 112087386-112087386 on Build GRCh38
rs781917485 Chr.6: 112087381-112087381 on Build GRCh38
rs782301514 Chr.6: 112087368-112087368 on Build GRCh38
rs782225039 Chr.6: 112087389-112087389 on Build GRCh38
rs782014165 Chr.6: 112087424-112087424 on Build GRCh38
rs587740657 Chr.6: 112087585-112087585 on Build GRCh38
rs202220859 Chr.6: 112087474-112087474 on Build GRCh38
rs1031653110 Chr.6: 112087552-112087552 on Build GRCh38
rs587667152 Chr.6: 112087790-112087790 on Build GRCh38
rs964218488 Chr.6: 112087634-112087634 on Build GRCh38
rs759580602 Chr.6: 112087510-112087510 on Build GRCh38
rs908191458 Chr.6: 112087755-112087755 on Build GRCh38
rs948905115 Chr.6: 112087804-112087804 on Build GRCh38
rs782636355 Chr.6: 112087366-112087366 on Build GRCh38
rs782267114 Chr.6: 112087651-112087651 on Build GRCh38
rs782103653 Chr.6: 112087417-112087417 on Build GRCh38
rs987540377 Chr.6: 112087612-112087612 on Build GRCh38
rs782336686 Chr.6: 112087390-112087390 on Build GRCh38
rs781834144 Chr.6: 112087384-112087384 on Build GRCh38
rs975390227 Chr.6: 112087666-112087666 on Build GRCh38
rs781862571 Chr.6: 112087460-112087460 on Build GRCh38
rs587674269 Chr.6: 112087725-112087725 on Build GRCh38
rs986649833 Chr.6: 112087396-112087396 on Build GRCh38

View Full Product Details