Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 4857735-4858276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAPOLB
Entrez Gene ID
56903
Gene Name
poly(A) polymerase beta
Gene Aliases
PAPT, TPAP
UniGene
Hs.487409
Gene Chromosome Location
Chr.7: 4857738-4861994 on Build GRCh38
Transcripts
NM_020144.4 NM_018059.4

Gene Symbol
RADIL
Entrez Gene ID
55698
Gene Name
Rap associating with DIL domain
Gene Aliases
RASIP2
UniGene
Hs.667336
Gene Chromosome Location
Chr.7: 4799109-4883704 on Build GRCh38
Transcripts
NM_020144.4 NM_018059.4

SNP Detail

SNP ID Chromosome Location
rs981583421 Chr.7: 4857955-4857956 on Build GRCh38
rs1027977844 Chr.7: 4858137-4858137 on Build GRCh38
rs776633669 Chr.7: 4858143-4858143 on Build GRCh38
rs529280090 Chr.7: 4857891-4857891 on Build GRCh38
rs1003974354 Chr.7: 4858104-4858104 on Build GRCh38
rs756185793 Chr.7: 4858047-4858047 on Build GRCh38
rs765846399 Chr.7: 4858172-4858172 on Build GRCh38
rs553811499 Chr.7: 4858110-4858110 on Build GRCh38
rs926604740 Chr.7: 4858249-4858249 on Build GRCh38
rs940823663 Chr.7: 4857938-4857941 on Build GRCh38
rs996251034 Chr.7: 4858072-4858072 on Build GRCh38
rs908139870 Chr.7: 4857903-4857903 on Build GRCh38
rs986345166 Chr.7: 4857957-4857957 on Build GRCh38
rs543293192 Chr.7: 4858147-4858147 on Build GRCh38
rs1046263149 Chr.7: 4857822-4857822 on Build GRCh38
rs1015143594 Chr.7: 4858103-4858103 on Build GRCh38
rs982102483 Chr.7: 4858243-4858243 on Build GRCh38
rs188215866 Chr.7: 4857934-4857934 on Build GRCh38
rs932100236 Chr.7: 4857771-4857771 on Build GRCh38
rs949460664 Chr.7: 4858240-4858240 on Build GRCh38
rs905035909 Chr.7: 4857848-4857848 on Build GRCh38
rs988600414 Chr.7: 4857998-4857998 on Build GRCh38
rs184233322 Chr.7: 4858141-4858141 on Build GRCh38
rs1004347908 Chr.7: 4857890-4857890 on Build GRCh38
rs902428841 Chr.7: 4858053-4858053 on Build GRCh38
rs138053291 Chr.7: 4857892-4857892 on Build GRCh38
rs761162276 Chr.7: 4858093-4858093 on Build GRCh38
rs1015721688 Chr.7: 4858108-4858108 on Build GRCh38
rs936738090 Chr.7: 4857798-4857798 on Build GRCh38
rs894796753 Chr.7: 4858016-4858016 on Build GRCh38
rs6966356 Chr.7: 4858129-4858129 on Build GRCh38
rs191717025 Chr.7: 4857793-4857793 on Build GRCh38
rs182070872 Chr.7: 4858202-4858202 on Build GRCh38
rs1027221610 Chr.7: 4857864-4857864 on Build GRCh38
rs1052469443 Chr.7: 4857800-4857800 on Build GRCh38
rs191422861 Chr.7: 4858139-4858139 on Build GRCh38
rs1022658101 Chr.7: 4857781-4857781 on Build GRCh38
rs913911308 Chr.7: 4858003-4858003 on Build GRCh38
rs547472149 Chr.7: 4857994-4857994 on Build GRCh38
rs552017549 Chr.7: 4858233-4858233 on Build GRCh38
rs879574497 Chr.7: 4858036-4858036 on Build GRCh38
rs77021497 Chr.7: 4858085-4858085 on Build GRCh38
rs958411251 Chr.7: 4858171-4858171 on Build GRCh38
rs771006610 Chr.7: 4857947-4857947 on Build GRCh38
rs369967134 Chr.7: 4858071-4858071 on Build GRCh38
rs925424667 Chr.7: 4857966-4857966 on Build GRCh38
rs901649314 Chr.7: 4858194-4858194 on Build GRCh38
rs190831903 Chr.7: 4858142-4858142 on Build GRCh38
rs780981087 Chr.7: 4857785-4857785 on Build GRCh38
rs928841848 Chr.7: 4857973-4857973 on Build GRCh38
rs772242783 Chr.7: 4858067-4858067 on Build GRCh38
rs888229384 Chr.7: 4857882-4857882 on Build GRCh38
rs1040345576 Chr.7: 4858185-4858185 on Build GRCh38
rs535404904 Chr.7: 4858109-4858109 on Build GRCh38
rs576629615 Chr.7: 4857862-4857862 on Build GRCh38
rs562026566 Chr.7: 4857813-4857813 on Build GRCh38
rs1032521945 Chr.7: 4858153-4858153 on Build GRCh38
rs564130752 Chr.7: 4857772-4857772 on Build GRCh38
rs561733292 Chr.7: 4857874-4857874 on Build GRCh38
rs901703075 Chr.7: 4858062-4858062 on Build GRCh38
rs35559543 Chr.7: 4857902-4857902 on Build GRCh38
rs949347370 Chr.7: 4858032-4858032 on Build GRCh38
rs777723712 Chr.7: 4857871-4857871 on Build GRCh38
rs751860007 Chr.7: 4858150-4858150 on Build GRCh38
rs183881237 Chr.7: 4857917-4857917 on Build GRCh38
rs780484622 Chr.7: 4858125-4858125 on Build GRCh38
rs1029506558 Chr.7: 4858079-4858079 on Build GRCh38
rs11972309 Chr.7: 4858099-4858099 on Build GRCh38
rs976334773 Chr.7: 4857898-4857898 on Build GRCh38
rs949314758 Chr.7: 4857818-4857818 on Build GRCh38
rs974856407 Chr.7: 4857851-4857851 on Build GRCh38
rs11972290 Chr.7: 4858056-4858056 on Build GRCh38
rs1043789781 Chr.7: 4858163-4858163 on Build GRCh38
rs561623736 Chr.7: 4858181-4858181 on Build GRCh38

View Full Product Details