Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 76328688-76329197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAG
Entrez Gene ID
7532
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma
Gene Aliases
14-3-3GAMMA, PPP1R170
UniGene
Hs.744840
Gene Chromosome Location
Chr.7: 76326791-76359025 on Build GRCh38
Transcripts
NM_012479.3

SNP Detail

SNP ID Chromosome Location
rs760535528 Chr.7: 76328988-76328988 on Build GRCh38
rs553097822 Chr.7: 76329123-76329123 on Build GRCh38
rs1015130896 Chr.7: 76328858-76328858 on Build GRCh38
rs947581910 Chr.7: 76328989-76328989 on Build GRCh38
rs11541901 Chr.7: 76329033-76329033 on Build GRCh38
rs550098710 Chr.7: 76328837-76328837 on Build GRCh38
rs902004261 Chr.7: 76328906-76328906 on Build GRCh38
rs571501867 Chr.7: 76328946-76328946 on Build GRCh38
rs566056923 Chr.7: 76329125-76329125 on Build GRCh38
rs776743778 Chr.7: 76329049-76329049 on Build GRCh38
rs566614979 Chr.7: 76329128-76329128 on Build GRCh38
rs1008695798 Chr.7: 76328732-76328732 on Build GRCh38
rs981588596 Chr.7: 76328866-76328866 on Build GRCh38
rs563326860 Chr.7: 76328739-76328739 on Build GRCh38
rs961910822 Chr.7: 76328865-76328865 on Build GRCh38
rs1040659534 Chr.7: 76329132-76329132 on Build GRCh38
rs996899173 Chr.7: 76329091-76329091 on Build GRCh38
rs1028853562 Chr.7: 76328836-76328836 on Build GRCh38
rs771897820 Chr.7: 76328947-76328947 on Build GRCh38
rs972499085 Chr.7: 76328814-76328814 on Build GRCh38
rs1053668885 Chr.7: 76329174-76329174 on Build GRCh38
rs1026918029 Chr.7: 76329093-76329093 on Build GRCh38
rs200338085 Chr.7: 76328799-76328799 on Build GRCh38
rs11541900 Chr.7: 76329080-76329080 on Build GRCh38
rs899955816 Chr.7: 76329058-76329058 on Build GRCh38
rs928452949 Chr.7: 76328779-76328779 on Build GRCh38
rs141819632 Chr.7: 76328806-76328807 on Build GRCh38
rs985167735 Chr.7: 76329088-76329088 on Build GRCh38
rs1002742231 Chr.7: 76329012-76329012 on Build GRCh38
rs749019654 Chr.7: 76328839-76328839 on Build GRCh38
rs143794132 Chr.7: 76328721-76328721 on Build GRCh38
rs770729884 Chr.7: 76329032-76329032 on Build GRCh38
rs547444143 Chr.7: 76329070-76329070 on Build GRCh38
rs998990053 Chr.7: 76328921-76328921 on Build GRCh38
rs934865377 Chr.7: 76329164-76329164 on Build GRCh38
rs772819920 Chr.7: 76328965-76328965 on Build GRCh38
rs919633998 Chr.7: 76328818-76328818 on Build GRCh38
rs928406331 Chr.7: 76328916-76328916 on Build GRCh38
rs893187833 Chr.7: 76328973-76328973 on Build GRCh38
rs996825661 Chr.7: 76328780-76328780 on Build GRCh38
rs777215284 Chr.7: 76329040-76329040 on Build GRCh38
rs969971072 Chr.7: 76328991-76328991 on Build GRCh38
rs969918565 Chr.7: 76328731-76328731 on Build GRCh38
rs888449760 Chr.7: 76329114-76329114 on Build GRCh38
rs1040607204 Chr.7: 76328890-76328890 on Build GRCh38
rs532569269 Chr.7: 76328977-76328977 on Build GRCh38
rs952557834 Chr.7: 76329084-76329084 on Build GRCh38
rs112558016 Chr.7: 76328755-76328755 on Build GRCh38
rs1011588824 Chr.7: 76328979-76328979 on Build GRCh38
rs984742738 Chr.7: 76328847-76328847 on Build GRCh38
rs768203272 Chr.7: 76329013-76329013 on Build GRCh38
rs78994924 Chr.7: 76328826-76328826 on Build GRCh38
rs868341084 Chr.7: 76328920-76328920 on Build GRCh38
rs923479681 Chr.7: 76329150-76329150 on Build GRCh38
rs964011535 Chr.7: 76328770-76328770 on Build GRCh38
rs571008155 Chr.7: 76328964-76328964 on Build GRCh38
rs147277079 Chr.7: 76328793-76328793 on Build GRCh38
rs202129187 Chr.7: 76328799-76328800 on Build GRCh38
rs68159547 Chr.7: 76329068-76329069 on Build GRCh38
rs11270080 Chr.7: 76328826-76328827 on Build GRCh38
rs186913497 Chr.7: 76328735-76328735 on Build GRCh38
rs33915038 Chr.7: 76328827-76328828 on Build GRCh38
rs550858903 Chr.7: 76328849-76328849 on Build GRCh38

View Full Product Details