Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 80761198-80761699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA3C
Entrez Gene ID
10512
Gene Name
semaphorin 3C
Gene Aliases
SEMAE, SemE
UniGene
Hs.269109
Gene Chromosome Location
Chr.7: 80742538-80919351 on Build GRCh38
Transcripts
NM_006379.3 XM_005250113.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3412330 c.1485G>T p.K495N Chr.7: 80761616-80761616 on Build GRCh38
4949113 c.1532C>A p.S511Y Chr.7: 80761623-80761623 on Build GRCh38
5652155 c.1520T>C p.M507T Chr.7: 80761635-80761635 on Build GRCh38
3412331 c.1539G>T p.K513N Chr.7: 80761616-80761616 on Build GRCh38
1452228 c.1478C>A p.S493Y Chr.7: 80761623-80761623 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551455674 Chr.7: 80761269-80761269 on Build GRCh38
rs200092266 Chr.7: 80761594-80761594 on Build GRCh38
rs1019438226 Chr.7: 80761389-80761389 on Build GRCh38
rs140093019 Chr.7: 80761337-80761337 on Build GRCh38
rs1026735841 Chr.7: 80761653-80761653 on Build GRCh38
rs934888005 Chr.7: 80761524-80761524 on Build GRCh38
rs778809396 Chr.7: 80761575-80761575 on Build GRCh38
rs1012647528 Chr.7: 80761382-80761382 on Build GRCh38
rs201774662 Chr.7: 80761582-80761582 on Build GRCh38
rs772041243 Chr.7: 80761591-80761591 on Build GRCh38
rs10230880 Chr.7: 80761486-80761486 on Build GRCh38
rs145505943 Chr.7: 80761523-80761523 on Build GRCh38
rs138970370 Chr.7: 80761650-80761650 on Build GRCh38
rs1009434928 Chr.7: 80761565-80761567 on Build GRCh38
rs147947863 Chr.7: 80761321-80761321 on Build GRCh38
rs370090437 Chr.7: 80761479-80761479 on Build GRCh38
rs776010387 Chr.7: 80761609-80761609 on Build GRCh38
rs755166857 Chr.7: 80761656-80761656 on Build GRCh38
rs547833975 Chr.7: 80761493-80761493 on Build GRCh38
rs776637350 Chr.7: 80761480-80761480 on Build GRCh38
rs910682787 Chr.7: 80761263-80761263 on Build GRCh38
rs915220210 Chr.7: 80761518-80761518 on Build GRCh38
rs111687310 Chr.7: 80761585-80761585 on Build GRCh38
rs751764231 Chr.7: 80761655-80761655 on Build GRCh38
rs890539033 Chr.7: 80761355-80761355 on Build GRCh38
rs971212589 Chr.7: 80761403-80761403 on Build GRCh38
rs1001759779 Chr.7: 80761417-80761432 on Build GRCh38
rs561038588 Chr.7: 80761433-80761433 on Build GRCh38
rs763277547 Chr.7: 80761648-80761648 on Build GRCh38
rs982860649 Chr.7: 80761456-80761456 on Build GRCh38
rs908168822 Chr.7: 80761464-80761464 on Build GRCh38
rs554169436 Chr.7: 80761226-80761226 on Build GRCh38
rs527516499 Chr.7: 80761402-80761402 on Build GRCh38
rs762343931 Chr.7: 80761628-80761628 on Build GRCh38
rs565039313 Chr.7: 80761393-80761393 on Build GRCh38
rs919631489 Chr.7: 80761222-80761222 on Build GRCh38
rs765687804 Chr.7: 80761642-80761642 on Build GRCh38
rs115740642 Chr.7: 80761267-80761267 on Build GRCh38
rs1050574759 Chr.7: 80761228-80761228 on Build GRCh38
rs537497532 Chr.7: 80761292-80761292 on Build GRCh38
rs1008947041 Chr.7: 80761362-80761362 on Build GRCh38
rs551393687 Chr.7: 80761277-80761277 on Build GRCh38
rs750981812 Chr.7: 80761647-80761647 on Build GRCh38
rs768156116 Chr.7: 80761608-80761608 on Build GRCh38
rs770775213 Chr.7: 80761572-80761572 on Build GRCh38
rs529281112 Chr.7: 80761469-80761469 on Build GRCh38
rs775096564 Chr.7: 80761528-80761528 on Build GRCh38
rs992589288 Chr.7: 80761476-80761476 on Build GRCh38
rs541273206 Chr.7: 80761408-80761408 on Build GRCh38
rs1031570142 Chr.7: 80761443-80761443 on Build GRCh38
rs890453811 Chr.7: 80761544-80761544 on Build GRCh38
rs746810488 Chr.7: 80761338-80761338 on Build GRCh38
rs531656830 Chr.7: 80761552-80761552 on Build GRCh38
rs10246081 Chr.7: 80761282-80761282 on Build GRCh38
rs904552486 Chr.7: 80761333-80761333 on Build GRCh38
rs749188360 Chr.7: 80761567-80761567 on Build GRCh38

View Full Product Details