Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81950011-81950507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA2D1
Entrez Gene ID
781
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 1
Gene Aliases
CACNA2, CACNL2A, CCHL2A, LINC01112, lncRNA-N3
UniGene
Hs.282151
Gene Chromosome Location
Chr.7: 81946444-82443921 on Build GRCh38
Transcripts
NM_000722.3 XM_005250570.2 XM_005250572.2 XM_005250573.2 XM_005250574.2 XM_006716118.2 XM_006716119.3 XM_006716120.3 XM_006716121.3 XM_011516571.2 XM_011516572.2 XM_017012588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4798064 c.3176G>A p.C1059Y Chr.7: 81950492-81950492 on Build GRCh38
7424870 c.3266G>A p.R1089H Chr.7: 81950402-81950402 on Build GRCh38
6202233 c.3238C>G p.L1080V Chr.7: 81950430-81950430 on Build GRCh38
5434340 c.3270G>A p.L1090L Chr.7: 81950398-81950398 on Build GRCh38
6178720 c.3256A>T p.S1086C Chr.7: 81950412-81950412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956966352 Chr.7: 81950456-81950456 on Build GRCh38
rs1022583971 Chr.7: 81950160-81950160 on Build GRCh38
rs897655199 Chr.7: 81950058-81950058 on Build GRCh38
rs747864706 Chr.7: 81950434-81950434 on Build GRCh38
rs988301316 Chr.7: 81950208-81950208 on Build GRCh38
rs878992489 Chr.7: 81950207-81950207 on Build GRCh38
rs370508103 Chr.7: 81950244-81950244 on Build GRCh38
rs376529606 Chr.7: 81950409-81950409 on Build GRCh38
rs1042396091 Chr.7: 81950291-81950291 on Build GRCh38
rs769366182 Chr.7: 81950350-81950350 on Build GRCh38
rs758341913 Chr.7: 81950416-81950416 on Build GRCh38
rs550794714 Chr.7: 81950403-81950403 on Build GRCh38
rs867740907 Chr.7: 81950059-81950059 on Build GRCh38
rs751857100 Chr.7: 81950380-81950381 on Build GRCh38
rs776265892 Chr.7: 81950341-81950341 on Build GRCh38
rs201437426 Chr.7: 81950337-81950337 on Build GRCh38
rs768028784 Chr.7: 81950467-81950467 on Build GRCh38
rs901169412 Chr.7: 81950309-81950309 on Build GRCh38
rs146250893 Chr.7: 81950450-81950450 on Build GRCh38
rs771497866 Chr.7: 81950458-81950458 on Build GRCh38
rs1027829408 Chr.7: 81950112-81950112 on Build GRCh38
rs759988243 Chr.7: 81950394-81950394 on Build GRCh38
rs555987555 Chr.7: 81950217-81950218 on Build GRCh38
rs181262965 Chr.7: 81950073-81950073 on Build GRCh38
rs897712038 Chr.7: 81950315-81950315 on Build GRCh38
rs901453927 Chr.7: 81950048-81950048 on Build GRCh38
rs1022635947 Chr.7: 81950400-81950400 on Build GRCh38
rs890005272 Chr.7: 81950393-81950393 on Build GRCh38
rs954347299 Chr.7: 81950076-81950076 on Build GRCh38
rs773549800 Chr.7: 81950354-81950354 on Build GRCh38
rs576908848 Chr.7: 81950232-81950232 on Build GRCh38
rs545573864 Chr.7: 81950236-81950236 on Build GRCh38
rs766856402 Chr.7: 81950389-81950389 on Build GRCh38
rs762438944 Chr.7: 81950455-81950455 on Build GRCh38
rs865966579 Chr.7: 81950270-81950270 on Build GRCh38
rs993320257 Chr.7: 81950095-81950095 on Build GRCh38
rs969647150 Chr.7: 81950182-81950182 on Build GRCh38
rs561652573 Chr.7: 81950330-81950330 on Build GRCh38
rs946779224 Chr.7: 81950251-81950251 on Build GRCh38
rs910562402 Chr.7: 81950269-81950269 on Build GRCh38
rs986162670 Chr.7: 81950252-81950252 on Build GRCh38
rs143491535 Chr.7: 81950370-81950370 on Build GRCh38
rs1035241865 Chr.7: 81950205-81950205 on Build GRCh38
rs557232938 Chr.7: 81950213-81950213 on Build GRCh38
rs777880132 Chr.7: 81950410-81950410 on Build GRCh38
rs759935042 Chr.7: 81950465-81950465 on Build GRCh38
rs887521771 Chr.7: 81950122-81950122 on Build GRCh38
rs138331326 Chr.7: 81950414-81950414 on Build GRCh38
rs768405533 Chr.7: 81950288-81950288 on Build GRCh38
rs753944670 Chr.7: 81950485-81950485 on Build GRCh38
rs1040332443 Chr.7: 81950274-81950274 on Build GRCh38
rs961086786 Chr.7: 81950178-81950178 on Build GRCh38
rs769383235 Chr.7: 81950440-81950440 on Build GRCh38
rs148861982 Chr.7: 81950316-81950316 on Build GRCh38
rs572486752 Chr.7: 81950275-81950275 on Build GRCh38
rs935833459 Chr.7: 81950235-81950235 on Build GRCh38
rs75370027 Chr.7: 81950198-81950198 on Build GRCh38
rs983927463 Chr.7: 81950090-81950090 on Build GRCh38
rs146833935 Chr.7: 81950039-81950039 on Build GRCh38
rs927185827 Chr.7: 81950215-81950215 on Build GRCh38
rs143631985 Chr.7: 81950402-81950402 on Build GRCh38
rs546875572 Chr.7: 81950140-81950142 on Build GRCh38
rs916899552 Chr.7: 81950185-81950185 on Build GRCh38
rs78368203 Chr.7: 81950272-81950272 on Build GRCh38
rs761167589 Chr.7: 81950342-81950342 on Build GRCh38
rs775016690 Chr.7: 81950459-81950459 on Build GRCh38
rs887563392 Chr.7: 81950372-81950372 on Build GRCh38
rs575156119 Chr.7: 81950148-81950148 on Build GRCh38
rs773980191 Chr.7: 81950426-81950426 on Build GRCh38
rs998826839 Chr.7: 81950052-81950052 on Build GRCh38
rs767791257 Chr.7: 81950395-81950395 on Build GRCh38
rs760927013 Chr.7: 81950469-81950469 on Build GRCh38
rs998482426 Chr.7: 81950310-81950310 on Build GRCh38
rs142846778 Chr.7: 81950430-81950430 on Build GRCh38
rs1024010075 Chr.7: 81950460-81950460 on Build GRCh38
rs1054264486 Chr.7: 81950247-81950247 on Build GRCh38
rs780771206 Chr.7: 81950433-81950433 on Build GRCh38
rs780032825 Chr.7: 81950421-81950421 on Build GRCh38
rs555316465 Chr.7: 81950102-81950102 on Build GRCh38

View Full Product Details