Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130275058-130275554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA2
Entrez Gene ID
1358
Gene Name
carboxypeptidase A2
Gene Aliases
-
UniGene
Hs.490038
Gene Chromosome Location
Chr.7: 130266863-130289798 on Build GRCh38
Transcripts
NM_001869.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1202119 c.577G>A p.A193T Chr.7: 130275239-130275239 on Build GRCh38
3922696 c.565G>A p.A189T Chr.7: 130275227-130275227 on Build GRCh38
5100082 c.585+7T>G p.? Chr.7: 130275254-130275254 on Build GRCh38
744563 c.497G>T p.G166V Chr.7: 130275159-130275159 on Build GRCh38
6039530 c.585+5delT p.? Chr.7: 130275252-130275252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373847393 Chr.7: 130275283-130275283 on Build GRCh38
rs952383761 Chr.7: 130275482-130275482 on Build GRCh38
rs758490346 Chr.7: 130275187-130275187 on Build GRCh38
rs980652203 Chr.7: 130275281-130275281 on Build GRCh38
rs546145508 Chr.7: 130275363-130275363 on Build GRCh38
rs201564383 Chr.7: 130275219-130275219 on Build GRCh38
rs1016855053 Chr.7: 130275434-130275434 on Build GRCh38
rs751623298 Chr.7: 130275291-130275291 on Build GRCh38
rs905305007 Chr.7: 130275185-130275185 on Build GRCh38
rs559954358 Chr.7: 130275338-130275338 on Build GRCh38
rs983415882 Chr.7: 130275504-130275504 on Build GRCh38
rs536756352 Chr.7: 130275454-130275454 on Build GRCh38
rs1041127821 Chr.7: 130275126-130275126 on Build GRCh38
rs932347279 Chr.7: 130275357-130275357 on Build GRCh38
rs926575192 Chr.7: 130275302-130275302 on Build GRCh38
rs913627594 Chr.7: 130275315-130275315 on Build GRCh38
rs768919734 Chr.7: 130275275-130275275 on Build GRCh38
rs556375275 Chr.7: 130275429-130275429 on Build GRCh38
rs371899087 Chr.7: 130275158-130275158 on Build GRCh38
rs963788480 Chr.7: 130275443-130275443 on Build GRCh38
rs61731658 Chr.7: 130275176-130275176 on Build GRCh38
rs182470836 Chr.7: 130275473-130275473 on Build GRCh38
rs370205787 Chr.7: 130275152-130275152 on Build GRCh38
rs540960723 Chr.7: 130275263-130275264 on Build GRCh38
rs71178570 Chr.7: 130275518-130275518 on Build GRCh38
rs771722562 Chr.7: 130275218-130275218 on Build GRCh38
rs761617917 Chr.7: 130275448-130275448 on Build GRCh38
rs1008075143 Chr.7: 130275397-130275397 on Build GRCh38
rs958213038 Chr.7: 130275525-130275525 on Build GRCh38
rs764099935 Chr.7: 130275191-130275191 on Build GRCh38
rs200668634 Chr.7: 130275208-130275208 on Build GRCh38
rs773305864 Chr.7: 130275127-130275127 on Build GRCh38
rs757775927 Chr.7: 130275198-130275198 on Build GRCh38
rs577061739 Chr.7: 130275244-130275244 on Build GRCh38
rs751234114 Chr.7: 130275203-130275203 on Build GRCh38
rs566939499 Chr.7: 130275474-130275474 on Build GRCh38
rs553500258 Chr.7: 130275199-130275199 on Build GRCh38
rs898019866 Chr.7: 130275469-130275469 on Build GRCh38
rs775700359 Chr.7: 130275239-130275239 on Build GRCh38
rs1011875521 Chr.7: 130275255-130275255 on Build GRCh38
rs567366821 Chr.7: 130275508-130275508 on Build GRCh38
rs1054030540 Chr.7: 130275308-130275308 on Build GRCh38
rs770891950 Chr.7: 130275226-130275226 on Build GRCh38
rs774371582 Chr.7: 130275352-130275352 on Build GRCh38
rs551448301 Chr.7: 130275298-130275298 on Build GRCh38
rs908663907 Chr.7: 130275151-130275151 on Build GRCh38
rs1033517561 Chr.7: 130275520-130275520 on Build GRCh38
rs767957872 Chr.7: 130275266-130275266 on Build GRCh38
rs936482848 Chr.7: 130275306-130275306 on Build GRCh38
rs190528888 Chr.7: 130275130-130275130 on Build GRCh38
rs931605907 Chr.7: 130275092-130275092 on Build GRCh38
rs560132249 Chr.7: 130275157-130275157 on Build GRCh38
rs975046743 Chr.7: 130275471-130275471 on Build GRCh38
rs527385842 Chr.7: 130275369-130275369 on Build GRCh38
rs892217327 Chr.7: 130275094-130275094 on Build GRCh38
rs754626497 Chr.7: 130275204-130275204 on Build GRCh38
rs970693864 Chr.7: 130275490-130275490 on Build GRCh38
rs746989318 Chr.7: 130275225-130275225 on Build GRCh38
rs563048357 Chr.7: 130275232-130275232 on Build GRCh38
rs771669165 Chr.7: 130275121-130275121 on Build GRCh38
rs562086993 Chr.7: 130275108-130275108 on Build GRCh38
rs746220232 Chr.7: 130275200-130275200 on Build GRCh38
rs376203585 Chr.7: 130275166-130275166 on Build GRCh38
rs747755858 Chr.7: 130275212-130275212 on Build GRCh38
rs747629893 Chr.7: 130275109-130275109 on Build GRCh38
rs867203654 Chr.7: 130275365-130275365 on Build GRCh38
rs766094216 Chr.7: 130275154-130275154 on Build GRCh38
rs35588699 Chr.7: 130275462-130275462 on Build GRCh38
rs145868138 Chr.7: 130275233-130275233 on Build GRCh38
rs768937185 Chr.7: 130275329-130275329 on Build GRCh38
rs765737679 Chr.7: 130275128-130275128 on Build GRCh38
rs757752564 Chr.7: 130275294-130275294 on Build GRCh38
rs776343338 Chr.7: 130275227-130275227 on Build GRCh38

View Full Product Details