Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 141743561-141744070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSBP1
Entrez Gene ID
6742
Gene Name
single stranded DNA binding protein 1
Gene Aliases
Mt-SSB, SOSS-B1, SSBP, mtSSB
UniGene
Hs.490394
Gene Chromosome Location
Chr.7: 141738309-141750488 on Build GRCh38
Transcripts
NM_001256510.1 NM_001256511.1 NM_001256512.1 NM_001256513.1 NM_003143.2 NR_046269.1 XM_005250048.4 XM_005250049.4 XM_005250050.4 XM_005250051.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6471703 c.130G>C p.V44L Chr.7: 141743605-141743605 on Build GRCh38
1086316 c.132C>T p.V44V Chr.7: 141743607-141743607 on Build GRCh38
1086317 c.271C>T p.R91W Chr.7: 141743946-141743946 on Build GRCh38
6471702 c.129T>A p.P43P Chr.7: 141743604-141743604 on Build GRCh38
137952 c.149G>A p.G50E Chr.7: 141743624-141743624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778082122 Chr.7: 141743854-141743854 on Build GRCh38
rs78598246 Chr.7: 141743699-141743699 on Build GRCh38
rs773115546 Chr.7: 141743912-141743912 on Build GRCh38
rs752190940 Chr.7: 141743963-141743963 on Build GRCh38
rs781336881 Chr.7: 141744007-141744007 on Build GRCh38
rs769933477 Chr.7: 141743879-141743879 on Build GRCh38
rs371743229 Chr.7: 141743851-141743851 on Build GRCh38
rs201482250 Chr.7: 141744016-141744016 on Build GRCh38
rs751523052 Chr.7: 141743688-141743688 on Build GRCh38
rs754582564 Chr.7: 141743743-141743743 on Build GRCh38
rs563970880 Chr.7: 141743832-141743832 on Build GRCh38
rs746126125 Chr.7: 141744008-141744008 on Build GRCh38
rs764823528 Chr.7: 141743961-141743961 on Build GRCh38
rs938506490 Chr.7: 141743987-141743987 on Build GRCh38
rs6957284 Chr.7: 141744038-141744038 on Build GRCh38
rs764849269 Chr.7: 141744040-141744040 on Build GRCh38
rs763440884 Chr.7: 141744033-141744033 on Build GRCh38
rs777188245 Chr.7: 141743624-141743625 on Build GRCh38
rs759724025 Chr.7: 141743678-141743678 on Build GRCh38
rs1055452900 Chr.7: 141743640-141743640 on Build GRCh38
rs201342338 Chr.7: 141743647-141743647 on Build GRCh38
rs181336944 Chr.7: 141743661-141743661 on Build GRCh38
rs575979891 Chr.7: 141743833-141743833 on Build GRCh38
rs750053920 Chr.7: 141743694-141743694 on Build GRCh38
rs367833670 Chr.7: 141743720-141743720 on Build GRCh38
rs757626917 Chr.7: 141743989-141743989 on Build GRCh38
rs772488076 Chr.7: 141743638-141743638 on Build GRCh38
rs3213666 Chr.7: 141743753-141743753 on Build GRCh38
rs747374622 Chr.7: 141743981-141743981 on Build GRCh38
rs765785387 Chr.7: 141743748-141743750 on Build GRCh38
rs202129795 Chr.7: 141743615-141743615 on Build GRCh38
rs767599376 Chr.7: 141743907-141743907 on Build GRCh38
rs913871378 Chr.7: 141743857-141743857 on Build GRCh38
rs528921793 Chr.7: 141743975-141743975 on Build GRCh38
rs760779735 Chr.7: 141743717-141743717 on Build GRCh38
rs760601343 Chr.7: 141743946-141743946 on Build GRCh38
rs759890740 Chr.7: 141743717-141743718 on Build GRCh38
rs766288953 Chr.7: 141744036-141744036 on Build GRCh38
rs868577257 Chr.7: 141743643-141743643 on Build GRCh38
rs748455345 Chr.7: 141743619-141743619 on Build GRCh38
rs765460267 Chr.7: 141744041-141744041 on Build GRCh38
rs148271472 Chr.7: 141743964-141743964 on Build GRCh38
rs186344144 Chr.7: 141743863-141743863 on Build GRCh38
rs745872096 Chr.7: 141743872-141743872 on Build GRCh38
rs796291801 Chr.7: 141744000-141744000 on Build GRCh38
rs376756147 Chr.7: 141743859-141743859 on Build GRCh38
rs773349100 Chr.7: 141744028-141744028 on Build GRCh38
rs773376873 Chr.7: 141743639-141743639 on Build GRCh38
rs989445821 Chr.7: 141744019-141744019 on Build GRCh38
rs200664859 Chr.7: 141743690-141743690 on Build GRCh38
rs1046199739 Chr.7: 141743746-141743746 on Build GRCh38
rs763937894 Chr.7: 141743684-141743684 on Build GRCh38
rs761839550 Chr.7: 141743884-141743884 on Build GRCh38
rs754387183 Chr.7: 141743952-141743952 on Build GRCh38
rs372289521 Chr.7: 141743873-141743873 on Build GRCh38
rs771128365 Chr.7: 141743861-141743861 on Build GRCh38
rs367792667 Chr.7: 141743747-141743747 on Build GRCh38
rs779532342 Chr.7: 141743617-141743617 on Build GRCh38
rs199799284 Chr.7: 141743744-141743744 on Build GRCh38
rs954855191 Chr.7: 141743763-141743763 on Build GRCh38
rs1055275773 Chr.7: 141743794-141743794 on Build GRCh38
rs774109769 Chr.7: 141743883-141743883 on Build GRCh38
rs373522073 Chr.7: 141744021-141744021 on Build GRCh38
rs769719345 Chr.7: 141744015-141744015 on Build GRCh38
rs757583499 Chr.7: 141743650-141743650 on Build GRCh38
rs761460057 Chr.7: 141743689-141743689 on Build GRCh38
rs776096235 Chr.7: 141744037-141744037 on Build GRCh38
rs760478932 Chr.7: 141744030-141744030 on Build GRCh38
rs553972941 Chr.7: 141743736-141743736 on Build GRCh38
rs921291875 Chr.7: 141743634-141743634 on Build GRCh38
rs965913310 Chr.7: 141743905-141743905 on Build GRCh38
rs771183484 Chr.7: 141743671-141743671 on Build GRCh38
rs776670908 Chr.7: 141743674-141743674 on Build GRCh38
rs11554109 Chr.7: 141743623-141743623 on Build GRCh38
rs775666793 Chr.7: 141743897-141743897 on Build GRCh38
rs1803993 Chr.7: 141743951-141743951 on Build GRCh38

View Full Product Details