Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143287888-143288385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP2
Entrez Gene ID
835
Gene Name
caspase 2
Gene Aliases
CASP-2, ICH1, NEDD-2, NEDD2, PPP1R57
UniGene
Hs.368982
Gene Chromosome Location
Chr.7: 143288215-143307696 on Build GRCh38
Transcripts
NM_032982.3 NM_032983.3 NM_001242773.2 NM_001242774.2 NM_001242775.2 NM_001282876.1 NM_001282877.1 NM_153345.3 NR_040003.2 NR_104250.1 NR_104251.1 NR_104252.1 NR_104253.1 NR_104254.1

Gene Symbol
TMEM139
Entrez Gene ID
135932
Gene Name
transmembrane protein 139
Gene Aliases
-
UniGene
Hs.17558
Gene Chromosome Location
Chr.7: 143284899-143288049 on Build GRCh38
Transcripts
NM_032982.3 NM_032983.3 NM_001242773.2 NM_001242774.2 NM_001242775.2 NM_001282876.1 NM_001282877.1 NM_153345.3 NR_040003.2 NR_104250.1 NR_104251.1 NR_104252.1 NR_104253.1 NR_104254.1

SNP Detail

SNP ID Chromosome Location
rs570190883 Chr.7: 143288319-143288319 on Build GRCh38
rs950999059 Chr.7: 143287928-143287928 on Build GRCh38
rs926847147 Chr.7: 143288149-143288149 on Build GRCh38
rs563498385 Chr.7: 143288161-143288161 on Build GRCh38
rs1007991981 Chr.7: 143288214-143288214 on Build GRCh38
rs915343041 Chr.7: 143288000-143288000 on Build GRCh38
rs988164315 Chr.7: 143287948-143287948 on Build GRCh38
rs988301909 Chr.7: 143288251-143288251 on Build GRCh38
rs1056811822 Chr.7: 143288081-143288081 on Build GRCh38
rs186744526 Chr.7: 143288226-143288226 on Build GRCh38
rs577651778 Chr.7: 143288111-143288111 on Build GRCh38
rs1008322080 Chr.7: 143288317-143288317 on Build GRCh38
rs958017944 Chr.7: 143288074-143288074 on Build GRCh38
rs1037698259 Chr.7: 143288164-143288164 on Build GRCh38
rs899287752 Chr.7: 143288181-143288181 on Build GRCh38
rs918613551 Chr.7: 143288245-143288245 on Build GRCh38
rs1048294629 Chr.7: 143288341-143288341 on Build GRCh38
rs905210159 Chr.7: 143287954-143287954 on Build GRCh38
rs544919393 Chr.7: 143288154-143288154 on Build GRCh38
rs11574688 Chr.7: 143288024-143288024 on Build GRCh38
rs920600769 Chr.7: 143288326-143288326 on Build GRCh38
rs898373397 Chr.7: 143288269-143288269 on Build GRCh38
rs559926330 Chr.7: 143288293-143288293 on Build GRCh38
rs866305289 Chr.7: 143288083-143288083 on Build GRCh38
rs925062574 Chr.7: 143288071-143288071 on Build GRCh38
rs573268017 Chr.7: 143288080-143288080 on Build GRCh38
rs556226916 Chr.7: 143288229-143288229 on Build GRCh38
rs554950664 Chr.7: 143288054-143288054 on Build GRCh38
rs964930550 Chr.7: 143288167-143288167 on Build GRCh38
rs899234181 Chr.7: 143287982-143287982 on Build GRCh38
rs562085140 Chr.7: 143288227-143288227 on Build GRCh38
rs10241727 Chr.7: 143288056-143288056 on Build GRCh38
rs1019513327 Chr.7: 143288217-143288217 on Build GRCh38
rs898257235 Chr.7: 143288093-143288093 on Build GRCh38
rs1037919764 Chr.7: 143288322-143288322 on Build GRCh38
rs968234624 Chr.7: 143288348-143288348 on Build GRCh38
rs774189223 Chr.7: 143287934-143287934 on Build GRCh38
rs866945534 Chr.7: 143288097-143288097 on Build GRCh38
rs147527355 Chr.7: 143288090-143288090 on Build GRCh38
rs963699325 Chr.7: 143288052-143288052 on Build GRCh38
rs895043617 Chr.7: 143288082-143288082 on Build GRCh38
rs111437190 Chr.7: 143288040-143288040 on Build GRCh38
rs575454509 Chr.7: 143288184-143288184 on Build GRCh38
rs867488037 Chr.7: 143287986-143287986 on Build GRCh38
rs1016382392 Chr.7: 143287978-143287978 on Build GRCh38
rs1047774456 Chr.7: 143288202-143288202 on Build GRCh38
rs1022433161 Chr.7: 143288183-143288183 on Build GRCh38
rs1025897756 Chr.7: 143287993-143287993 on Build GRCh38
rs866473057 Chr.7: 143288231-143288231 on Build GRCh38
rs144511554 Chr.7: 143288362-143288362 on Build GRCh38
rs537702608 Chr.7: 143288029-143288029 on Build GRCh38
rs113268906 Chr.7: 143288340-143288340 on Build GRCh38
rs184689717 Chr.7: 143288103-143288103 on Build GRCh38
rs756924105 Chr.7: 143287980-143287982 on Build GRCh38
rs1008044191 Chr.7: 143288358-143288358 on Build GRCh38
rs541418331 Chr.7: 143288240-143288240 on Build GRCh38
rs1031520268 Chr.7: 143288354-143288354 on Build GRCh38
rs1042418001 Chr.7: 143288041-143288041 on Build GRCh38
rs886953940 Chr.7: 143288213-143288213 on Build GRCh38
rs890815884 Chr.7: 143288125-143288125 on Build GRCh38
rs975030158 Chr.7: 143288070-143288070 on Build GRCh38
rs149238655 Chr.7: 143288192-143288192 on Build GRCh38
rs1019461182 Chr.7: 143288047-143288047 on Build GRCh38
rs996473133 Chr.7: 143288335-143288336 on Build GRCh38
rs978263399 Chr.7: 143288350-143288350 on Build GRCh38
rs949543846 Chr.7: 143288104-143288104 on Build GRCh38
rs993838560 Chr.7: 143288096-143288096 on Build GRCh38
rs912134885 Chr.7: 143287966-143287966 on Build GRCh38
rs982264733 Chr.7: 143288298-143288298 on Build GRCh38
rs1046702897 Chr.7: 143288118-143288118 on Build GRCh38
rs1031964569 Chr.7: 143288237-143288237 on Build GRCh38
rs929295408 Chr.7: 143288330-143288330 on Build GRCh38
rs551684797 Chr.7: 143288316-143288316 on Build GRCh38
rs533462215 Chr.7: 143288296-143288296 on Build GRCh38
rs527401653 Chr.7: 143287970-143287970 on Build GRCh38
rs890527187 Chr.7: 143288342-143288342 on Build GRCh38
rs1038137921 Chr.7: 143288092-143288092 on Build GRCh38

View Full Product Details