Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143300115-143300609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP2
Entrez Gene ID
835
Gene Name
caspase 2
Gene Aliases
CASP-2, ICH1, NEDD-2, NEDD2, PPP1R57
UniGene
Hs.368982
Gene Chromosome Location
Chr.7: 143288215-143307696 on Build GRCh38
Transcripts
NM_001224.4 NM_032982.3 NM_032983.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6854882 c.924A>G p.L308L Chr.7: 143300251-143300251 on Build GRCh38
6620573 c.907G>A p.A303T Chr.7: 143300234-143300234 on Build GRCh38
6252623 c.877-1G>T p.? Chr.7: 143300203-143300203 on Build GRCh38
5712435 c.916C>T p.P306S Chr.7: 143300243-143300243 on Build GRCh38
1699507 c.917C>T p.P306L Chr.7: 143300244-143300244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368449253 Chr.7: 143300289-143300289 on Build GRCh38
rs752018469 Chr.7: 143300151-143300151 on Build GRCh38
rs564574124 Chr.7: 143300535-143300535 on Build GRCh38
rs541450291 Chr.7: 143300550-143300550 on Build GRCh38
rs200353443 Chr.7: 143300238-143300238 on Build GRCh38
rs4647319 Chr.7: 143300459-143300459 on Build GRCh38
rs769361986 Chr.7: 143300309-143300309 on Build GRCh38
rs931205807 Chr.7: 143300548-143300548 on Build GRCh38
rs141723844 Chr.7: 143300512-143300512 on Build GRCh38
rs4647321 Chr.7: 143300533-143300533 on Build GRCh38
rs377218240 Chr.7: 143300262-143300262 on Build GRCh38
rs756201685 Chr.7: 143300234-143300234 on Build GRCh38
rs763709140 Chr.7: 143300385-143300385 on Build GRCh38
rs777570112 Chr.7: 143300287-143300287 on Build GRCh38
rs760739852 Chr.7: 143300375-143300375 on Build GRCh38
rs762930613 Chr.7: 143300392-143300392 on Build GRCh38
rs745356050 Chr.7: 143300186-143300186 on Build GRCh38
rs776129080 Chr.7: 143300200-143300200 on Build GRCh38
rs769262939 Chr.7: 143300329-143300329 on Build GRCh38
rs766474545 Chr.7: 143300382-143300382 on Build GRCh38
rs757957052 Chr.7: 143300342-143300342 on Build GRCh38
rs770571274 Chr.7: 143300174-143300174 on Build GRCh38
rs4647320 Chr.7: 143300501-143300501 on Build GRCh38
rs899980722 Chr.7: 143300352-143300352 on Build GRCh38
rs766307129 Chr.7: 143300237-143300237 on Build GRCh38
rs973362293 Chr.7: 143300567-143300567 on Build GRCh38
rs943789258 Chr.7: 143300573-143300573 on Build GRCh38
rs372813468 Chr.7: 143300319-143300319 on Build GRCh38
rs748541175 Chr.7: 143300424-143300424 on Build GRCh38
rs372542250 Chr.7: 143300199-143300199 on Build GRCh38
rs748774523 Chr.7: 143300415-143300415 on Build GRCh38
rs559161604 Chr.7: 143300426-143300426 on Build GRCh38
rs761799919 Chr.7: 143300224-143300225 on Build GRCh38
rs774896123 Chr.7: 143300344-143300344 on Build GRCh38
rs374230679 Chr.7: 143300239-143300239 on Build GRCh38
rs886746756 Chr.7: 143300565-143300565 on Build GRCh38
rs886860384 Chr.7: 143300246-143300246 on Build GRCh38
rs763497851 Chr.7: 143300206-143300206 on Build GRCh38
rs767378860 Chr.7: 143300487-143300487 on Build GRCh38
rs776734557 Chr.7: 143300457-143300457 on Build GRCh38
rs760971994 Chr.7: 143300453-143300453 on Build GRCh38
rs751336595 Chr.7: 143300288-143300288 on Build GRCh38
rs745428846 Chr.7: 143300304-143300304 on Build GRCh38
rs1040799678 Chr.7: 143300583-143300583 on Build GRCh38
rs767747070 Chr.7: 143300230-143300230 on Build GRCh38
rs746698551 Chr.7: 143300173-143300173 on Build GRCh38
rs762173952 Chr.7: 143300223-143300223 on Build GRCh38
rs773493211 Chr.7: 143300372-143300372 on Build GRCh38
rs991399152 Chr.7: 143300513-143300513 on Build GRCh38
rs779548643 Chr.7: 143300416-143300416 on Build GRCh38
rs772236672 Chr.7: 143300358-143300358 on Build GRCh38
rs779757480 Chr.7: 143300414-143300414 on Build GRCh38
rs999504066 Chr.7: 143300485-143300485 on Build GRCh38
rs754519632 Chr.7: 143300251-143300251 on Build GRCh38
rs751589935 Chr.7: 143300219-143300220 on Build GRCh38
rs765386737 Chr.7: 143300169-143300178 on Build GRCh38
rs761590773 Chr.7: 143300466-143300466 on Build GRCh38
rs573888034 Chr.7: 143300400-143300400 on Build GRCh38
rs893493434 Chr.7: 143300456-143300456 on Build GRCh38
rs774806131 Chr.7: 143300213-143300213 on Build GRCh38
rs771122177 Chr.7: 143300455-143300455 on Build GRCh38
rs755800033 Chr.7: 143300578-143300578 on Build GRCh38
rs780792819 Chr.7: 143300409-143300409 on Build GRCh38
rs780893395 Chr.7: 143300295-143300295 on Build GRCh38
rs991606308 Chr.7: 143300521-143300521 on Build GRCh38
rs997127318 Chr.7: 143300404-143300404 on Build GRCh38
rs779646871 Chr.7: 143300312-143300312 on Build GRCh38
rs764560560 Chr.7: 143300212-143300212 on Build GRCh38
rs767441689 Chr.7: 143300263-143300263 on Build GRCh38
rs555135029 Chr.7: 143300338-143300338 on Build GRCh38
rs773406561 Chr.7: 143300436-143300436 on Build GRCh38
rs751529323 Chr.7: 143300465-143300465 on Build GRCh38
rs550436353 Chr.7: 143300553-143300553 on Build GRCh38
rs776057327 Chr.7: 143300177-143300177 on Build GRCh38
rs568058185 Chr.7: 143300405-143300405 on Build GRCh38
rs559358465 Chr.7: 143300508-143300508 on Build GRCh38
rs750534880 Chr.7: 143300233-143300233 on Build GRCh38
rs373585704 Chr.7: 143300461-143300461 on Build GRCh38
rs751426739 Chr.7: 143300403-143300403 on Build GRCh38
rs368359334 Chr.7: 143300160-143300160 on Build GRCh38
rs757061833 Chr.7: 143300407-143300407 on Build GRCh38
rs982136836 Chr.7: 143300484-143300484 on Build GRCh38
rs928494001 Chr.7: 143300505-143300505 on Build GRCh38
rs777476131 Chr.7: 143300161-143300161 on Build GRCh38
rs750095620 Chr.7: 143300411-143300411 on Build GRCh38

View Full Product Details