Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143388409-143388915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZYX
Entrez Gene ID
7791
Gene Name
zyxin
Gene Aliases
ESP-2, HED-2
UniGene
Hs.490415
Gene Chromosome Location
Chr.7: 143381174-143391111 on Build GRCh38
Transcripts
NM_001010972.1 NM_003461.4 XM_011516569.2 XM_017012587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6834681 c.1178G>A p.R393Q Chr.7: 143388522-143388522 on Build GRCh38
1187457 c.1262G>T p.G421V Chr.7: 143388606-143388606 on Build GRCh38
5872116 c.1384G>A p.A462T Chr.7: 143388836-143388836 on Build GRCh38
5872115 c.1216C>T p.H406Y Chr.7: 143388560-143388560 on Build GRCh38
5872114 c.1157G>A p.R386Q Chr.7: 143388501-143388501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146983863 Chr.7: 143388871-143388871 on Build GRCh38
rs368356918 Chr.7: 143388704-143388704 on Build GRCh38
rs778220389 Chr.7: 143388720-143388720 on Build GRCh38
rs146749977 Chr.7: 143388858-143388858 on Build GRCh38
rs570358698 Chr.7: 143388666-143388675 on Build GRCh38
rs777931036 Chr.7: 143388781-143388781 on Build GRCh38
rs758472576 Chr.7: 143388847-143388847 on Build GRCh38
rs771255181 Chr.7: 143388793-143388793 on Build GRCh38
rs773934127 Chr.7: 143388816-143388816 on Build GRCh38
rs113113492 Chr.7: 143388660-143388660 on Build GRCh38
rs76291337 Chr.7: 143388544-143388544 on Build GRCh38
rs559675418 Chr.7: 143388634-143388634 on Build GRCh38
rs142129852 Chr.7: 143388436-143388436 on Build GRCh38
rs781027905 Chr.7: 143388701-143388701 on Build GRCh38
rs929104308 Chr.7: 143388815-143388815 on Build GRCh38
rs770296649 Chr.7: 143388632-143388632 on Build GRCh38
rs758330921 Chr.7: 143388559-143388559 on Build GRCh38
rs769383580 Chr.7: 143388808-143388808 on Build GRCh38
rs753911301 Chr.7: 143388667-143388667 on Build GRCh38
rs748499610 Chr.7: 143388591-143388591 on Build GRCh38
rs1047269639 Chr.7: 143388758-143388758 on Build GRCh38
rs769718724 Chr.7: 143388522-143388522 on Build GRCh38
rs769975439 Chr.7: 143388876-143388876 on Build GRCh38
rs758657866 Chr.7: 143388776-143388776 on Build GRCh38
rs544381844 Chr.7: 143388812-143388812 on Build GRCh38
rs768352370 Chr.7: 143388813-143388813 on Build GRCh38
rs112926466 Chr.7: 143388765-143388765 on Build GRCh38
rs199617231 Chr.7: 143388756-143388756 on Build GRCh38
rs754897443 Chr.7: 143388515-143388515 on Build GRCh38
rs767477318 Chr.7: 143388752-143388752 on Build GRCh38
rs913062234 Chr.7: 143388500-143388500 on Build GRCh38
rs755109421 Chr.7: 143388685-143388685 on Build GRCh38
rs151166003 Chr.7: 143388440-143388440 on Build GRCh38
rs755019600 Chr.7: 143388759-143388759 on Build GRCh38
rs777554393 Chr.7: 143388565-143388565 on Build GRCh38
rs762242416 Chr.7: 143388539-143388539 on Build GRCh38
rs41277428 Chr.7: 143388729-143388729 on Build GRCh38
rs181913993 Chr.7: 143388637-143388637 on Build GRCh38
rs571369385 Chr.7: 143388828-143388828 on Build GRCh38
rs774624715 Chr.7: 143388577-143388577 on Build GRCh38
rs748820036 Chr.7: 143388895-143388895 on Build GRCh38
rs762721087 Chr.7: 143388740-143388740 on Build GRCh38
rs879834568 Chr.7: 143388523-143388523 on Build GRCh38
rs6954724 Chr.7: 143388726-143388726 on Build GRCh38
rs749155185 Chr.7: 143388455-143388458 on Build GRCh38
rs142288548 Chr.7: 143388892-143388892 on Build GRCh38
rs374951395 Chr.7: 143388643-143388643 on Build GRCh38
rs779247184 Chr.7: 143388497-143388497 on Build GRCh38
rs775156061 Chr.7: 143388810-143388810 on Build GRCh38
rs768343435 Chr.7: 143388535-143388535 on Build GRCh38
rs756577754 Chr.7: 143388454-143388454 on Build GRCh38
rs749076750 Chr.7: 143388530-143388530 on Build GRCh38
rs960012948 Chr.7: 143388526-143388526 on Build GRCh38
rs773977946 Chr.7: 143388536-143388536 on Build GRCh38
rs762814445 Chr.7: 143388644-143388644 on Build GRCh38
rs371072304 Chr.7: 143388541-143388541 on Build GRCh38
rs146818692 Chr.7: 143388602-143388602 on Build GRCh38
rs372754746 Chr.7: 143388732-143388732 on Build GRCh38
rs370303425 Chr.7: 143388512-143388512 on Build GRCh38
rs576382219 Chr.7: 143388540-143388540 on Build GRCh38
rs761582543 Chr.7: 143388749-143388749 on Build GRCh38
rs963368750 Chr.7: 143388768-143388768 on Build GRCh38
rs756110512 Chr.7: 143388572-143388572 on Build GRCh38
rs778994196 Chr.7: 143388772-143388772 on Build GRCh38
rs112389985 Chr.7: 143388661-143388661 on Build GRCh38
rs372852797 Chr.7: 143388790-143388790 on Build GRCh38
rs751775395 Chr.7: 143388861-143388861 on Build GRCh38
rs377556976 Chr.7: 143388496-143388496 on Build GRCh38
rs1012203062 Chr.7: 143388665-143388665 on Build GRCh38
rs768872930 Chr.7: 143388576-143388576 on Build GRCh38
rs761647537 Chr.7: 143388817-143388817 on Build GRCh38
rs752776674 Chr.7: 143388846-143388846 on Build GRCh38
rs200252570 Chr.7: 143388731-143388731 on Build GRCh38
rs373373502 Chr.7: 143388459-143388459 on Build GRCh38
rs751407162 Chr.7: 143388648-143388648 on Build GRCh38
rs781295880 Chr.7: 143388872-143388872 on Build GRCh38
rs41277425 Chr.7: 143388875-143388875 on Build GRCh38
rs71528913 Chr.7: 143388545-143388545 on Build GRCh38
rs201007727 Chr.7: 143388636-143388636 on Build GRCh38
rs772851326 Chr.7: 143388785-143388785 on Build GRCh38
rs781348336 Chr.7: 143388521-143388521 on Build GRCh38
rs369557649 Chr.7: 143388859-143388859 on Build GRCh38
rs765063788 Chr.7: 143388547-143388547 on Build GRCh38
rs113729249 Chr.7: 143388487-143388487 on Build GRCh38
rs964494631 Chr.7: 143388614-143388614 on Build GRCh38
rs145663530 Chr.7: 143388844-143388844 on Build GRCh38
rs140343735 Chr.7: 143388738-143388738 on Build GRCh38
rs112041427 Chr.7: 143388766-143388766 on Build GRCh38
rs111867630 Chr.7: 143388513-143388513 on Build GRCh38
rs747414710 Chr.7: 143388722-143388722 on Build GRCh38
rs541672435 Chr.7: 143388592-143388592 on Build GRCh38
rs775175829 Chr.7: 143388528-143388528 on Build GRCh38
rs778518202 Chr.7: 143388799-143388799 on Build GRCh38
rs373656218 Chr.7: 143388556-143388556 on Build GRCh38
rs1035787606 Chr.7: 143388893-143388893 on Build GRCh38
rs754182486 Chr.7: 143388468-143388468 on Build GRCh38
rs772384803 Chr.7: 143388716-143388716 on Build GRCh38
rs945826708 Chr.7: 143388724-143388724 on Build GRCh38
rs367718052 Chr.7: 143388566-143388566 on Build GRCh38
rs201468462 Chr.7: 143388744-143388744 on Build GRCh38
rs749414962 Chr.7: 143388575-143388575 on Build GRCh38
rs770852133 Chr.7: 143388665-143388666 on Build GRCh38
rs373566522 Chr.7: 143388689-143388689 on Build GRCh38
rs376217001 Chr.7: 143388794-143388794 on Build GRCh38
rs748409519 Chr.7: 143388703-143388703 on Build GRCh38
rs771791771 Chr.7: 143388667-143388668 on Build GRCh38
rs752866447 Chr.7: 143388773-143388773 on Build GRCh38
rs199792709 Chr.7: 143388741-143388741 on Build GRCh38
rs75965463 Chr.7: 143388829-143388829 on Build GRCh38
rs113589902 Chr.7: 143388700-143388700 on Build GRCh38
rs534030625 Chr.7: 143388711-143388711 on Build GRCh38
rs779934079 Chr.7: 143388666-143388666 on Build GRCh38
rs757235429 Chr.7: 143388656-143388656 on Build GRCh38
rs975833871 Chr.7: 143388437-143388437 on Build GRCh38

View Full Product Details