Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42710104-42710603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB3
Entrez Gene ID
1142
Gene Name
cholinergic receptor nicotinic beta 3 subunit
Gene Aliases
-
UniGene
Hs.654576
Gene Chromosome Location
Chr.8: 42697419-42737407 on Build GRCh38
Transcripts
NM_000749.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7080584 c.241C>T p.L81F Chr.8: 42710426-42710426 on Build GRCh38
3900305 c.242T>C p.L81P Chr.8: 42710427-42710427 on Build GRCh38
4449330 c.205-2A>C p.? Chr.8: 42710388-42710388 on Build GRCh38
3996069 c.225G>A p.M75I Chr.8: 42710410-42710410 on Build GRCh38
3699096 c.225G>T p.M75I Chr.8: 42710410-42710410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55786845 Chr.8: 42710371-42710371 on Build GRCh38
rs903268787 Chr.8: 42710248-42710248 on Build GRCh38
rs910128248 Chr.8: 42710216-42710216 on Build GRCh38
rs975777302 Chr.8: 42710155-42710155 on Build GRCh38
rs903259065 Chr.8: 42710561-42710561 on Build GRCh38
rs879059102 Chr.8: 42710301-42710301 on Build GRCh38
rs1045823161 Chr.8: 42710243-42710243 on Build GRCh38
rs1055948117 Chr.8: 42710231-42710231 on Build GRCh38
rs1017537496 Chr.8: 42710362-42710362 on Build GRCh38
rs558636081 Chr.8: 42710145-42710146 on Build GRCh38
rs188806970 Chr.8: 42710437-42710437 on Build GRCh38
rs1007920444 Chr.8: 42710315-42710315 on Build GRCh38
rs893036434 Chr.8: 42710314-42710314 on Build GRCh38
rs541879490 Chr.8: 42710156-42710156 on Build GRCh38
rs907490746 Chr.8: 42710349-42710349 on Build GRCh38
rs919026179 Chr.8: 42710168-42710168 on Build GRCh38
rs10108555 Chr.8: 42710221-42710221 on Build GRCh38
rs1025986532 Chr.8: 42710458-42710458 on Build GRCh38
rs894664471 Chr.8: 42710233-42710233 on Build GRCh38
rs759488366 Chr.8: 42710410-42710410 on Build GRCh38
rs76654847 Chr.8: 42710580-42710580 on Build GRCh38
rs79465339 Chr.8: 42710351-42710351 on Build GRCh38
rs916182165 Chr.8: 42710513-42710513 on Build GRCh38
rs950155076 Chr.8: 42710529-42710529 on Build GRCh38
rs745905290 Chr.8: 42710421-42710421 on Build GRCh38
rs1054449560 Chr.8: 42710582-42710582 on Build GRCh38
rs1033468302 Chr.8: 42710290-42710290 on Build GRCh38
rs919005934 Chr.8: 42710190-42710190 on Build GRCh38
rs998821269 Chr.8: 42710264-42710264 on Build GRCh38
rs770752100 Chr.8: 42710218-42710218 on Build GRCh38
rs776514234 Chr.8: 42710468-42710468 on Build GRCh38
rs756503769 Chr.8: 42710341-42710341 on Build GRCh38
rs74730626 Chr.8: 42710232-42710232 on Build GRCh38
rs749986594 Chr.8: 42710374-42710375 on Build GRCh38
rs373220649 Chr.8: 42710395-42710395 on Build GRCh38
rs78172416 Chr.8: 42710500-42710500 on Build GRCh38
rs535435269 Chr.8: 42710189-42710189 on Build GRCh38
rs1048811421 Chr.8: 42710205-42710205 on Build GRCh38
rs184205173 Chr.8: 42710157-42710157 on Build GRCh38
rs950505162 Chr.8: 42710473-42710473 on Build GRCh38
rs769555013 Chr.8: 42710479-42710479 on Build GRCh38
rs533641355 Chr.8: 42710240-42710240 on Build GRCh38
rs369617386 Chr.8: 42710367-42710367 on Build GRCh38
rs575707254 Chr.8: 42710366-42710366 on Build GRCh38
rs75496257 Chr.8: 42710293-42710293 on Build GRCh38
rs767400288 Chr.8: 42710480-42710480 on Build GRCh38
rs762510558 Chr.8: 42710482-42710482 on Build GRCh38
rs138880882 Chr.8: 42710495-42710495 on Build GRCh38
rs762090517 Chr.8: 42710202-42710202 on Build GRCh38
rs747959801 Chr.8: 42710469-42710469 on Build GRCh38
rs573002998 Chr.8: 42710134-42710134 on Build GRCh38
rs142960043 Chr.8: 42710199-42710199 on Build GRCh38
rs1012033943 Chr.8: 42710380-42710380 on Build GRCh38
rs992122644 Chr.8: 42710502-42710502 on Build GRCh38
rs80221890 Chr.8: 42710398-42710398 on Build GRCh38
rs549141799 Chr.8: 42710560-42710560 on Build GRCh38
rs1052485804 Chr.8: 42710256-42710256 on Build GRCh38

View Full Product Details