Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 9136918-9137416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R3B
Entrez Gene ID
79660
Gene Name
protein phosphatase 1 regulatory subunit 3B
Gene Aliases
GL, PPP1R4, PTG
UniGene
Hs.458513
Gene Chromosome Location
Chr.8: 9136254-9151642 on Build GRCh38
Transcripts
NM_001201329.1 NM_024607.3 XM_006716253.3 XM_017013835.1

SNP Detail

SNP ID Chromosome Location
rs73197767 Chr.8: 9137001-9137001 on Build GRCh38
rs950881196 Chr.8: 9137251-9137251 on Build GRCh38
rs796601291 Chr.8: 9136946-9136946 on Build GRCh38
rs537432545 Chr.8: 9137249-9137249 on Build GRCh38
rs987886572 Chr.8: 9137165-9137165 on Build GRCh38
rs539288021 Chr.8: 9137215-9137215 on Build GRCh38
rs879001222 Chr.8: 9136992-9136992 on Build GRCh38
rs953653059 Chr.8: 9137033-9137033 on Build GRCh38
rs964848188 Chr.8: 9137356-9137356 on Build GRCh38
rs919463755 Chr.8: 9136982-9136982 on Build GRCh38
rs754918467 Chr.8: 9137185-9137185 on Build GRCh38
rs1014891366 Chr.8: 9136981-9136981 on Build GRCh38
rs933279784 Chr.8: 9137076-9137081 on Build GRCh38
rs963323969 Chr.8: 9136993-9136993 on Build GRCh38
rs765944901 Chr.8: 9137036-9137036 on Build GRCh38
rs975146612 Chr.8: 9137357-9137357 on Build GRCh38
rs953786967 Chr.8: 9137385-9137385 on Build GRCh38
rs143629894 Chr.8: 9136948-9136948 on Build GRCh38
rs895910056 Chr.8: 9136974-9136974 on Build GRCh38
rs899178745 Chr.8: 9137349-9137349 on Build GRCh38
rs987712484 Chr.8: 9137386-9137386 on Build GRCh38
rs752482222 Chr.8: 9137394-9137394 on Build GRCh38
rs558965252 Chr.8: 9137227-9137227 on Build GRCh38
rs933206369 Chr.8: 9137371-9137371 on Build GRCh38
rs1044743177 Chr.8: 9137231-9137231 on Build GRCh38
rs909193858 Chr.8: 9137028-9137028 on Build GRCh38
rs533207186 Chr.8: 9137007-9137007 on Build GRCh38
rs891639978 Chr.8: 9137107-9137107 on Build GRCh38
rs935091360 Chr.8: 9137164-9137164 on Build GRCh38
rs1039065514 Chr.8: 9137060-9137060 on Build GRCh38
rs551442417 Chr.8: 9137158-9137158 on Build GRCh38
rs1056727306 Chr.8: 9137256-9137256 on Build GRCh38
rs536895705 Chr.8: 9137130-9137130 on Build GRCh38
rs946332156 Chr.8: 9137209-9137209 on Build GRCh38
rs1034856146 Chr.8: 9136971-9136971 on Build GRCh38
rs574149798 Chr.8: 9137367-9137367 on Build GRCh38
rs1014922378 Chr.8: 9137348-9137348 on Build GRCh38
rs886917394 Chr.8: 9137258-9137258 on Build GRCh38
rs151012124 Chr.8: 9137169-9137169 on Build GRCh38
rs559229486 Chr.8: 9137182-9137185 on Build GRCh38
rs905498678 Chr.8: 9137147-9137147 on Build GRCh38
rs1034359856 Chr.8: 9136961-9136963 on Build GRCh38
rs373344513 Chr.8: 9137218-9137218 on Build GRCh38
rs530792634 Chr.8: 9136954-9136954 on Build GRCh38
rs994837902 Chr.8: 9137353-9137353 on Build GRCh38
rs1028933509 Chr.8: 9137364-9137364 on Build GRCh38
rs554290247 Chr.8: 9137298-9137298 on Build GRCh38
rs9949 Chr.8: 9137002-9137002 on Build GRCh38
rs923172551 Chr.8: 9137071-9137071 on Build GRCh38
rs1052876575 Chr.8: 9137099-9137099 on Build GRCh38
rs111753738 Chr.8: 9137232-9137232 on Build GRCh38
rs1043216282 Chr.8: 9137136-9137136 on Build GRCh38
rs1003819652 Chr.8: 9136961-9136961 on Build GRCh38
rs1004956296 Chr.8: 9136978-9136978 on Build GRCh38

View Full Product Details