Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 54622025-54622537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RP1
Entrez Gene ID
6101
Gene Name
retinitis pigmentosa 1 (autosomal dominant)
Gene Aliases
DCDC4A, ORP1
UniGene
Hs.732820
Gene Chromosome Location
Chr.8: 54509448-54630834 on Build GRCh38
Transcripts
NM_006269.1 XM_017013721.1 XM_017013722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1100537 c.690A>G p.G230G Chr.8: 54622191-54622191 on Build GRCh38
1700093 c.148C>T p.P50S Chr.8: 54622219-54622219 on Build GRCh38
3649766 c.760G>A p.G254R Chr.8: 54622261-54622261 on Build GRCh38
225260 c.761G>A p.G254E Chr.8: 54622262-54622262 on Build GRCh38
3663801 c.702C>A p.I234I Chr.8: 54622203-54622203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032798791 Chr.8: 54622477-54622477 on Build GRCh38
rs761808094 Chr.8: 54622328-54622328 on Build GRCh38
rs914790120 Chr.8: 54622475-54622475 on Build GRCh38
rs373574136 Chr.8: 54622247-54622247 on Build GRCh38
rs781445794 Chr.8: 54622220-54622220 on Build GRCh38
rs201892201 Chr.8: 54622084-54622084 on Build GRCh38
rs146337514 Chr.8: 54622119-54622119 on Build GRCh38
rs967270448 Chr.8: 54622454-54622454 on Build GRCh38
rs756392635 Chr.8: 54622328-54622329 on Build GRCh38
rs190484195 Chr.8: 54622199-54622199 on Build GRCh38
rs186571865 Chr.8: 54622111-54622111 on Build GRCh38
rs207469187 Chr.8: 54622065-54622065 on Build GRCh38
rs769156393 Chr.8: 54622243-54622243 on Build GRCh38
rs550073225 Chr.8: 54622237-54622237 on Build GRCh38
rs145691085 Chr.8: 54622153-54622153 on Build GRCh38
rs758604193 Chr.8: 54622176-54622176 on Build GRCh38
rs534993935 Chr.8: 54622414-54622414 on Build GRCh38
rs139570650 Chr.8: 54622370-54622370 on Build GRCh38
rs892801523 Chr.8: 54622062-54622062 on Build GRCh38
rs780362977 Chr.8: 54622089-54622089 on Build GRCh38
rs863223340 Chr.8: 54622180-54622180 on Build GRCh38
rs780569571 Chr.8: 54622332-54622333 on Build GRCh38
rs990933958 Chr.8: 54622489-54622489 on Build GRCh38
rs1001046875 Chr.8: 54622467-54622467 on Build GRCh38
rs577635143 Chr.8: 54622340-54622340 on Build GRCh38
rs747236589 Chr.8: 54622102-54622102 on Build GRCh38
rs770516808 Chr.8: 54622136-54622136 on Build GRCh38
rs990840478 Chr.8: 54622134-54622134 on Build GRCh38
rs535728289 Chr.8: 54622362-54622362 on Build GRCh38
rs267601948 Chr.8: 54622140-54622140 on Build GRCh38
rs181614973 Chr.8: 54622484-54622484 on Build GRCh38
rs974031024 Chr.8: 54622294-54622294 on Build GRCh38
rs770300288 Chr.8: 54622246-54622246 on Build GRCh38
rs768964344 Chr.8: 54622105-54622105 on Build GRCh38
rs759197247 Chr.8: 54622145-54622145 on Build GRCh38
rs951414813 Chr.8: 54622363-54622363 on Build GRCh38
rs755182172 Chr.8: 54622216-54622216 on Build GRCh38
rs770414773 Chr.8: 54622137-54622137 on Build GRCh38
rs112524889 Chr.8: 54622396-54622396 on Build GRCh38
rs1011228800 Chr.8: 54622066-54622066 on Build GRCh38
rs779213123 Chr.8: 54622110-54622110 on Build GRCh38
rs772717240 Chr.8: 54622121-54622121 on Build GRCh38
rs16920614 Chr.8: 54622253-54622253 on Build GRCh38
rs745452078 Chr.8: 54622164-54622164 on Build GRCh38
rs759131798 Chr.8: 54622265-54622265 on Build GRCh38
rs558204863 Chr.8: 54622452-54622452 on Build GRCh38
rs570242074 Chr.8: 54622322-54622322 on Build GRCh38
rs527236105 Chr.8: 54622150-54622150 on Build GRCh38
rs779154813 Chr.8: 54622070-54622070 on Build GRCh38
rs749753331 Chr.8: 54622333-54622333 on Build GRCh38
rs902899929 Chr.8: 54622100-54622100 on Build GRCh38
rs979920705 Chr.8: 54622051-54622051 on Build GRCh38
rs753890350 Chr.8: 54622327-54622327 on Build GRCh38
rs113763758 Chr.8: 54622287-54622287 on Build GRCh38
rs771715697 Chr.8: 54622271-54622271 on Build GRCh38
rs770470055 Chr.8: 54622249-54622249 on Build GRCh38
rs760485111 Chr.8: 54622299-54622299 on Build GRCh38
rs752485908 Chr.8: 54622157-54622157 on Build GRCh38
rs959499478 Chr.8: 54622482-54622482 on Build GRCh38
rs563819315 Chr.8: 54622141-54622141 on Build GRCh38
rs773889365 Chr.8: 54622259-54622259 on Build GRCh38
rs55933920 Chr.8: 54622103-54622103 on Build GRCh38
rs760561079 Chr.8: 54622163-54622163 on Build GRCh38
rs751781610 Chr.8: 54622212-54622212 on Build GRCh38
rs1040350129 Chr.8: 54622460-54622460 on Build GRCh38
rs369804244 Chr.8: 54622165-54622165 on Build GRCh38
rs750540431 Chr.8: 54622167-54622167 on Build GRCh38
rs867820372 Chr.8: 54622128-54622128 on Build GRCh38
rs762393398 Chr.8: 54622123-54622123 on Build GRCh38

View Full Product Details