Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 73021859-73022379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TERF1
Entrez Gene ID
7013
Gene Name
telomeric repeat binding factor 1
Gene Aliases
PIN2, TRBF1, TRF, TRF1, hTRF1-AS, t-TRF1
UniGene
Hs.442707
Gene Chromosome Location
Chr.8: 73008862-73047752 on Build GRCh38
Transcripts
NM_003218.3 NM_017489.2 XM_005251291.3 XM_005251292.3 XM_011517582.2 XM_017013792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3901485 c.613A>G p.N205D Chr.8: 73022291-73022291 on Build GRCh38
3901487 c.614A>C p.N205T Chr.8: 73022292-73022292 on Build GRCh38
1700263 c.553A>T p.M185L Chr.8: 73022231-73022231 on Build GRCh38
1196783 c.617C>A p.S206Y Chr.8: 73022295-73022295 on Build GRCh38
3901484 c.613A>G p.N205D Chr.8: 73022291-73022291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961123504 Chr.8: 73022113-73022113 on Build GRCh38
rs762213009 Chr.8: 73022305-73022313 on Build GRCh38
rs559198469 Chr.8: 73021985-73021985 on Build GRCh38
rs761306149 Chr.8: 73022336-73022336 on Build GRCh38
rs767652543 Chr.8: 73022171-73022171 on Build GRCh38
rs899715595 Chr.8: 73022120-73022120 on Build GRCh38
rs569397053 Chr.8: 73022122-73022123 on Build GRCh38
rs919944096 Chr.8: 73022049-73022049 on Build GRCh38
rs375420725 Chr.8: 73022042-73022042 on Build GRCh38
rs367991518 Chr.8: 73022198-73022198 on Build GRCh38
rs1042721913 Chr.8: 73022134-73022134 on Build GRCh38
rs374424459 Chr.8: 73022322-73022322 on Build GRCh38
rs550941750 Chr.8: 73022126-73022126 on Build GRCh38
rs943927768 Chr.8: 73022117-73022117 on Build GRCh38
rs754143908 Chr.8: 73022190-73022190 on Build GRCh38
rs370648005 Chr.8: 73021999-73021999 on Build GRCh38
rs776820604 Chr.8: 73022262-73022262 on Build GRCh38
rs934503889 Chr.8: 73021964-73021964 on Build GRCh38
rs1027178011 Chr.8: 73022138-73022138 on Build GRCh38
rs772278457 Chr.8: 73022324-73022324 on Build GRCh38
rs974060064 Chr.8: 73022039-73022039 on Build GRCh38
rs754857546 Chr.8: 73022280-73022280 on Build GRCh38
rs1019790026 Chr.8: 73022086-73022086 on Build GRCh38
rs79263711 Chr.8: 73021912-73021912 on Build GRCh38
rs200522789 Chr.8: 73022167-73022167 on Build GRCh38
rs199736840 Chr.8: 73022193-73022193 on Build GRCh38
rs889489433 Chr.8: 73022077-73022077 on Build GRCh38
rs776074966 Chr.8: 73022183-73022183 on Build GRCh38
rs1040021838 Chr.8: 73022118-73022118 on Build GRCh38
rs201882345 Chr.8: 73022295-73022295 on Build GRCh38
rs376812116 Chr.8: 73022343-73022343 on Build GRCh38
rs781089542 Chr.8: 73022285-73022285 on Build GRCh38
rs1007855620 Chr.8: 73022082-73022082 on Build GRCh38
rs746970786 Chr.8: 73022248-73022248 on Build GRCh38
rs767781920 Chr.8: 73022349-73022350 on Build GRCh38
rs779482581 Chr.8: 73022206-73022206 on Build GRCh38
rs1011478581 Chr.8: 73022041-73022041 on Build GRCh38
rs912379186 Chr.8: 73022111-73022111 on Build GRCh38
rs147608592 Chr.8: 73022291-73022291 on Build GRCh38
rs375336420 Chr.8: 73022184-73022184 on Build GRCh38
rs756956379 Chr.8: 73022191-73022191 on Build GRCh38
rs994761336 Chr.8: 73022136-73022136 on Build GRCh38
rs892955294 Chr.8: 73021973-73021973 on Build GRCh38
rs537682390 Chr.8: 73021892-73021892 on Build GRCh38
rs761323502 Chr.8: 73022045-73022045 on Build GRCh38
rs777097942 Chr.8: 73022351-73022351 on Build GRCh38
rs569185919 Chr.8: 73022148-73022148 on Build GRCh38
rs775423787 Chr.8: 73022335-73022335 on Build GRCh38
rs764403654 Chr.8: 73022195-73022195 on Build GRCh38
rs759717946 Chr.8: 73022169-73022169 on Build GRCh38
rs772319565 Chr.8: 73021993-73021993 on Build GRCh38
rs571502597 Chr.8: 73021970-73021970 on Build GRCh38
rs774773856 Chr.8: 73022257-73022259 on Build GRCh38
rs369059905 Chr.8: 73022195-73022195 on Build GRCh38
rs1047164668 Chr.8: 73022097-73022097 on Build GRCh38

View Full Product Details