Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17576127-17576669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDGFRL
Entrez Gene ID
5157
Gene Name
platelet derived growth factor receptor like
Gene Aliases
PDGRL, PRLTS
UniGene
Hs.458573
Gene Chromosome Location
Chr.8: 17576433-17643133 on Build GRCh38
Transcripts
NM_006207.2

SNP Detail

SNP ID Chromosome Location
rs568898026 Chr.8: 17576532-17576532 on Build GRCh38
rs1051136029 Chr.8: 17576453-17576453 on Build GRCh38
rs1017382300 Chr.8: 17576469-17576469 on Build GRCh38
rs550615921 Chr.8: 17576450-17576450 on Build GRCh38
rs1041405005 Chr.8: 17576238-17576238 on Build GRCh38
rs983940831 Chr.8: 17576193-17576193 on Build GRCh38
rs917000550 Chr.8: 17576498-17576498 on Build GRCh38
rs977054409 Chr.8: 17576406-17576406 on Build GRCh38
rs917129775 Chr.8: 17576399-17576399 on Build GRCh38
rs916889709 Chr.8: 17576415-17576415 on Build GRCh38
rs182826659 Chr.8: 17576497-17576497 on Build GRCh38
rs1047332809 Chr.8: 17576452-17576452 on Build GRCh38
rs967567930 Chr.8: 17576514-17576514 on Build GRCh38
rs937106423 Chr.8: 17576195-17576195 on Build GRCh38
rs532110520 Chr.8: 17576300-17576300 on Build GRCh38
rs530690716 Chr.8: 17576216-17576216 on Build GRCh38
rs550711054 Chr.8: 17576234-17576234 on Build GRCh38
rs984593034 Chr.8: 17576375-17576375 on Build GRCh38
rs900031037 Chr.8: 17576446-17576446 on Build GRCh38
rs888712781 Chr.8: 17576611-17576611 on Build GRCh38
rs952927107 Chr.8: 17576190-17576190 on Build GRCh38
rs998249820 Chr.8: 17576283-17576283 on Build GRCh38
rs774584981 Chr.8: 17576297-17576297 on Build GRCh38
rs140338356 Chr.8: 17576493-17576493 on Build GRCh38
rs1034478354 Chr.8: 17576376-17576376 on Build GRCh38
rs992596675 Chr.8: 17576414-17576414 on Build GRCh38
rs935721559 Chr.8: 17576412-17576412 on Build GRCh38
rs1039831474 Chr.8: 17576444-17576444 on Build GRCh38
rs564202702 Chr.8: 17576620-17576620 on Build GRCh38
rs553745033 Chr.8: 17576572-17576572 on Build GRCh38
rs936280790 Chr.8: 17576428-17576428 on Build GRCh38
rs549611863 Chr.8: 17576418-17576418 on Build GRCh38
rs1014088926 Chr.8: 17576623-17576623 on Build GRCh38
rs762508423 Chr.8: 17576543-17576543 on Build GRCh38
rs941879987 Chr.8: 17576180-17576180 on Build GRCh38
rs977103001 Chr.8: 17576524-17576524 on Build GRCh38
rs751409696 Chr.8: 17576560-17576560 on Build GRCh38
rs992666694 Chr.8: 17576397-17576397 on Build GRCh38
rs546611429 Chr.8: 17576290-17576290 on Build GRCh38
rs577702627 Chr.8: 17576526-17576526 on Build GRCh38
rs1051241000 Chr.8: 17576562-17576562 on Build GRCh38
rs1025613536 Chr.8: 17576303-17576303 on Build GRCh38
rs958778696 Chr.8: 17576490-17576490 on Build GRCh38
rs748257228 Chr.8: 17576153-17576153 on Build GRCh38
rs907674849 Chr.8: 17576175-17576175 on Build GRCh38
rs145456931 Chr.8: 17576586-17576586 on Build GRCh38
rs963311009 Chr.8: 17576470-17576470 on Build GRCh38
rs1004732240 Chr.8: 17576571-17576571 on Build GRCh38
rs373199076 Chr.8: 17576593-17576593 on Build GRCh38
rs150004866 Chr.8: 17576187-17576187 on Build GRCh38
rs963662683 Chr.8: 17576331-17576331 on Build GRCh38
rs765986042 Chr.8: 17576556-17576556 on Build GRCh38
rs191564603 Chr.8: 17576394-17576394 on Build GRCh38
rs1014844180 Chr.8: 17576312-17576312 on Build GRCh38
rs541764712 Chr.8: 17576591-17576591 on Build GRCh38
rs561733942 Chr.8: 17576202-17576202 on Build GRCh38
rs778109974 Chr.8: 17576211-17576211 on Build GRCh38
rs764629229 Chr.8: 17576517-17576517 on Build GRCh38
rs945735605 Chr.8: 17576416-17576416 on Build GRCh38
rs567200905 Chr.8: 17576301-17576301 on Build GRCh38
rs1041985651 Chr.8: 17576225-17576225 on Build GRCh38
rs1034275789 Chr.8: 17576614-17576614 on Build GRCh38
rs1005924256 Chr.8: 17576612-17576612 on Build GRCh38
rs1034811097 Chr.8: 17576475-17576475 on Build GRCh38
rs925593179 Chr.8: 17576409-17576409 on Build GRCh38
rs973251821 Chr.8: 17576392-17576392 on Build GRCh38
rs887067716 Chr.8: 17576458-17576458 on Build GRCh38
rs573042972 Chr.8: 17576173-17576173 on Build GRCh38
rs1056804750 Chr.8: 17576198-17576198 on Build GRCh38
rs904650370 Chr.8: 17576244-17576244 on Build GRCh38
rs973520510 Chr.8: 17576183-17576183 on Build GRCh38
rs1024589558 Chr.8: 17576628-17576628 on Build GRCh38
rs796376750 Chr.8: 17576382-17576382 on Build GRCh38
rs2237819 Chr.8: 17576246-17576246 on Build GRCh38
rs762241836 Chr.8: 17576203-17576206 on Build GRCh38
rs967136951 Chr.8: 17576638-17576638 on Build GRCh38
rs945658379 Chr.8: 17576237-17576240 on Build GRCh38
rs995185430 Chr.8: 17576478-17576478 on Build GRCh38
rs1039948636 Chr.8: 17576538-17576538 on Build GRCh38
rs958541018 Chr.8: 17576489-17576489 on Build GRCh38
rs775937685 Chr.8: 17576370-17576370 on Build GRCh38
rs75865284 Chr.8: 17576607-17576607 on Build GRCh38
rs772592552 Chr.8: 17576344-17576344 on Build GRCh38
rs1034159862 Chr.8: 17576484-17576484 on Build GRCh38
rs921399937 Chr.8: 17576550-17576550 on Build GRCh38
rs926219428 Chr.8: 17576425-17576425 on Build GRCh38
rs1004211751 Chr.8: 17576464-17576464 on Build GRCh38
rs977595065 Chr.8: 17576647-17576647 on Build GRCh38
rs771397076 Chr.8: 17576273-17576273 on Build GRCh38
rs569370293 Chr.8: 17576482-17576482 on Build GRCh38
rs575370573 Chr.8: 17576608-17576608 on Build GRCh38
rs528787321 Chr.8: 17576355-17576356 on Build GRCh38
rs532984584 Chr.8: 17576247-17576247 on Build GRCh38
rs754768193 Chr.8: 17576568-17576568 on Build GRCh38
rs769733844 Chr.8: 17576185-17576185 on Build GRCh38
rs927138916 Chr.8: 17576393-17576393 on Build GRCh38
rs186166389 Chr.8: 17576539-17576539 on Build GRCh38
rs1050209441 Chr.8: 17576272-17576272 on Build GRCh38
rs544398153 Chr.8: 17576618-17576618 on Build GRCh38

View Full Product Details