Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17873917-17874429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGL1
Entrez Gene ID
2267
Gene Name
fibrinogen like 1
Gene Aliases
HFREP1, HP-041, LFIRE-1, LFIRE1
UniGene
Hs.491143
Gene Chromosome Location
Chr.8: 17864380-17909983 on Build GRCh38
Transcripts
NM_004467.3 NM_147203.2 NM_201552.1 NM_201553.1 XM_017013232.1 XM_017013233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1569127 c.390T>A p.S130R Chr.8: 17874376-17874376 on Build GRCh38
1455867 c.436A>C p.N146H Chr.8: 17874085-17874085 on Build GRCh38
4684640 c.397T>G p.F133V Chr.8: 17874369-17874369 on Build GRCh38
4684639 c.476A>C p.K159T Chr.8: 17874045-17874045 on Build GRCh38
6663866 c.346G>A p.D116N Chr.8: 17874420-17874420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753764266 Chr.8: 17874343-17874343 on Build GRCh38
rs750635580 Chr.8: 17874352-17874352 on Build GRCh38
rs752679118 Chr.8: 17874139-17874139 on Build GRCh38
rs770470957 Chr.8: 17874372-17874372 on Build GRCh38
rs1015092883 Chr.8: 17873944-17873944 on Build GRCh38
rs775315817 Chr.8: 17874382-17874382 on Build GRCh38
rs921112442 Chr.8: 17874206-17874206 on Build GRCh38
rs915865316 Chr.8: 17874066-17874066 on Build GRCh38
rs765287617 Chr.8: 17874346-17874346 on Build GRCh38
rs988226157 Chr.8: 17874363-17874363 on Build GRCh38
rs779027445 Chr.8: 17874077-17874077 on Build GRCh38
rs367543613 Chr.8: 17874328-17874328 on Build GRCh38
rs376838318 Chr.8: 17874074-17874074 on Build GRCh38
rs113038731 Chr.8: 17874361-17874361 on Build GRCh38
rs781198781 Chr.8: 17874118-17874119 on Build GRCh38
rs750126281 Chr.8: 17873979-17873979 on Build GRCh38
rs963915746 Chr.8: 17873951-17873951 on Build GRCh38
rs374709305 Chr.8: 17874345-17874345 on Build GRCh38
rs747374996 Chr.8: 17874158-17874158 on Build GRCh38
rs201465199 Chr.8: 17874166-17874166 on Build GRCh38
rs754894707 Chr.8: 17873991-17873991 on Build GRCh38
rs112170258 Chr.8: 17874360-17874360 on Build GRCh38
rs751525129 Chr.8: 17874175-17874175 on Build GRCh38
rs778636832 Chr.8: 17874366-17874367 on Build GRCh38
rs188608946 Chr.8: 17874184-17874184 on Build GRCh38
rs948157915 Chr.8: 17873964-17873964 on Build GRCh38
rs770035105 Chr.8: 17874005-17874005 on Build GRCh38
rs771196321 Chr.8: 17874024-17874024 on Build GRCh38
rs748350363 Chr.8: 17874315-17874315 on Build GRCh38
rs968333930 Chr.8: 17874398-17874398 on Build GRCh38
rs573301828 Chr.8: 17873963-17873963 on Build GRCh38
rs568931562 Chr.8: 17874114-17874114 on Build GRCh38
rs758744653 Chr.8: 17874153-17874153 on Build GRCh38
rs951998699 Chr.8: 17874037-17874037 on Build GRCh38
rs961830746 Chr.8: 17874009-17874009 on Build GRCh38
rs774184250 Chr.8: 17874213-17874213 on Build GRCh38
rs111376859 Chr.8: 17874230-17874230 on Build GRCh38
rs538079958 Chr.8: 17874160-17874160 on Build GRCh38
rs566312769 Chr.8: 17874079-17874079 on Build GRCh38
rs758102179 Chr.8: 17873980-17873980 on Build GRCh38
rs746134514 Chr.8: 17874151-17874151 on Build GRCh38
rs371152619 Chr.8: 17874147-17874147 on Build GRCh38
rs752732031 Chr.8: 17874060-17874060 on Build GRCh38
rs113862747 Chr.8: 17874018-17874018 on Build GRCh38
rs34239530 Chr.8: 17874405-17874405 on Build GRCh38
rs199781222 Chr.8: 17874379-17874379 on Build GRCh38
rs533463660 Chr.8: 17874402-17874402 on Build GRCh38
rs982043562 Chr.8: 17873965-17873965 on Build GRCh38
rs751443993 Chr.8: 17873983-17873983 on Build GRCh38
rs779167895 Chr.8: 17874148-17874148 on Build GRCh38
rs377713527 Chr.8: 17874140-17874140 on Build GRCh38
rs1035089432 Chr.8: 17874375-17874375 on Build GRCh38
rs756250214 Chr.8: 17874064-17874064 on Build GRCh38
rs368503160 Chr.8: 17874354-17874354 on Build GRCh38
rs372364435 Chr.8: 17874376-17874376 on Build GRCh38
rs765950818 Chr.8: 17874050-17874050 on Build GRCh38
rs973500724 Chr.8: 17873956-17873956 on Build GRCh38
rs138392872 Chr.8: 17873972-17873972 on Build GRCh38
rs370532467 Chr.8: 17874330-17874330 on Build GRCh38
rs34353097 Chr.8: 17873955-17873955 on Build GRCh38
rs750028870 Chr.8: 17874042-17874042 on Build GRCh38
rs933726703 Chr.8: 17874207-17874207 on Build GRCh38
rs765222489 Chr.8: 17874392-17874392 on Build GRCh38
rs116032579 Chr.8: 17874198-17874198 on Build GRCh38
rs762730555 Chr.8: 17874047-17874047 on Build GRCh38
rs759833729 Chr.8: 17874027-17874027 on Build GRCh38
rs759329624 Chr.8: 17874325-17874325 on Build GRCh38
rs1024220083 Chr.8: 17874331-17874331 on Build GRCh38
rs755328401 Chr.8: 17874364-17874364 on Build GRCh38
rs780460701 Chr.8: 17874154-17874154 on Build GRCh38
rs927999715 Chr.8: 17874094-17874094 on Build GRCh38
rs747323904 Chr.8: 17874362-17874362 on Build GRCh38
rs982054830 Chr.8: 17874347-17874347 on Build GRCh38
rs760572785 Chr.8: 17874339-17874339 on Build GRCh38
rs764055375 Chr.8: 17874341-17874341 on Build GRCh38
rs757358521 Chr.8: 17874106-17874106 on Build GRCh38
rs374685173 Chr.8: 17874040-17874040 on Build GRCh38
rs564476986 Chr.8: 17874028-17874028 on Build GRCh38
rs749584044 Chr.8: 17874019-17874019 on Build GRCh38
rs779954253 Chr.8: 17873981-17873981 on Build GRCh38
rs766006638 Chr.8: 17874123-17874123 on Build GRCh38
rs773905546 Chr.8: 17874125-17874125 on Build GRCh38
rs781234321 Chr.8: 17874056-17874056 on Build GRCh38
rs1012735089 Chr.8: 17874286-17874286 on Build GRCh38
rs774713490 Chr.8: 17874025-17874025 on Build GRCh38
rs767478579 Chr.8: 17874132-17874132 on Build GRCh38
rs768389622 Chr.8: 17874389-17874389 on Build GRCh38
rs573104574 Chr.8: 17874248-17874248 on Build GRCh38
rs764832375 Chr.8: 17873976-17873976 on Build GRCh38
rs1011170219 Chr.8: 17873953-17873953 on Build GRCh38
rs772469816 Chr.8: 17874101-17874101 on Build GRCh38
rs35431851 Chr.8: 17874102-17874102 on Build GRCh38
rs1050663755 Chr.8: 17874176-17874176 on Build GRCh38
rs200686011 Chr.8: 17874359-17874359 on Build GRCh38
rs771804269 Chr.8: 17874381-17874381 on Build GRCh38
rs748189753 Chr.8: 17874003-17874003 on Build GRCh38
rs761278944 Chr.8: 17874032-17874032 on Build GRCh38
rs995102614 Chr.8: 17874302-17874302 on Build GRCh38
rs956731771 Chr.8: 17874172-17874172 on Build GRCh38
rs946666759 Chr.8: 17874190-17874190 on Build GRCh38
rs749940982 Chr.8: 17873969-17873969 on Build GRCh38
rs114704900 Chr.8: 17874279-17874279 on Build GRCh38
rs1022801063 Chr.8: 17874310-17874310 on Build GRCh38
rs34871936 Chr.8: 17874367-17874367 on Build GRCh38
rs541914162 Chr.8: 17873968-17873968 on Build GRCh38
rs1023813225 Chr.8: 17874133-17874133 on Build GRCh38
rs886761909 Chr.8: 17874253-17874253 on Build GRCh38
rs1008535991 Chr.8: 17874197-17874197 on Build GRCh38
rs145346939 Chr.8: 17873954-17873955 on Build GRCh38
rs993276081 Chr.8: 17874233-17874233 on Build GRCh38
rs576102008 Chr.8: 17874221-17874221 on Build GRCh38
rs375560744 Chr.8: 17874011-17874011 on Build GRCh38
rs373250063 Chr.8: 17874072-17874072 on Build GRCh38
rs748791444 Chr.8: 17874313-17874313 on Build GRCh38
rs749194783 Chr.8: 17874157-17874157 on Build GRCh38
rs371189982 Chr.8: 17874095-17874095 on Build GRCh38
rs142466248 Chr.8: 17873993-17873993 on Build GRCh38
rs760490854 Chr.8: 17874388-17874388 on Build GRCh38
rs754841694 Chr.8: 17874054-17874054 on Build GRCh38
rs369302932 Chr.8: 17874052-17874052 on Build GRCh38
rs899384615 Chr.8: 17874290-17874290 on Build GRCh38
rs61744652 Chr.8: 17874390-17874390 on Build GRCh38
rs150113828 Chr.8: 17874113-17874113 on Build GRCh38
rs988055954 Chr.8: 17874174-17874176 on Build GRCh38
rs147591340 Chr.8: 17874068-17874068 on Build GRCh38
rs145205778 Chr.8: 17874036-17874036 on Build GRCh38
rs578014942 Chr.8: 17874173-17874173 on Build GRCh38
rs754874876 Chr.8: 17874364-17874369 on Build GRCh38
rs748661649 Chr.8: 17874370-17874370 on Build GRCh38
rs374582234 Chr.8: 17874126-17874126 on Build GRCh38
rs769287820 Chr.8: 17874109-17874109 on Build GRCh38
rs544437871 Chr.8: 17873955-17873956 on Build GRCh38

View Full Product Details