Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23432798-23433299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD4
Entrez Gene ID
9583
Gene Name
ectonucleoside triphosphate diphosphohydrolase 4
Gene Aliases
LALP70, LAP70, LYSAL1, NTPDase-4, UDPase
UniGene
Hs.444389
Gene Chromosome Location
Chr.8: 23429152-23457731 on Build GRCh38
Transcripts
NM_001128930.2 NM_004901.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
300470 c.1798G>A p.A600T Chr.8: 23432979-23432979 on Build GRCh38
5459358 c.1723C>G p.L575V Chr.8: 23433054-23433054 on Build GRCh38
4751209 c.1661A>T p.H554L Chr.8: 23433116-23433116 on Build GRCh38
1456164 c.1756C>T p.R586W Chr.8: 23433021-23433021 on Build GRCh38
2786674 c.1750C>T p.R584W Chr.8: 23433027-23433027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556405061 Chr.8: 23432979-23432979 on Build GRCh38
rs762950631 Chr.8: 23433100-23433100 on Build GRCh38
rs755651058 Chr.8: 23432996-23432996 on Build GRCh38
rs779338335 Chr.8: 23433227-23433227 on Build GRCh38
rs752930736 Chr.8: 23433186-23433186 on Build GRCh38
rs1033927281 Chr.8: 23432990-23432990 on Build GRCh38
rs971814133 Chr.8: 23433026-23433026 on Build GRCh38
rs770959664 Chr.8: 23433018-23433023 on Build GRCh38
rs565281949 Chr.8: 23433067-23433067 on Build GRCh38
rs202242195 Chr.8: 23433017-23433017 on Build GRCh38
rs765670701 Chr.8: 23432949-23432949 on Build GRCh38
rs750252103 Chr.8: 23433020-23433020 on Build GRCh38
rs368920976 Chr.8: 23433181-23433182 on Build GRCh38
rs751262101 Chr.8: 23433106-23433106 on Build GRCh38
rs866761610 Chr.8: 23432928-23432928 on Build GRCh38
rs200186299 Chr.8: 23433005-23433005 on Build GRCh38
rs747049248 Chr.8: 23432940-23432940 on Build GRCh38
rs368598181 Chr.8: 23433167-23433167 on Build GRCh38
rs548817345 Chr.8: 23432878-23432878 on Build GRCh38
rs781204181 Chr.8: 23433110-23433110 on Build GRCh38
rs3736021 Chr.8: 23433256-23433256 on Build GRCh38
rs570420189 Chr.8: 23432952-23432952 on Build GRCh38
rs753952215 Chr.8: 23433222-23433222 on Build GRCh38
rs779360023 Chr.8: 23433132-23433132 on Build GRCh38
rs749052028 Chr.8: 23432926-23432926 on Build GRCh38
rs778569333 Chr.8: 23432934-23432934 on Build GRCh38
rs537811155 Chr.8: 23433264-23433264 on Build GRCh38
rs986553603 Chr.8: 23433001-23433001 on Build GRCh38
rs369574181 Chr.8: 23433027-23433027 on Build GRCh38
rs368571477 Chr.8: 23432983-23432983 on Build GRCh38
rs1025861169 Chr.8: 23433215-23433215 on Build GRCh38
rs756245715 Chr.8: 23433119-23433119 on Build GRCh38
rs893870045 Chr.8: 23432959-23432959 on Build GRCh38
rs769692133 Chr.8: 23432860-23432861 on Build GRCh38
rs761518791 Chr.8: 23433148-23433148 on Build GRCh38
rs753867940 Chr.8: 23433203-23433203 on Build GRCh38
rs751275484 Chr.8: 23432888-23432888 on Build GRCh38
rs370967876 Chr.8: 23432918-23432918 on Build GRCh38
rs776240690 Chr.8: 23433082-23433082 on Build GRCh38
rs778905365 Chr.8: 23432980-23432980 on Build GRCh38
rs749836030 Chr.8: 23433129-23433129 on Build GRCh38
rs781352847 Chr.8: 23433058-23433058 on Build GRCh38
rs367984909 Chr.8: 23432986-23432986 on Build GRCh38
rs764237088 Chr.8: 23433219-23433219 on Build GRCh38
rs760240348 Chr.8: 23433006-23433006 on Build GRCh38
rs767329862 Chr.8: 23433185-23433185 on Build GRCh38
rs549530914 Chr.8: 23433271-23433271 on Build GRCh38
rs749783482 Chr.8: 23433069-23433069 on Build GRCh38
rs17089244 Chr.8: 23432938-23432938 on Build GRCh38
rs760427490 Chr.8: 23433079-23433079 on Build GRCh38
rs749546191 Chr.8: 23432938-23432939 on Build GRCh38
rs776093882 Chr.8: 23433142-23433142 on Build GRCh38
rs772822867 Chr.8: 23433161-23433161 on Build GRCh38
rs979183358 Chr.8: 23433228-23433228 on Build GRCh38
rs908953004 Chr.8: 23432868-23432868 on Build GRCh38
rs560678563 Chr.8: 23433225-23433225 on Build GRCh38
rs774498491 Chr.8: 23432986-23432987 on Build GRCh38
rs763501000 Chr.8: 23432975-23432975 on Build GRCh38
rs773542726 Chr.8: 23432951-23432951 on Build GRCh38
rs530441511 Chr.8: 23433189-23433189 on Build GRCh38
rs138511953 Chr.8: 23432987-23432987 on Build GRCh38
rs747293465 Chr.8: 23433025-23433025 on Build GRCh38
rs372556071 Chr.8: 23433060-23433060 on Build GRCh38
rs770818367 Chr.8: 23432931-23432931 on Build GRCh38
rs375242682 Chr.8: 23433021-23433021 on Build GRCh38
rs376644779 Chr.8: 23433088-23433088 on Build GRCh38
rs1017209730 Chr.8: 23432982-23432982 on Build GRCh38
rs763003308 Chr.8: 23433163-23433163 on Build GRCh38
rs753880920 Chr.8: 23432977-23432977 on Build GRCh38
rs548755865 Chr.8: 23433223-23433223 on Build GRCh38
rs776430787 Chr.8: 23433053-23433054 on Build GRCh38
rs774669675 Chr.8: 23433077-23433077 on Build GRCh38
rs934979952 Chr.8: 23433239-23433239 on Build GRCh38
rs777825999 Chr.8: 23433120-23433120 on Build GRCh38
rs889456576 Chr.8: 23432838-23432838 on Build GRCh38
rs769054012 Chr.8: 23432942-23432942 on Build GRCh38
rs751371953 Chr.8: 23433171-23433171 on Build GRCh38
rs768724699 Chr.8: 23432983-23432989 on Build GRCh38
rs770338186 Chr.8: 23433061-23433061 on Build GRCh38
rs755257966 Chr.8: 23433109-23433109 on Build GRCh38
rs765229113 Chr.8: 23433019-23433019 on Build GRCh38
rs1046443934 Chr.8: 23432898-23432908 on Build GRCh38
rs746180754 Chr.8: 23433134-23433134 on Build GRCh38
rs138795546 Chr.8: 23432953-23432953 on Build GRCh38
rs1001519881 Chr.8: 23433160-23433160 on Build GRCh38
rs759194265 Chr.8: 23433184-23433184 on Build GRCh38
rs201075031 Chr.8: 23432907-23432907 on Build GRCh38
rs752835265 Chr.8: 23433111-23433111 on Build GRCh38
rs552225506 Chr.8: 23433255-23433255 on Build GRCh38
rs376465431 Chr.8: 23432913-23432913 on Build GRCh38
rs544314711 Chr.8: 23433086-23433086 on Build GRCh38
rs752151301 Chr.8: 23432909-23432909 on Build GRCh38
rs760898941 Chr.8: 23432939-23432939 on Build GRCh38
rs773595870 Chr.8: 23432997-23432997 on Build GRCh38
rs1050807690 Chr.8: 23432830-23432830 on Build GRCh38
rs757245710 Chr.8: 23433018-23433018 on Build GRCh38
rs768362885 Chr.8: 23433139-23433139 on Build GRCh38
rs771406410 Chr.8: 23433131-23433131 on Build GRCh38
rs771352806 Chr.8: 23433070-23433070 on Build GRCh38
rs767171536 Chr.8: 23433000-23433000 on Build GRCh38
rs755593892 Chr.8: 23432971-23432971 on Build GRCh38
rs574404418 Chr.8: 23432981-23432981 on Build GRCh38
rs201149309 Chr.8: 23433081-23433081 on Build GRCh38
rs754593207 Chr.8: 23432900-23432900 on Build GRCh38
rs919094634 Chr.8: 23433039-23433039 on Build GRCh38
rs762218327 Chr.8: 23432948-23432948 on Build GRCh38
rs761741968 Chr.8: 23432996-23432997 on Build GRCh38
rs769465195 Chr.8: 23433158-23433158 on Build GRCh38
rs780475296 Chr.8: 23432909-23432909 on Build GRCh38
rs779986459 Chr.8: 23433024-23433024 on Build GRCh38
rs761165634 Chr.8: 23433083-23433083 on Build GRCh38
rs766323650 Chr.8: 23433105-23433105 on Build GRCh38
rs533879942 Chr.8: 23432925-23432925 on Build GRCh38
rs942307847 Chr.8: 23432866-23432866 on Build GRCh38
rs763700848 Chr.8: 23433007-23433007 on Build GRCh38
rs527921883 Chr.8: 23433235-23433235 on Build GRCh38
rs1053696257 Chr.8: 23433202-23433202 on Build GRCh38
rs752206452 Chr.8: 23432958-23432958 on Build GRCh38
rs373833962 Chr.8: 23432945-23432945 on Build GRCh38

View Full Product Details