Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144510253-144510757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD3
Entrez Gene ID
113655
Gene Name
major facilitator superfamily domain containing 3
Gene Aliases
-
UniGene
Hs.7678
Gene Chromosome Location
Chr.8: 144508081-144511228 on Build GRCh38
Transcripts
NM_138431.2 NR_130120.1 XM_011516806.2 XM_017013005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6313594 c.750G>T p.L250F Chr.8: 144510427-144510427 on Build GRCh38
3898298 c.948C>G p.I316M Chr.8: 144510715-144510715 on Build GRCh38
5979647 c.778G>T p.V260F Chr.8: 144510455-144510455 on Build GRCh38
6030130 c.705G>A p.L235L Chr.8: 144510382-144510382 on Build GRCh38
6112931 c.873C>T p.L291L Chr.8: 144510640-144510640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750699928 Chr.8: 144510721-144510721 on Build GRCh38
rs1053123306 Chr.8: 144510446-144510446 on Build GRCh38
rs779110997 Chr.8: 144510599-144510599 on Build GRCh38
rs765943534 Chr.8: 144510718-144510719 on Build GRCh38
rs180675502 Chr.8: 144510489-144510489 on Build GRCh38
rs35464919 Chr.8: 144510728-144510728 on Build GRCh38
rs770281720 Chr.8: 144510349-144510349 on Build GRCh38
rs770191819 Chr.8: 144510400-144510400 on Build GRCh38
rs1008476507 Chr.8: 144510497-144510497 on Build GRCh38
rs202096147 Chr.8: 144510368-144510368 on Build GRCh38
rs565059578 Chr.8: 144510308-144510308 on Build GRCh38
rs767091045 Chr.8: 144510364-144510364 on Build GRCh38
rs781453183 Chr.8: 144510438-144510438 on Build GRCh38
rs928876633 Chr.8: 144510418-144510418 on Build GRCh38
rs753878066 Chr.8: 144510376-144510376 on Build GRCh38
rs769426171 Chr.8: 144510557-144510557 on Build GRCh38
rs967888697 Chr.8: 144510658-144510658 on Build GRCh38
rs763288657 Chr.8: 144510704-144510704 on Build GRCh38
rs762733065 Chr.8: 144510562-144510562 on Build GRCh38
rs749669063 Chr.8: 144510458-144510458 on Build GRCh38
rs756059165 Chr.8: 144510335-144510335 on Build GRCh38
rs769807864 Chr.8: 144510342-144510342 on Build GRCh38
rs1001531004 Chr.8: 144510350-144510350 on Build GRCh38
rs144605322 Chr.8: 144510635-144510635 on Build GRCh38
rs746040263 Chr.8: 144510441-144510441 on Build GRCh38
rs371459771 Chr.8: 144510348-144510348 on Build GRCh38
rs762077494 Chr.8: 144510585-144510586 on Build GRCh38
rs754198388 Chr.8: 144510523-144510523 on Build GRCh38
rs887510785 Chr.8: 144510288-144510288 on Build GRCh38
rs975925947 Chr.8: 144510660-144510660 on Build GRCh38
rs749757086 Chr.8: 144510407-144510407 on Build GRCh38
rs755255821 Chr.8: 144510593-144510593 on Build GRCh38
rs952339925 Chr.8: 144510566-144510566 on Build GRCh38
rs367661925 Chr.8: 144510394-144510394 on Build GRCh38
rs756875758 Chr.8: 144510613-144510613 on Build GRCh38
rs953319199 Chr.8: 144510676-144510676 on Build GRCh38
rs760813047 Chr.8: 144510575-144510575 on Build GRCh38
rs35905340 Chr.8: 144510699-144510699 on Build GRCh38
rs763703358 Chr.8: 144510321-144510321 on Build GRCh38
rs55797497 Chr.8: 144510619-144510619 on Build GRCh38
rs758956615 Chr.8: 144510607-144510607 on Build GRCh38
rs376881663 Chr.8: 144510347-144510347 on Build GRCh38
rs368331613 Chr.8: 144510727-144510727 on Build GRCh38
rs779455089 Chr.8: 144510533-144510533 on Build GRCh38
rs756270582 Chr.8: 144510521-144510521 on Build GRCh38
rs888145221 Chr.8: 144510725-144510725 on Build GRCh38
rs749166353 Chr.8: 144510623-144510623 on Build GRCh38
rs753687366 Chr.8: 144510427-144510427 on Build GRCh38
rs370613210 Chr.8: 144510327-144510327 on Build GRCh38
rs34730139 Chr.8: 144510718-144510718 on Build GRCh38
rs750900055 Chr.8: 144510659-144510660 on Build GRCh38
rs746573139 Chr.8: 144510541-144510541 on Build GRCh38
rs540663905 Chr.8: 144510332-144510332 on Build GRCh38
rs372083866 Chr.8: 144510474-144510474 on Build GRCh38
rs536613878 Chr.8: 144510467-144510467 on Build GRCh38
rs1036078871 Chr.8: 144510532-144510532 on Build GRCh38
rs747211889 Chr.8: 144510346-144510346 on Build GRCh38
rs773995987 Chr.8: 144510412-144510412 on Build GRCh38
rs376179435 Chr.8: 144510729-144510729 on Build GRCh38
rs776556982 Chr.8: 144510689-144510689 on Build GRCh38
rs541790061 Chr.8: 144510680-144510680 on Build GRCh38
rs199612909 Chr.8: 144510395-144510395 on Build GRCh38
rs766560298 Chr.8: 144510582-144510582 on Build GRCh38
rs543586036 Chr.8: 144510283-144510283 on Build GRCh38
rs774314606 Chr.8: 144510630-144510630 on Build GRCh38
rs766231600 Chr.8: 144510425-144510425 on Build GRCh38
rs769194537 Chr.8: 144510466-144510466 on Build GRCh38
rs138616461 Chr.8: 144510629-144510629 on Build GRCh38
rs61741211 Chr.8: 144510484-144510484 on Build GRCh38
rs779825312 Chr.8: 144510663-144510663 on Build GRCh38
rs760527755 Chr.8: 144510424-144510424 on Build GRCh38
rs563382310 Chr.8: 144510362-144510362 on Build GRCh38
rs775107905 Chr.8: 144510551-144510551 on Build GRCh38
rs777840926 Chr.8: 144510389-144510389 on Build GRCh38
rs756044796 Chr.8: 144510371-144510371 on Build GRCh38
rs373768263 Chr.8: 144510375-144510375 on Build GRCh38
rs765344428 Chr.8: 144510697-144510697 on Build GRCh38
rs772875570 Chr.8: 144510480-144510480 on Build GRCh38
rs908916788 Chr.8: 144510518-144510518 on Build GRCh38
rs768194476 Chr.8: 144510616-144510617 on Build GRCh38
rs755341003 Chr.8: 144510524-144510524 on Build GRCh38
rs759614660 Chr.8: 144510636-144510636 on Build GRCh38
rs750506918 Chr.8: 144510520-144510520 on Build GRCh38
rs143475431 Chr.8: 144510655-144510655 on Build GRCh38
rs773854713 Chr.8: 144510416-144510416 on Build GRCh38
rs768737318 Chr.8: 144510508-144510509 on Build GRCh38
rs199772518 Chr.8: 144510508-144510508 on Build GRCh38
rs923192525 Chr.8: 144510483-144510483 on Build GRCh38
rs752061795 Chr.8: 144510645-144510645 on Build GRCh38
rs780500504 Chr.8: 144510647-144510647 on Build GRCh38
rs994047768 Chr.8: 144510722-144510722 on Build GRCh38
rs774817675 Chr.8: 144510523-144510524 on Build GRCh38
rs747176961 Chr.8: 144510733-144510733 on Build GRCh38
rs759312515 Chr.8: 144510494-144510494 on Build GRCh38
rs766956423 Chr.8: 144510522-144510522 on Build GRCh38
rs139574369 Chr.8: 144510363-144510363 on Build GRCh38
rs2306387 Chr.8: 144510641-144510641 on Build GRCh38
rs767543732 Chr.8: 144510512-144510512 on Build GRCh38
rs781328648 Chr.8: 144510637-144510637 on Build GRCh38
rs748559172 Chr.8: 144510536-144510536 on Build GRCh38
rs1053218934 Chr.8: 144510295-144510295 on Build GRCh38
rs770700362 Chr.8: 144510542-144510542 on Build GRCh38
rs762426427 Chr.8: 144510357-144510357 on Build GRCh38
rs35971959 Chr.8: 144510703-144510703 on Build GRCh38
rs775090208 Chr.8: 144510622-144510622 on Build GRCh38
rs755708860 Chr.8: 144510657-144510657 on Build GRCh38
rs754993952 Chr.8: 144510387-144510387 on Build GRCh38
rs897049707 Chr.8: 144510519-144510519 on Build GRCh38
rs757319064 Chr.8: 144510437-144510437 on Build GRCh38
rs776551858 Chr.8: 144510493-144510493 on Build GRCh38
rs202144010 Chr.8: 144510621-144510621 on Build GRCh38
rs766400259 Chr.8: 144510634-144510634 on Build GRCh38
rs747069149 Chr.8: 144510392-144510392 on Build GRCh38
rs760262492 Chr.8: 144510704-144510729 on Build GRCh38
rs1006008507 Chr.8: 144510294-144510294 on Build GRCh38
rs566101371 Chr.8: 144510435-144510435 on Build GRCh38
rs142874249 Chr.8: 144510681-144510681 on Build GRCh38
rs7837302 Chr.8: 144510694-144510694 on Build GRCh38
rs772321274 Chr.8: 144510344-144510344 on Build GRCh38
rs765188043 Chr.8: 144510502-144510502 on Build GRCh38
rs776462031 Chr.8: 144510546-144510546 on Build GRCh38
rs941679172 Chr.8: 144510530-144510530 on Build GRCh38
rs777357624 Chr.8: 144510540-144510540 on Build GRCh38
rs1020301113 Chr.8: 144510654-144510654 on Build GRCh38
rs768129458 Chr.8: 144510716-144510716 on Build GRCh38
rs111396958 Chr.8: 144510341-144510342 on Build GRCh38
rs753460200 Chr.8: 144510731-144510735 on Build GRCh38
rs747953965 Chr.8: 144510734-144510734 on Build GRCh38
rs754060057 Chr.8: 144510586-144510586 on Build GRCh38
rs764511752 Chr.8: 144510705-144510705 on Build GRCh38
rs34906397 Chr.8: 144510343-144510343 on Build GRCh38
rs778488465 Chr.8: 144510608-144510608 on Build GRCh38
rs61734979 Chr.8: 144510410-144510410 on Build GRCh38
rs34073765 Chr.8: 144510309-144510309 on Build GRCh38
rs757870611 Chr.8: 144510646-144510646 on Build GRCh38
rs780329971 Chr.8: 144510455-144510455 on Build GRCh38
rs762516411 Chr.8: 144510314-144510314 on Build GRCh38
rs761288446 Chr.8: 144510421-144510421 on Build GRCh38
rs772005078 Chr.8: 144510688-144510688 on Build GRCh38
rs753115454 Chr.8: 144510601-144510601 on Build GRCh38
rs763612704 Chr.8: 144510361-144510361 on Build GRCh38
rs375058104 Chr.8: 144510723-144510723 on Build GRCh38
rs148527481 Chr.8: 144510673-144510673 on Build GRCh38
rs772669722 Chr.8: 144510538-144510538 on Build GRCh38
rs746483333 Chr.8: 144510612-144510612 on Build GRCh38
rs141941617 Chr.8: 144510712-144510712 on Build GRCh38
rs35731908 Chr.8: 144510431-144510431 on Build GRCh38
rs751652681 Chr.8: 144510433-144510433 on Build GRCh38
rs201878765 Chr.8: 144510714-144510714 on Build GRCh38
rs752996137 Chr.8: 144510640-144510640 on Build GRCh38
rs1019551259 Chr.8: 144510499-144510499 on Build GRCh38
rs773043880 Chr.8: 144510422-144510422 on Build GRCh38
rs775853883 Chr.8: 144510404-144510404 on Build GRCh38
rs750490585 Chr.8: 144510567-144510567 on Build GRCh38
rs571830575 Chr.8: 144510392-144510394 on Build GRCh38
rs186355356 Chr.8: 144510342-144510342 on Build GRCh38
rs948864680 Chr.8: 144510406-144510406 on Build GRCh38
rs752624252 Chr.8: 144510506-144510506 on Build GRCh38
rs756360431 Chr.8: 144510451-144510451 on Build GRCh38
rs746200071 Chr.8: 144510398-144510398 on Build GRCh38
rs780869893 Chr.8: 144510615-144510615 on Build GRCh38
rs201469367 Chr.8: 144510430-144510430 on Build GRCh38
rs764032496 Chr.8: 144510564-144510564 on Build GRCh38
rs775386169 Chr.8: 144510558-144510558 on Build GRCh38
rs893214510 Chr.8: 144510306-144510306 on Build GRCh38
rs763186152 Chr.8: 144510419-144510419 on Build GRCh38
rs751273651 Chr.8: 144510323-144510323 on Build GRCh38

View Full Product Details