Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5029657-5030185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSL6
Entrez Gene ID
11172
Gene Name
insulin like 6
Gene Aliases
RIF1
UniGene
Hs.632648
Gene Chromosome Location
Chr.9: 4992408-5185706 on Build GRCh38
Transcripts
XM_011517702.2 NM_001322194.1 NM_001322195.1 NM_001322196.1 NM_001322198.1 NM_001322199.1 NM_004972.3

Gene Symbol
JAK2
Entrez Gene ID
3717
Gene Name
Janus kinase 2
Gene Aliases
JTK10, THCYT3
UniGene
Hs.656213
Gene Chromosome Location
Chr.9: 4985086-5128183 on Build GRCh38
Transcripts
XM_011517702.2 NM_001322194.1 NM_001322195.1 NM_001322196.1 NM_001322198.1 NM_001322199.1 NM_004972.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6972267 c.245A>T p.H82L Chr.9: 5029801-5029801 on Build GRCh38
5876294 c.296A>G p.N99S Chr.9: 5029852-5029852 on Build GRCh38
455997 c.344G>T p.R115I Chr.9: 5029900-5029900 on Build GRCh38
6488673 c.274G>A p.E92K Chr.9: 5029830-5029830 on Build GRCh38
6929228 c.301G>A p.V101I Chr.9: 5029857-5029857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967660278 Chr.9: 5030031-5030031 on Build GRCh38
rs771625963 Chr.9: 5029923-5029923 on Build GRCh38
rs756548720 Chr.9: 5029770-5029770 on Build GRCh38
rs868488092 Chr.9: 5030160-5030160 on Build GRCh38
rs768067545 Chr.9: 5029880-5029880 on Build GRCh38
rs760494411 Chr.9: 5029739-5029739 on Build GRCh38
rs1028719793 Chr.9: 5030012-5030012 on Build GRCh38
rs758461525 Chr.9: 5029755-5029755 on Build GRCh38
rs183799456 Chr.9: 5030060-5030060 on Build GRCh38
rs139093384 Chr.9: 5029787-5029787 on Build GRCh38
rs757756942 Chr.9: 5029764-5029764 on Build GRCh38
rs772815990 Chr.9: 5029939-5029940 on Build GRCh38
rs200860354 Chr.9: 5029737-5029737 on Build GRCh38
rs761978965 Chr.9: 5029879-5029879 on Build GRCh38
rs756530639 Chr.9: 5029902-5029902 on Build GRCh38
rs771969118 Chr.9: 5029936-5029937 on Build GRCh38
rs982734832 Chr.9: 5029990-5029990 on Build GRCh38
rs773772702 Chr.9: 5029807-5029807 on Build GRCh38
rs763128346 Chr.9: 5029872-5029872 on Build GRCh38
rs766188228 Chr.9: 5029942-5029945 on Build GRCh38
rs753508497 Chr.9: 5029912-5029912 on Build GRCh38
rs925952048 Chr.9: 5030063-5030063 on Build GRCh38
rs752078903 Chr.9: 5029761-5029761 on Build GRCh38
rs116316410 Chr.9: 5029745-5029745 on Build GRCh38
rs765377116 Chr.9: 5029741-5029741 on Build GRCh38
rs764149019 Chr.9: 5029760-5029760 on Build GRCh38
rs951340485 Chr.9: 5029984-5029984 on Build GRCh38
rs751548297 Chr.9: 5029878-5029878 on Build GRCh38
rs927794877 Chr.9: 5030092-5030092 on Build GRCh38
rs915966955 Chr.9: 5030085-5030085 on Build GRCh38
rs947345256 Chr.9: 5030061-5030061 on Build GRCh38
rs767311663 Chr.9: 5029733-5029733 on Build GRCh38
rs201709745 Chr.9: 5029962-5029962 on Build GRCh38
rs778649664 Chr.9: 5029915-5029915 on Build GRCh38
rs868276307 Chr.9: 5030004-5030004 on Build GRCh38
rs1013063937 Chr.9: 5030104-5030104 on Build GRCh38
rs74715056 Chr.9: 5030080-5030080 on Build GRCh38
rs895976288 Chr.9: 5030097-5030097 on Build GRCh38
rs975475520 Chr.9: 5030019-5030019 on Build GRCh38
rs201075771 Chr.9: 5029917-5029917 on Build GRCh38
rs943880654 Chr.9: 5030009-5030009 on Build GRCh38
rs904957031 Chr.9: 5030140-5030140 on Build GRCh38
rs921376064 Chr.9: 5030049-5030049 on Build GRCh38
rs775688165 Chr.9: 5029865-5029865 on Build GRCh38
rs771864517 Chr.9: 5029847-5029847 on Build GRCh38
rs946266987 Chr.9: 5029698-5029698 on Build GRCh38
rs374591930 Chr.9: 5029914-5029914 on Build GRCh38
rs760684817 Chr.9: 5029940-5029941 on Build GRCh38
rs760406225 Chr.9: 5029858-5029858 on Build GRCh38
rs780630654 Chr.9: 5029908-5029908 on Build GRCh38
rs764215783 Chr.9: 5029875-5029875 on Build GRCh38
rs753745588 Chr.9: 5029948-5029959 on Build GRCh38
rs371104486 Chr.9: 5029856-5029856 on Build GRCh38
rs143103233 Chr.9: 5029893-5029893 on Build GRCh38
rs1050502873 Chr.9: 5030157-5030160 on Build GRCh38
rs777676408 Chr.9: 5029924-5029924 on Build GRCh38
rs953686513 Chr.9: 5030016-5030016 on Build GRCh38
rs749005257 Chr.9: 5029780-5029780 on Build GRCh38
rs912389883 Chr.9: 5030003-5030003 on Build GRCh38
rs562092300 Chr.9: 5030090-5030090 on Build GRCh38
rs1014579311 Chr.9: 5029997-5029997 on Build GRCh38
rs770772508 Chr.9: 5029864-5029864 on Build GRCh38
rs987666398 Chr.9: 5030018-5030018 on Build GRCh38
rs150760370 Chr.9: 5029706-5029706 on Build GRCh38
rs781735942 Chr.9: 5029765-5029765 on Build GRCh38
rs957353307 Chr.9: 5030073-5030073 on Build GRCh38
rs903395086 Chr.9: 5030086-5030086 on Build GRCh38
rs977661032 Chr.9: 5030050-5030050 on Build GRCh38
rs774194613 Chr.9: 5029738-5029739 on Build GRCh38
rs753722068 Chr.9: 5030146-5030146 on Build GRCh38
rs1055847130 Chr.9: 5029772-5029776 on Build GRCh38
rs761542465 Chr.9: 5029910-5029913 on Build GRCh38
rs531385688 Chr.9: 5030113-5030113 on Build GRCh38
rs1019104275 Chr.9: 5030023-5030023 on Build GRCh38
rs574820223 Chr.9: 5030128-5030128 on Build GRCh38
rs969443461 Chr.9: 5029884-5029884 on Build GRCh38
rs950121677 Chr.9: 5030088-5030088 on Build GRCh38
rs201577635 Chr.9: 5029776-5029776 on Build GRCh38
rs995548660 Chr.9: 5030143-5030143 on Build GRCh38
rs746191004 Chr.9: 5029767-5029767 on Build GRCh38
rs925972458 Chr.9: 5029717-5029717 on Build GRCh38
rs188176896 Chr.9: 5030089-5030089 on Build GRCh38
rs748095804 Chr.9: 5029833-5029833 on Build GRCh38

View Full Product Details