Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5908720-5909216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA2026
Entrez Gene ID
158358
Gene Name
KIAA2026
Gene Aliases
-
UniGene
Hs.535060
Gene Chromosome Location
Chr.9: 5860254-6008479 on Build GRCh38
Transcripts
XM_011517759.2 XM_011517760.2 XM_017014335.1 NM_005511.1

Gene Symbol
MLANA
Entrez Gene ID
2315
Gene Name
melan-A
Gene Aliases
MART-1, MART1
UniGene
Hs.154069
Gene Chromosome Location
Chr.9: 5890909-5909822 on Build GRCh38
Transcripts
XM_011517759.2 XM_011517760.2 XM_017014335.1 NM_005511.1

SNP Detail

SNP ID Chromosome Location
rs949238034 Chr.9: 5908918-5908918 on Build GRCh38
rs1022704561 Chr.9: 5909004-5909004 on Build GRCh38
rs559650504 Chr.9: 5909029-5909029 on Build GRCh38
rs886798444 Chr.9: 5909005-5909005 on Build GRCh38
rs543854573 Chr.9: 5908845-5908845 on Build GRCh38
rs992840182 Chr.9: 5909114-5909114 on Build GRCh38
rs886855127 Chr.9: 5909156-5909156 on Build GRCh38
rs533410956 Chr.9: 5909053-5909053 on Build GRCh38
rs924582525 Chr.9: 5908867-5908867 on Build GRCh38
rs1015677536 Chr.9: 5909173-5909173 on Build GRCh38
rs529289261 Chr.9: 5908883-5908883 on Build GRCh38
rs971118572 Chr.9: 5909017-5909017 on Build GRCh38
rs957497266 Chr.9: 5908957-5908957 on Build GRCh38
rs949249297 Chr.9: 5909126-5909126 on Build GRCh38
rs546101193 Chr.9: 5908933-5908933 on Build GRCh38
rs1027031712 Chr.9: 5908771-5908771 on Build GRCh38
rs932504655 Chr.9: 5909087-5909087 on Build GRCh38
rs1056796 Chr.9: 5909152-5909152 on Build GRCh38
rs924704793 Chr.9: 5909108-5909108 on Build GRCh38
rs749425206 Chr.9: 5908754-5908754 on Build GRCh38
rs942768504 Chr.9: 5908889-5908889 on Build GRCh38
rs1028415091 Chr.9: 5909092-5909092 on Build GRCh38
rs1033615446 Chr.9: 5908944-5908944 on Build GRCh38
rs760378999 Chr.9: 5909151-5909151 on Build GRCh38
rs560795480 Chr.9: 5908873-5908873 on Build GRCh38
rs934689382 Chr.9: 5909113-5909113 on Build GRCh38
rs547385238 Chr.9: 5908928-5908928 on Build GRCh38
rs866966055 Chr.9: 5908909-5908909 on Build GRCh38
rs1008939913 Chr.9: 5908884-5908884 on Build GRCh38
rs979277553 Chr.9: 5909107-5909107 on Build GRCh38
rs947437786 Chr.9: 5908853-5908853 on Build GRCh38
rs921389989 Chr.9: 5908828-5908828 on Build GRCh38
rs917841985 Chr.9: 5909118-5909118 on Build GRCh38
rs2233180 Chr.9: 5908772-5908772 on Build GRCh38
rs1013078637 Chr.9: 5909149-5909149 on Build GRCh38
rs931117410 Chr.9: 5909139-5909139 on Build GRCh38
rs144676130 Chr.9: 5909076-5909076 on Build GRCh38
rs528818889 Chr.9: 5909080-5909080 on Build GRCh38
rs898304379 Chr.9: 5908869-5908869 on Build GRCh38
rs965600317 Chr.9: 5909056-5909056 on Build GRCh38
rs577979836 Chr.9: 5908881-5908881 on Build GRCh38
rs896395827 Chr.9: 5908904-5908904 on Build GRCh38
rs772419510 Chr.9: 5908822-5908822 on Build GRCh38
rs912796369 Chr.9: 5908780-5908780 on Build GRCh38
rs1040793341 Chr.9: 5909115-5909115 on Build GRCh38
rs991176936 Chr.9: 5908787-5908787 on Build GRCh38
rs768933181 Chr.9: 5908967-5908967 on Build GRCh38
rs1003869687 Chr.9: 5909026-5909026 on Build GRCh38
rs981538039 Chr.9: 5909021-5909021 on Build GRCh38
rs1020321861 Chr.9: 5909043-5909043 on Build GRCh38
rs371936965 Chr.9: 5909097-5909097 on Build GRCh38
rs192396843 Chr.9: 5908779-5908779 on Build GRCh38
rs773606117 Chr.9: 5908966-5908966 on Build GRCh38
rs548855037 Chr.9: 5909163-5909163 on Build GRCh38
rs904949800 Chr.9: 5908949-5908949 on Build GRCh38
rs199693682 Chr.9: 5909162-5909162 on Build GRCh38
rs908495237 Chr.9: 5908831-5908831 on Build GRCh38
rs149543135 Chr.9: 5908786-5908786 on Build GRCh38
rs1044931392 Chr.9: 5909128-5909128 on Build GRCh38
rs981065977 Chr.9: 5908823-5908823 on Build GRCh38
rs184580758 Chr.9: 5909129-5909129 on Build GRCh38
rs930716366 Chr.9: 5908870-5908870 on Build GRCh38
rs975547879 Chr.9: 5908806-5908806 on Build GRCh38
rs1033170792 Chr.9: 5908783-5908783 on Build GRCh38
rs1026670926 Chr.9: 5908985-5908985 on Build GRCh38
rs187821687 Chr.9: 5909168-5909168 on Build GRCh38
rs376551516 Chr.9: 5909058-5909058 on Build GRCh38
rs763310379 Chr.9: 5908923-5908923 on Build GRCh38
rs1052808374 Chr.9: 5908891-5908891 on Build GRCh38
rs1019240383 Chr.9: 5909184-5909184 on Build GRCh38
rs901442602 Chr.9: 5909190-5909190 on Build GRCh38
rs999517166 Chr.9: 5909122-5909122 on Build GRCh38
rs991207923 Chr.9: 5909002-5909002 on Build GRCh38
rs768795850 Chr.9: 5908759-5908759 on Build GRCh38
rs939940361 Chr.9: 5908835-5908835 on Build GRCh38
rs531736398 Chr.9: 5908799-5908799 on Build GRCh38
rs770330206 Chr.9: 5908861-5908861 on Build GRCh38

View Full Product Details